Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16258745-16259295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISPD
Entrez Gene ID
729920
Gene Name
isoprenoid synthase domain containing
Gene Aliases
MDDGA7, MDDGC7, Nip, hCG_1745121
UniGene
Hs.636502
Gene Chromosome Location
Chr.7: 16087527-16422562 on Build GRCh38
Transcripts
NM_001101417.3 NM_001101426.3 XM_006715770.3 XM_011515499.2 XM_011515500.2 XM_011515508.2 XM_011515509.2 XM_017012575.1 XM_017012577.1 XM_017012578.1 NR_038946.1 NR_038947.1

Gene Symbol
ISPD-AS1
Entrez Gene ID
100506025
Gene Name
ISPD antisense RNA 1
Gene Aliases
-
UniGene
Hs.649029
Gene Chromosome Location
Chr.7: 16210486-16270604 on Build GRCh38
Transcripts
NM_001101417.3 NM_001101426.3 XM_006715770.3 XM_011515499.2 XM_011515500.2 XM_011515508.2 XM_011515509.2 XM_017012575.1 XM_017012577.1 XM_017012578.1 NR_038946.1 NR_038947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5997148 c.976C>T p.H326Y Chr.7: 16258970-16258970 on Build GRCh38
5500416 c.934-10T>C p.? Chr.7: 16259022-16259022 on Build GRCh38
7372506 c.947C>A p.T316K Chr.7: 16258999-16258999 on Build GRCh38
1449745 c.1013T>G p.F338C Chr.7: 16258933-16258933 on Build GRCh38
1088047 c.990C>A p.I330I Chr.7: 16258956-16258956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768164446 Chr.7: 16258923-16258923 on Build GRCh38
rs113227942 Chr.7: 16258875-16258875 on Build GRCh38
rs773791866 Chr.7: 16259015-16259015 on Build GRCh38
rs888126960 Chr.7: 16259103-16259103 on Build GRCh38
rs550533620 Chr.7: 16259131-16259131 on Build GRCh38
rs547690848 Chr.7: 16259175-16259175 on Build GRCh38
rs763179938 Chr.7: 16259049-16259049 on Build GRCh38
rs767455446 Chr.7: 16258931-16258931 on Build GRCh38
rs747466513 Chr.7: 16258887-16258887 on Build GRCh38
rs144982982 Chr.7: 16259192-16259194 on Build GRCh38
rs587780361 Chr.7: 16258967-16258967 on Build GRCh38
rs7799150 Chr.7: 16258813-16258813 on Build GRCh38
rs573577667 Chr.7: 16258962-16258962 on Build GRCh38
rs5882563 Chr.7: 16259141-16259142 on Build GRCh38
rs887511192 Chr.7: 16259201-16259201 on Build GRCh38
rs1021551358 Chr.7: 16259173-16259173 on Build GRCh38
rs768631434 Chr.7: 16259162-16259162 on Build GRCh38
rs565474383 Chr.7: 16259133-16259133 on Build GRCh38
rs935544667 Chr.7: 16259130-16259130 on Build GRCh38
rs371264127 Chr.7: 16259039-16259039 on Build GRCh38
rs1002831695 Chr.7: 16259069-16259069 on Build GRCh38
rs762349005 Chr.7: 16258915-16258915 on Build GRCh38
rs901037542 Chr.7: 16258968-16258968 on Build GRCh38
rs781782597 Chr.7: 16258965-16258965 on Build GRCh38
rs73684109 Chr.7: 16259222-16259222 on Build GRCh38
rs375739127 Chr.7: 16258878-16258878 on Build GRCh38
rs762323912 Chr.7: 16258873-16258874 on Build GRCh38
rs756731680 Chr.7: 16258966-16258966 on Build GRCh38
rs760530856 Chr.7: 16258946-16258946 on Build GRCh38
rs928489314 Chr.7: 16258833-16258833 on Build GRCh38
rs532701222 Chr.7: 16259150-16259150 on Build GRCh38
rs939907602 Chr.7: 16259122-16259122 on Build GRCh38
rs750914736 Chr.7: 16259060-16259060 on Build GRCh38
rs909531981 Chr.7: 16259265-16259266 on Build GRCh38
rs979393180 Chr.7: 16259096-16259096 on Build GRCh38
rs771988779 Chr.7: 16259021-16259021 on Build GRCh38
rs370052768 Chr.7: 16258974-16258974 on Build GRCh38
rs561291707 Chr.7: 16259112-16259112 on Build GRCh38
rs766147885 Chr.7: 16259041-16259041 on Build GRCh38
rs753009162 Chr.7: 16258950-16258950 on Build GRCh38
rs889158441 Chr.7: 16259144-16259144 on Build GRCh38
rs780620823 Chr.7: 16258998-16258998 on Build GRCh38
rs566456109 Chr.7: 16258839-16258839 on Build GRCh38
rs995906242 Chr.7: 16259009-16259009 on Build GRCh38
rs1050639989 Chr.7: 16259188-16259188 on Build GRCh38
rs114363936 Chr.7: 16258999-16258999 on Build GRCh38
rs149948893 Chr.7: 16259064-16259064 on Build GRCh38
rs370374480 Chr.7: 16259087-16259087 on Build GRCh38
rs886062166 Chr.7: 16258910-16258910 on Build GRCh38
rs775248815 Chr.7: 16258911-16258911 on Build GRCh38
rs939450573 Chr.7: 16258842-16258842 on Build GRCh38
rs376909665 Chr.7: 16258947-16258947 on Build GRCh38
rs369832637 Chr.7: 16258936-16258936 on Build GRCh38
rs1006011546 Chr.7: 16259211-16259211 on Build GRCh38
rs992975870 Chr.7: 16259224-16259224 on Build GRCh38
rs924822515 Chr.7: 16258816-16258816 on Build GRCh38
rs33970227 Chr.7: 16259142-16259143 on Build GRCh38
rs886044693 Chr.7: 16258955-16258955 on Build GRCh38
rs190443306 Chr.7: 16259126-16259126 on Build GRCh38
rs916555400 Chr.7: 16258882-16258882 on Build GRCh38
rs886043337 Chr.7: 16259017-16259017 on Build GRCh38
rs969768875 Chr.7: 16259244-16259245 on Build GRCh38
rs1031960933 Chr.7: 16259204-16259204 on Build GRCh38
rs183829084 Chr.7: 16259258-16259258 on Build GRCh38
rs370409636 Chr.7: 16259193-16259195 on Build GRCh38
rs374251724 Chr.7: 16259044-16259044 on Build GRCh38
rs779271557 Chr.7: 16259193-16259193 on Build GRCh38
rs761713653 Chr.7: 16259089-16259089 on Build GRCh38
rs1035491513 Chr.7: 16259076-16259076 on Build GRCh38
rs958797870 Chr.7: 16259078-16259078 on Build GRCh38
rs897499534 Chr.7: 16259237-16259237 on Build GRCh38
rs867449738 Chr.7: 16259254-16259254 on Build GRCh38
rs762898634 Chr.7: 16259265-16259269 on Build GRCh38
rs376875864 Chr.7: 16259006-16259006 on Build GRCh38
rs373422736 Chr.7: 16258956-16258956 on Build GRCh38
rs773174159 Chr.7: 16259026-16259026 on Build GRCh38
rs938401166 Chr.7: 16258855-16258855 on Build GRCh38
rs968130163 Chr.7: 16259093-16259093 on Build GRCh38
rs202126749 Chr.7: 16258961-16258961 on Build GRCh38
rs1005872310 Chr.7: 16259052-16259052 on Build GRCh38
rs139478143 Chr.7: 16258874-16258875 on Build GRCh38
rs571902097 Chr.7: 16258788-16258788 on Build GRCh38
rs1048070284 Chr.7: 16259102-16259102 on Build GRCh38
rs757453295 Chr.7: 16259055-16259055 on Build GRCh38
rs769331832 Chr.7: 16258905-16258905 on Build GRCh38
rs902010047 Chr.7: 16259003-16259003 on Build GRCh38
rs577463503 Chr.7: 16259018-16259018 on Build GRCh38
rs369141466 Chr.7: 16258954-16258954 on Build GRCh38
rs555932095 Chr.7: 16259011-16259011 on Build GRCh38
rs1295153 Chr.7: 16258809-16258809 on Build GRCh38
rs781644120 Chr.7: 16258903-16258903 on Build GRCh38
rs914860696 Chr.7: 16258800-16258800 on Build GRCh38
rs751686177 Chr.7: 16259054-16259054 on Build GRCh38
rs773884120 Chr.7: 16258929-16258929 on Build GRCh38
rs764470709 Chr.7: 16259050-16259050 on Build GRCh38
rs991316967 Chr.7: 16258868-16258868 on Build GRCh38
rs771571339 Chr.7: 16258898-16258898 on Build GRCh38

View Full Product Details