Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41929385-41929919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT6A
Entrez Gene ID
7994
Gene Name
lysine acetyltransferase 6A
Gene Aliases
MOZ, MRD32, MYST-3, MYST3, RUNXBP2, ZC2HC6A, ZNF220
UniGene
Hs.491577
Gene Chromosome Location
Chr.8: 41929479-42052026 on Build GRCh38
Transcripts
NM_006766.4 XM_011544656.2 XM_011544657.2 XM_011544658.2 XM_011544659.2 XM_017013863.1 XM_017013864.1

SNP Detail

SNP ID Chromosome Location
rs11551808 Chr.8: 41929638-41929638 on Build GRCh38
rs888333495 Chr.8: 41929684-41929684 on Build GRCh38
rs11551807 Chr.8: 41929562-41929562 on Build GRCh38
rs1025551593 Chr.8: 41929641-41929641 on Build GRCh38
rs542020052 Chr.8: 41929668-41929668 on Build GRCh38
rs979652035 Chr.8: 41929443-41929443 on Build GRCh38
rs949420434 Chr.8: 41929530-41929530 on Build GRCh38
rs968215304 Chr.8: 41929426-41929426 on Build GRCh38
rs1005483789 Chr.8: 41929698-41929698 on Build GRCh38
rs572420649 Chr.8: 41929445-41929445 on Build GRCh38
rs952508926 Chr.8: 41929436-41929436 on Build GRCh38
rs975139135 Chr.8: 41929502-41929503 on Build GRCh38
rs976741947 Chr.8: 41929735-41929735 on Build GRCh38
rs1054339529 Chr.8: 41929585-41929585 on Build GRCh38
rs563890357 Chr.8: 41929694-41929694 on Build GRCh38
rs1025608671 Chr.8: 41929420-41929420 on Build GRCh38
rs1002608519 Chr.8: 41929605-41929605 on Build GRCh38
rs577228426 Chr.8: 41929636-41929636 on Build GRCh38
rs978034506 Chr.8: 41929813-41929813 on Build GRCh38
rs534955890 Chr.8: 41929508-41929508 on Build GRCh38
rs900838194 Chr.8: 41929628-41929628 on Build GRCh38
rs186022719 Chr.8: 41929731-41929731 on Build GRCh38
rs988713407 Chr.8: 41929421-41929425 on Build GRCh38
rs2939983 Chr.8: 41929776-41929776 on Build GRCh38
rs905149149 Chr.8: 41929593-41929593 on Build GRCh38
rs561013203 Chr.8: 41929801-41929801 on Build GRCh38
rs147476663 Chr.8: 41929577-41929577 on Build GRCh38
rs143230746 Chr.8: 41929526-41929526 on Build GRCh38
rs1040851962 Chr.8: 41929617-41929617 on Build GRCh38
rs1021084476 Chr.8: 41929421-41929421 on Build GRCh38
rs145487556 Chr.8: 41929786-41929786 on Build GRCh38
rs941214464 Chr.8: 41929575-41929575 on Build GRCh38
rs982526561 Chr.8: 41929751-41929751 on Build GRCh38
rs1034198492 Chr.8: 41929451-41929451 on Build GRCh38
rs563806864 Chr.8: 41929614-41929614 on Build GRCh38
rs941186833 Chr.8: 41929785-41929785 on Build GRCh38
rs910829278 Chr.8: 41929447-41929447 on Build GRCh38
rs1038202314 Chr.8: 41929796-41929796 on Build GRCh38
rs923961882 Chr.8: 41929867-41929867 on Build GRCh38
rs191309256 Chr.8: 41929742-41929742 on Build GRCh38
rs924218646 Chr.8: 41929568-41929568 on Build GRCh38
rs529944897 Chr.8: 41929805-41929805 on Build GRCh38
rs139955916 Chr.8: 41929610-41929610 on Build GRCh38
rs879084445 Chr.8: 41929616-41929616 on Build GRCh38
rs1056793829 Chr.8: 41929806-41929806 on Build GRCh38
rs778250090 Chr.8: 41929426-41929428 on Build GRCh38
rs766215949 Chr.8: 41929670-41929670 on Build GRCh38
rs990921797 Chr.8: 41929495-41929495 on Build GRCh38
rs968189490 Chr.8: 41929653-41929653 on Build GRCh38
rs377285350 Chr.8: 41929532-41929532 on Build GRCh38
rs955226785 Chr.8: 41929750-41929750 on Build GRCh38
rs767495433 Chr.8: 41929529-41929529 on Build GRCh38
rs1017861485 Chr.8: 41929686-41929686 on Build GRCh38
rs959695978 Chr.8: 41929480-41929480 on Build GRCh38
rs867662965 Chr.8: 41929594-41929594 on Build GRCh38
rs960858735 Chr.8: 41929413-41929413 on Build GRCh38
rs908352582 Chr.8: 41929548-41929549 on Build GRCh38
rs964997512 Chr.8: 41929734-41929734 on Build GRCh38
rs889964773 Chr.8: 41929609-41929609 on Build GRCh38
rs559757991 Chr.8: 41929725-41929725 on Build GRCh38

View Full Product Details