Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22604790-22605332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCAR2
Entrez Gene ID
57805
Gene Name
cell cycle and apoptosis regulator 2
Gene Aliases
DBC-1, DBC1, KIAA1967, NET35, p30 DBC, p30DBC
UniGene
Hs.744848
Gene Chromosome Location
Chr.8: 22604632-22620471 on Build GRCh38
Transcripts
NM_021174.5 NR_033902.1 XM_011544603.1 XM_011544604.1 XM_017013717.1 XM_017014122.1

Gene Symbol
LOC107986876
Entrez Gene ID
107986876
Gene Name
proline-rich proteoglycan 2-like
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.8: 22602708-22605328 on Build GRCh38
Transcripts
NM_021174.5 NR_033902.1 XM_011544603.1 XM_011544604.1 XM_017013717.1 XM_017014122.1

SNP Detail

SNP ID Chromosome Location
rs922454147 Chr.8: 22605000-22605000 on Build GRCh38
rs985740475 Chr.8: 22605247-22605247 on Build GRCh38
rs1048427270 Chr.8: 22604814-22604814 on Build GRCh38
rs1013994973 Chr.8: 22605116-22605116 on Build GRCh38
rs961366625 Chr.8: 22605043-22605043 on Build GRCh38
rs200570116 Chr.8: 22605223-22605223 on Build GRCh38
rs764862215 Chr.8: 22605034-22605034 on Build GRCh38
rs950998647 Chr.8: 22605142-22605142 on Build GRCh38
rs772657108 Chr.8: 22604877-22604877 on Build GRCh38
rs1010879877 Chr.8: 22604813-22604813 on Build GRCh38
rs1042395560 Chr.8: 22604953-22604953 on Build GRCh38
rs536188377 Chr.8: 22604980-22604980 on Build GRCh38
rs201399089 Chr.8: 22605224-22605224 on Build GRCh38
rs754487578 Chr.8: 22604880-22604887 on Build GRCh38
rs564255980 Chr.8: 22605288-22605288 on Build GRCh38
rs1049921944 Chr.8: 22604907-22604907 on Build GRCh38
rs905579512 Chr.8: 22605305-22605305 on Build GRCh38
rs201335134 Chr.8: 22605286-22605286 on Build GRCh38
rs549779482 Chr.8: 22604959-22604959 on Build GRCh38
rs931265953 Chr.8: 22604813-22604814 on Build GRCh38
rs201600355 Chr.8: 22604820-22604820 on Build GRCh38
rs961631384 Chr.8: 22604882-22604882 on Build GRCh38
rs199833878 Chr.8: 22605265-22605265 on Build GRCh38
rs949710937 Chr.8: 22605271-22605271 on Build GRCh38
rs755838760 Chr.8: 22604855-22604856 on Build GRCh38
rs904456636 Chr.8: 22604869-22604869 on Build GRCh38
rs946772777 Chr.8: 22605105-22605105 on Build GRCh38
rs769666593 Chr.8: 22604850-22604850 on Build GRCh38
rs961710860 Chr.8: 22605180-22605180 on Build GRCh38
rs1013129686 Chr.8: 22604987-22604987 on Build GRCh38
rs11555543 Chr.8: 22604833-22604833 on Build GRCh38
rs926820886 Chr.8: 22604897-22604897 on Build GRCh38
rs1049584208 Chr.8: 22605032-22605032 on Build GRCh38
rs567939163 Chr.8: 22604965-22604965 on Build GRCh38
rs11777808 Chr.8: 22605242-22605242 on Build GRCh38
rs939310959 Chr.8: 22605139-22605139 on Build GRCh38
rs3735901 Chr.8: 22604861-22604861 on Build GRCh38
rs998224699 Chr.8: 22605241-22605241 on Build GRCh38
rs1045419062 Chr.8: 22605287-22605287 on Build GRCh38
rs79550139 Chr.8: 22605245-22605245 on Build GRCh38
rs753710853 Chr.8: 22604879-22604879 on Build GRCh38
rs116765617 Chr.8: 22605159-22605159 on Build GRCh38
rs907486276 Chr.8: 22604948-22604948 on Build GRCh38
rs543823029 Chr.8: 22604829-22604829 on Build GRCh38
rs199766031 Chr.8: 22605220-22605220 on Build GRCh38
rs774610379 Chr.8: 22604868-22604868 on Build GRCh38
rs952440139 Chr.8: 22605018-22605018 on Build GRCh38
rs1022765096 Chr.8: 22604993-22604993 on Build GRCh38
rs200166208 Chr.8: 22605058-22605058 on Build GRCh38
rs915437915 Chr.8: 22605094-22605094 on Build GRCh38
rs541736319 Chr.8: 22604875-22604875 on Build GRCh38
rs977027837 Chr.8: 22605045-22605045 on Build GRCh38
rs1003535158 Chr.8: 22604954-22604954 on Build GRCh38
rs200389746 Chr.8: 22605147-22605147 on Build GRCh38
rs796561329 Chr.8: 22605227-22605228 on Build GRCh38
rs902541657 Chr.8: 22604961-22604961 on Build GRCh38
rs377754041 Chr.8: 22604819-22604819 on Build GRCh38
rs201911737 Chr.8: 22605263-22605263 on Build GRCh38
rs976788554 Chr.8: 22605208-22605208 on Build GRCh38
rs952957594 Chr.8: 22604935-22604935 on Build GRCh38
rs891501022 Chr.8: 22605074-22605074 on Build GRCh38
rs200592497 Chr.8: 22605199-22605199 on Build GRCh38
rs560439467 Chr.8: 22604883-22604883 on Build GRCh38
rs531273213 Chr.8: 22604950-22604950 on Build GRCh38
rs200058989 Chr.8: 22604816-22604816 on Build GRCh38
rs539647252 Chr.8: 22605040-22605040 on Build GRCh38
rs201475220 Chr.8: 22605216-22605216 on Build GRCh38
rs201082198 Chr.8: 22604885-22604885 on Build GRCh38
rs771123507 Chr.8: 22604863-22604863 on Build GRCh38
rs1022465807 Chr.8: 22605248-22605248 on Build GRCh38
rs1044190867 Chr.8: 22605177-22605177 on Build GRCh38
rs1015796869 Chr.8: 22605226-22605226 on Build GRCh38
rs202161379 Chr.8: 22605135-22605135 on Build GRCh38
rs985247351 Chr.8: 22605243-22605243 on Build GRCh38
rs118007235 Chr.8: 22605066-22605066 on Build GRCh38
rs777791656 Chr.8: 22604859-22604859 on Build GRCh38
rs1025138044 Chr.8: 22604884-22604884 on Build GRCh38
rs915302322 Chr.8: 22604941-22604941 on Build GRCh38
rs946803800 Chr.8: 22604945-22604945 on Build GRCh38
rs201975249 Chr.8: 22604834-22604834 on Build GRCh38
rs142286316 Chr.8: 22605029-22605029 on Build GRCh38
rs199504245 Chr.8: 22605185-22605185 on Build GRCh38
rs952472116 Chr.8: 22604836-22604836 on Build GRCh38
rs552362439 Chr.8: 22604893-22604893 on Build GRCh38
rs759466257 Chr.8: 22604983-22604983 on Build GRCh38
rs555761332 Chr.8: 22605155-22605155 on Build GRCh38
rs1033863187 Chr.8: 22605141-22605141 on Build GRCh38
rs939463617 Chr.8: 22605294-22605294 on Build GRCh38
rs896903590 Chr.8: 22605231-22605231 on Build GRCh38
rs983722388 Chr.8: 22604857-22604857 on Build GRCh38
rs998775213 Chr.8: 22604963-22604963 on Build GRCh38
rs905612083 Chr.8: 22605130-22605130 on Build GRCh38
rs1042530473 Chr.8: 22605117-22605117 on Build GRCh38
rs201605352 Chr.8: 22604814-22604814 on Build GRCh38
rs751491917 Chr.8: 22604818-22604818 on Build GRCh38
rs1050143408 Chr.8: 22604865-22604872 on Build GRCh38
rs960179479 Chr.8: 22604831-22604831 on Build GRCh38
rs765960979 Chr.8: 22604879-22604880 on Build GRCh38
rs750245754 Chr.8: 22604849-22604850 on Build GRCh38
rs554034785 Chr.8: 22604986-22604986 on Build GRCh38
rs985604618 Chr.8: 22605080-22605080 on Build GRCh38
rs570567213 Chr.8: 22604924-22604924 on Build GRCh38
rs1013048562 Chr.8: 22604855-22604855 on Build GRCh38
rs895736693 Chr.8: 22604966-22604966 on Build GRCh38
rs923988972 Chr.8: 22605278-22605278 on Build GRCh38

View Full Product Details