Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92210238-92210747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IARS
Entrez Gene ID
3376
Gene Name
isoleucyl-tRNA synthetase
Gene Aliases
IARS1, ILERS, ILRS, IRS, PRO0785
UniGene
Hs.445403
Gene Chromosome Location
Chr.9: 92210207-92293756 on Build GRCh38
Transcripts
NM_002161.5 NM_013417.3 NR_073446.1

SNP Detail

SNP ID Chromosome Location
rs1027703138 Chr.9: 92210329-92210329 on Build GRCh38
rs928168192 Chr.9: 92210344-92210344 on Build GRCh38
rs1055960014 Chr.9: 92210564-92210564 on Build GRCh38
rs941377090 Chr.9: 92210369-92210369 on Build GRCh38
rs1014295193 Chr.9: 92210278-92210278 on Build GRCh38
rs943233372 Chr.9: 92210699-92210699 on Build GRCh38
rs1014535626 Chr.9: 92210595-92210595 on Build GRCh38
rs985922182 Chr.9: 92210398-92210398 on Build GRCh38
rs987790247 Chr.9: 92210431-92210431 on Build GRCh38
rs1055388525 Chr.9: 92210362-92210362 on Build GRCh38
rs190530483 Chr.9: 92210609-92210609 on Build GRCh38
rs187123938 Chr.9: 92210322-92210322 on Build GRCh38
rs965826483 Chr.9: 92210701-92210701 on Build GRCh38
rs765863316 Chr.9: 92210570-92210570 on Build GRCh38
rs1034728755 Chr.9: 92210548-92210548 on Build GRCh38
rs1020636280 Chr.9: 92210490-92210490 on Build GRCh38
rs965920965 Chr.9: 92210430-92210430 on Build GRCh38
rs941605800 Chr.9: 92210611-92210611 on Build GRCh38
rs1017507093 Chr.9: 92210462-92210462 on Build GRCh38
rs1803840 Chr.9: 92210579-92210579 on Build GRCh38
rs1058726 Chr.9: 92210684-92210684 on Build GRCh38
rs774462678 Chr.9: 92210484-92210485 on Build GRCh38
rs953453768 Chr.9: 92210683-92210683 on Build GRCh38
rs966468492 Chr.9: 92210319-92210319 on Build GRCh38
rs1058751 Chr.9: 92210634-92210634 on Build GRCh38
rs528107267 Chr.9: 92210374-92210374 on Build GRCh38
rs529435156 Chr.9: 92210602-92210602 on Build GRCh38
rs148380707 Chr.9: 92210400-92210400 on Build GRCh38
rs562294921 Chr.9: 92210536-92210536 on Build GRCh38
rs546352686 Chr.9: 92210427-92210427 on Build GRCh38
rs575809462 Chr.9: 92210272-92210272 on Build GRCh38
rs570400938 Chr.9: 92210696-92210696 on Build GRCh38
rs893710685 Chr.9: 92210475-92210475 on Build GRCh38
rs946374063 Chr.9: 92210463-92210463 on Build GRCh38
rs1015324042 Chr.9: 92210294-92210294 on Build GRCh38
rs113546461 Chr.9: 92210680-92210680 on Build GRCh38
rs959567350 Chr.9: 92210573-92210573 on Build GRCh38
rs546176144 Chr.9: 92210306-92210306 on Build GRCh38
rs913576285 Chr.9: 92210444-92210444 on Build GRCh38
rs953335021 Chr.9: 92210368-92210368 on Build GRCh38
rs536759425 Chr.9: 92210662-92210662 on Build GRCh38
rs994807468 Chr.9: 92210658-92210658 on Build GRCh38
rs937527137 Chr.9: 92210542-92210542 on Build GRCh38
rs547008536 Chr.9: 92210526-92210526 on Build GRCh38
rs890287646 Chr.9: 92210432-92210432 on Build GRCh38
rs896096139 Chr.9: 92210584-92210584 on Build GRCh38
rs938127438 Chr.9: 92210345-92210345 on Build GRCh38
rs1038823665 Chr.9: 92210718-92210718 on Build GRCh38
rs973373507 Chr.9: 92210627-92210627 on Build GRCh38

View Full Product Details