Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92240689-92241233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IARS
Entrez Gene ID
3376
Gene Name
isoleucyl-tRNA synthetase
Gene Aliases
IARS1, ILERS, ILRS, IRS, PRO0785
UniGene
Hs.445403
Gene Chromosome Location
Chr.9: 92210207-92293756 on Build GRCh38
Transcripts
NM_002161.5 NM_013417.3 NR_073446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
381222 c.3258C>G p.N1086K Chr.9: 92240881-92240881 on Build GRCh38
1210204 c.3183G>T p.K1061N Chr.9: 92240956-92240956 on Build GRCh38
6116640 c.3283G>A p.G1095S Chr.9: 92240856-92240856 on Build GRCh38
4814744 c.441A>T p.Q147H Chr.9: 92240857-92240857 on Build GRCh38
1111044 c.3272G>A p.G1091D Chr.9: 92240867-92240867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747137603 Chr.9: 92241069-92241069 on Build GRCh38
rs550830895 Chr.9: 92241019-92241019 on Build GRCh38
rs897297669 Chr.9: 92241183-92241183 on Build GRCh38
rs754479236 Chr.9: 92240881-92240881 on Build GRCh38
rs766016986 Chr.9: 92240971-92240971 on Build GRCh38
rs868219056 Chr.9: 92240854-92240854 on Build GRCh38
rs371709177 Chr.9: 92240872-92240872 on Build GRCh38
rs950953949 Chr.9: 92241193-92241193 on Build GRCh38
rs749618886 Chr.9: 92241013-92241013 on Build GRCh38
rs112284692 Chr.9: 92240970-92240970 on Build GRCh38
rs552432572 Chr.9: 92240778-92240778 on Build GRCh38
rs771046038 Chr.9: 92241164-92241164 on Build GRCh38
rs987511599 Chr.9: 92241208-92241208 on Build GRCh38
rs973632706 Chr.9: 92240770-92240770 on Build GRCh38
rs760506058 Chr.9: 92240833-92240833 on Build GRCh38
rs745509732 Chr.9: 92240985-92240988 on Build GRCh38
rs772046234 Chr.9: 92240947-92240947 on Build GRCh38
rs772851102 Chr.9: 92240808-92240808 on Build GRCh38
rs1013109094 Chr.9: 92240730-92240730 on Build GRCh38
rs748507246 Chr.9: 92240731-92240731 on Build GRCh38
rs1018334318 Chr.9: 92240873-92240873 on Build GRCh38
rs912007878 Chr.9: 92240864-92240864 on Build GRCh38
rs199645059 Chr.9: 92240961-92240961 on Build GRCh38
rs762976612 Chr.9: 92241037-92241037 on Build GRCh38
rs748680208 Chr.9: 92240893-92240893 on Build GRCh38
rs774168823 Chr.9: 92241175-92241175 on Build GRCh38
rs540398402 Chr.9: 92240751-92240751 on Build GRCh38
rs987846944 Chr.9: 92240805-92240805 on Build GRCh38
rs149779047 Chr.9: 92240918-92240918 on Build GRCh38
rs949775286 Chr.9: 92241089-92241089 on Build GRCh38
rs770269194 Chr.9: 92240902-92240902 on Build GRCh38
rs191211575 Chr.9: 92240754-92240754 on Build GRCh38
rs139191387 Chr.9: 92240903-92240903 on Build GRCh38
rs979068255 Chr.9: 92241005-92241005 on Build GRCh38
rs376331249 Chr.9: 92241148-92241148 on Build GRCh38
rs535322462 Chr.9: 92240801-92240801 on Build GRCh38
rs998551447 Chr.9: 92241039-92241039 on Build GRCh38
rs374939512 Chr.9: 92240888-92240888 on Build GRCh38
rs878999308 Chr.9: 92240830-92240830 on Build GRCh38
rs1055444484 Chr.9: 92241048-92241048 on Build GRCh38
rs765025476 Chr.9: 92240856-92240856 on Build GRCh38
rs763323746 Chr.9: 92240807-92240807 on Build GRCh38
rs764040984 Chr.9: 92240839-92240839 on Build GRCh38
rs568318028 Chr.9: 92240932-92240932 on Build GRCh38
rs1027011530 Chr.9: 92240781-92240781 on Build GRCh38
rs899342369 Chr.9: 92241135-92241137 on Build GRCh38
rs776570033 Chr.9: 92240949-92240949 on Build GRCh38
rs569245699 Chr.9: 92241161-92241161 on Build GRCh38
rs371678044 Chr.9: 92240969-92240969 on Build GRCh38
rs754452383 Chr.9: 92240978-92240978 on Build GRCh38
rs774527491 Chr.9: 92240819-92240819 on Build GRCh38
rs750270216 Chr.9: 92240967-92240967 on Build GRCh38
rs895577471 Chr.9: 92241050-92241050 on Build GRCh38
rs976560854 Chr.9: 92240877-92240877 on Build GRCh38
rs753748025 Chr.9: 92240847-92240847 on Build GRCh38
rs764990499 Chr.9: 92240962-92240962 on Build GRCh38
rs746481683 Chr.9: 92240879-92240879 on Build GRCh38
rs771326067 Chr.9: 92240929-92240929 on Build GRCh38
rs150130428 Chr.9: 92240818-92240818 on Build GRCh38
rs756975046 Chr.9: 92240851-92240851 on Build GRCh38
rs201460676 Chr.9: 92240776-92240776 on Build GRCh38
rs747861580 Chr.9: 92240725-92240725 on Build GRCh38
rs1043015907 Chr.9: 92240917-92240917 on Build GRCh38
rs759289776 Chr.9: 92241199-92241199 on Build GRCh38
rs960045804 Chr.9: 92240713-92240713 on Build GRCh38
rs925670441 Chr.9: 92241020-92241020 on Build GRCh38
rs751178239 Chr.9: 92240977-92240977 on Build GRCh38
rs535481901 Chr.9: 92240979-92240979 on Build GRCh38
rs946022681 Chr.9: 92241000-92241000 on Build GRCh38
rs778257886 Chr.9: 92240997-92240997 on Build GRCh38
rs759618440 Chr.9: 92240936-92240936 on Build GRCh38
rs766429670 Chr.9: 92240819-92240819 on Build GRCh38
rs770287616 Chr.9: 92240739-92240739 on Build GRCh38
rs187102071 Chr.9: 92240740-92240740 on Build GRCh38
rs113761432 Chr.9: 92240860-92240860 on Build GRCh38
rs937002019 Chr.9: 92241022-92241022 on Build GRCh38
rs777679320 Chr.9: 92240777-92240777 on Build GRCh38
rs146284911 Chr.9: 92240867-92240867 on Build GRCh38
rs1022677997 Chr.9: 92241128-92241128 on Build GRCh38
rs753251179 Chr.9: 92240993-92240993 on Build GRCh38
rs1013071497 Chr.9: 92241096-92241096 on Build GRCh38
rs1057183768 Chr.9: 92241048-92241050 on Build GRCh38
rs750075842 Chr.9: 92240862-92240862 on Build GRCh38
rs779411972 Chr.9: 92240874-92240874 on Build GRCh38

View Full Product Details