Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97930737-97931251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEMGN
Entrez Gene ID
55363
Gene Name
hemogen
Gene Aliases
CT155, EDAG, EDAG-1, NDR
UniGene
Hs.176626
Gene Chromosome Location
Chr.9: 97926791-97944915 on Build GRCh38
Transcripts
NM_018437.4 NM_197978.2 XM_005252086.1 XM_011518845.2 XM_011518846.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2793229 c.640C>T p.H214Y Chr.9: 97930755-97930755 on Build GRCh38
4498478 c.520C>T p.P174S Chr.9: 97930875-97930875 on Build GRCh38
7370971 c.394C>G p.H132D Chr.9: 97931001-97931001 on Build GRCh38
5912918 c.206G>A p.G69E Chr.9: 97931189-97931189 on Build GRCh38
5954531 c.547C>A p.L183I Chr.9: 97930848-97930848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044657825 Chr.9: 97930900-97930900 on Build GRCh38
rs576790299 Chr.9: 97930809-97930809 on Build GRCh38
rs768261992 Chr.9: 97930771-97930771 on Build GRCh38
rs376645183 Chr.9: 97930837-97930837 on Build GRCh38
rs777381979 Chr.9: 97931024-97931024 on Build GRCh38
rs941293540 Chr.9: 97930906-97930906 on Build GRCh38
rs761762464 Chr.9: 97930948-97930948 on Build GRCh38
rs750485705 Chr.9: 97931209-97931209 on Build GRCh38
rs146853281 Chr.9: 97931008-97931008 on Build GRCh38
rs776710745 Chr.9: 97931198-97931198 on Build GRCh38
rs145386159 Chr.9: 97931088-97931088 on Build GRCh38
rs143681169 Chr.9: 97930941-97930941 on Build GRCh38
rs748934699 Chr.9: 97931033-97931033 on Build GRCh38
rs201982775 Chr.9: 97931159-97931159 on Build GRCh38
rs766637508 Chr.9: 97931097-97931097 on Build GRCh38
rs953425759 Chr.9: 97930815-97930815 on Build GRCh38
rs554987978 Chr.9: 97930800-97930800 on Build GRCh38
rs759296272 Chr.9: 97930937-97930938 on Build GRCh38
rs765383909 Chr.9: 97930954-97930954 on Build GRCh38
rs191644480 Chr.9: 97930836-97930836 on Build GRCh38
rs764155775 Chr.9: 97930806-97930806 on Build GRCh38
rs745597622 Chr.9: 97931049-97931049 on Build GRCh38
rs867154235 Chr.9: 97930899-97930899 on Build GRCh38
rs1031519231 Chr.9: 97931185-97931186 on Build GRCh38
rs201689350 Chr.9: 97930799-97930799 on Build GRCh38
rs776067752 Chr.9: 97930784-97930784 on Build GRCh38
rs769374349 Chr.9: 97930896-97930896 on Build GRCh38
rs775245293 Chr.9: 97931068-97931068 on Build GRCh38
rs990530163 Chr.9: 97930824-97930824 on Build GRCh38
rs779943930 Chr.9: 97931177-97931177 on Build GRCh38
rs747541400 Chr.9: 97930889-97930889 on Build GRCh38
rs751910562 Chr.9: 97931007-97931007 on Build GRCh38
rs914595266 Chr.9: 97930830-97930830 on Build GRCh38
rs541174960 Chr.9: 97930924-97930924 on Build GRCh38
rs755598168 Chr.9: 97930879-97930879 on Build GRCh38
rs955198864 Chr.9: 97930782-97930782 on Build GRCh38
rs370935913 Chr.9: 97931135-97931135 on Build GRCh38
rs565474361 Chr.9: 97930909-97930909 on Build GRCh38
rs772918303 Chr.9: 97930795-97930795 on Build GRCh38
rs569650277 Chr.9: 97931110-97931110 on Build GRCh38
rs144965245 Chr.9: 97931105-97931105 on Build GRCh38
rs986563473 Chr.9: 97930829-97930829 on Build GRCh38
rs781508998 Chr.9: 97931010-97931010 on Build GRCh38
rs762091173 Chr.9: 97931093-97931093 on Build GRCh38
rs756709528 Chr.9: 97931140-97931140 on Build GRCh38
rs758737111 Chr.9: 97930985-97930985 on Build GRCh38
rs771154310 Chr.9: 97931207-97931207 on Build GRCh38
rs202156150 Chr.9: 97931173-97931173 on Build GRCh38
rs771744071 Chr.9: 97931172-97931172 on Build GRCh38
rs898151041 Chr.9: 97931092-97931092 on Build GRCh38
rs760717668 Chr.9: 97931070-97931070 on Build GRCh38
rs896247630 Chr.9: 97930986-97930986 on Build GRCh38
rs267602331 Chr.9: 97931040-97931040 on Build GRCh38
rs772654568 Chr.9: 97930897-97930897 on Build GRCh38
rs772691220 Chr.9: 97930907-97930907 on Build GRCh38
rs530693956 Chr.9: 97931132-97931132 on Build GRCh38
rs778937138 Chr.9: 97930814-97930814 on Build GRCh38
rs532692786 Chr.9: 97930921-97930921 on Build GRCh38
rs763084947 Chr.9: 97931206-97931206 on Build GRCh38
rs770629296 Chr.9: 97931039-97931039 on Build GRCh38
rs774200186 Chr.9: 97930935-97930935 on Build GRCh38
rs371151700 Chr.9: 97930796-97930796 on Build GRCh38
rs767265129 Chr.9: 97931178-97931178 on Build GRCh38
rs139808473 Chr.9: 97930804-97930804 on Build GRCh38
rs1030400684 Chr.9: 97931168-97931168 on Build GRCh38
rs918429608 Chr.9: 97930932-97930932 on Build GRCh38
rs759840748 Chr.9: 97931209-97931209 on Build GRCh38
rs990233097 Chr.9: 97931208-97931209 on Build GRCh38
rs142085732 Chr.9: 97931208-97931208 on Build GRCh38
rs370588490 Chr.9: 97931072-97931072 on Build GRCh38
rs751475385 Chr.9: 97930803-97930803 on Build GRCh38
rs111240053 Chr.9: 97930995-97930995 on Build GRCh38
rs144511790 Chr.9: 97931084-97931084 on Build GRCh38
rs373987605 Chr.9: 97931130-97931130 on Build GRCh38
rs370929191 Chr.9: 97931094-97931094 on Build GRCh38
rs147951470 Chr.9: 97931145-97931145 on Build GRCh38
rs948723546 Chr.9: 97930857-97930857 on Build GRCh38
rs761510736 Chr.9: 97930785-97930785 on Build GRCh38
rs750692896 Chr.9: 97930823-97930823 on Build GRCh38
rs765066126 Chr.9: 97931073-97931073 on Build GRCh38
rs767972315 Chr.9: 97931133-97931133 on Build GRCh38
rs867323476 Chr.9: 97931189-97931189 on Build GRCh38
rs201990544 Chr.9: 97931105-97931107 on Build GRCh38
rs559750808 Chr.9: 97930964-97930964 on Build GRCh38
rs956159427 Chr.9: 97930822-97930822 on Build GRCh38
rs765203808 Chr.9: 97931074-97931074 on Build GRCh38
rs200834552 Chr.9: 97931047-97931047 on Build GRCh38
rs762389703 Chr.9: 97930919-97930919 on Build GRCh38
rs139664531 Chr.9: 97931063-97931063 on Build GRCh38
rs777135825 Chr.9: 97931146-97931146 on Build GRCh38
rs748709763 Chr.9: 97931017-97931017 on Build GRCh38
rs755555750 Chr.9: 97931023-97931023 on Build GRCh38
rs956296629 Chr.9: 97931201-97931201 on Build GRCh38
rs745544667 Chr.9: 97931171-97931171 on Build GRCh38
rs753287664 Chr.9: 97931149-97931149 on Build GRCh38
rs751768399 Chr.9: 97931072-97931073 on Build GRCh38
rs778499186 Chr.9: 97931163-97931163 on Build GRCh38
rs759349655 Chr.9: 97931091-97931091 on Build GRCh38
rs747490832 Chr.9: 97930854-97930854 on Build GRCh38
rs375684699 Chr.9: 97930810-97930810 on Build GRCh38

View Full Product Details