Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83972761-83973272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPK
Entrez Gene ID
3190
Gene Name
heterogeneous nuclear ribonucleoprotein K
Gene Aliases
AUKS, CSBP, HNRPK, TUNP
UniGene
Hs.522257
Gene Chromosome Location
Chr.9: 83968083-83980782 on Build GRCh38
Transcripts
NM_001318186.1 NM_001318187.1 NM_001318188.1 NM_002140.4 NM_031262.3 NM_031263.3 XM_005251960.2 XM_005251963.3 XM_005251965.2 XM_011518616.1 XM_017014668.1 XM_017014669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1314982 c.534C>G p.I178M Chr.9: 83972955-83972955 on Build GRCh38
4758442 c.598G>A p.D200N Chr.9: 83972891-83972891 on Build GRCh38
6681326 c.395G>T p.R132M Chr.9: 83972887-83972887 on Build GRCh38
3734482 c.593A>G p.K198R Chr.9: 83972896-83972896 on Build GRCh38
5716615 c.580C>T p.L194F Chr.9: 83972909-83972909 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764391860 Chr.9: 83973112-83973112 on Build GRCh38
rs374707438 Chr.9: 83972811-83972811 on Build GRCh38
rs772129876 Chr.9: 83973072-83973072 on Build GRCh38
rs993126825 Chr.9: 83972986-83972986 on Build GRCh38
rs761608526 Chr.9: 83973111-83973111 on Build GRCh38
rs777309624 Chr.9: 83973235-83973235 on Build GRCh38
rs758182685 Chr.9: 83972852-83972852 on Build GRCh38
rs766276220 Chr.9: 83972856-83972856 on Build GRCh38
rs201881795 Chr.9: 83972961-83972961 on Build GRCh38
rs971490131 Chr.9: 83973177-83973177 on Build GRCh38
rs767433017 Chr.9: 83973000-83973000 on Build GRCh38
rs754725549 Chr.9: 83972978-83972979 on Build GRCh38
rs966935818 Chr.9: 83973137-83973137 on Build GRCh38
rs771203047 Chr.9: 83973171-83973171 on Build GRCh38
rs984702076 Chr.9: 83973089-83973089 on Build GRCh38
rs753485538 Chr.9: 83973240-83973240 on Build GRCh38
rs764397197 Chr.9: 83972848-83972848 on Build GRCh38
rs760823353 Chr.9: 83973087-83973090 on Build GRCh38
rs550190413 Chr.9: 83973172-83973172 on Build GRCh38
rs898831589 Chr.9: 83972981-83972981 on Build GRCh38
rs187780635 Chr.9: 83973033-83973033 on Build GRCh38
rs538271548 Chr.9: 83973133-83973133 on Build GRCh38
rs372452585 Chr.9: 83973150-83973150 on Build GRCh38
rs1028623088 Chr.9: 83973084-83973084 on Build GRCh38
rs747814393 Chr.9: 83972952-83972952 on Build GRCh38
rs765317598 Chr.9: 83972997-83972997 on Build GRCh38
rs75380026 Chr.9: 83972987-83972987 on Build GRCh38
rs779318160 Chr.9: 83972968-83972968 on Build GRCh38
rs948597831 Chr.9: 83972821-83972821 on Build GRCh38
rs965276661 Chr.9: 83973061-83973061 on Build GRCh38
rs200447609 Chr.9: 83972790-83972790 on Build GRCh38
rs748000179 Chr.9: 83972995-83972997 on Build GRCh38
rs927965570 Chr.9: 83973143-83973143 on Build GRCh38
rs899048905 Chr.9: 83973217-83973217 on Build GRCh38
rs143811423 Chr.9: 83972970-83972970 on Build GRCh38
rs754517582 Chr.9: 83972895-83972895 on Build GRCh38
rs907381095 Chr.9: 83973147-83973147 on Build GRCh38
rs749661846 Chr.9: 83972964-83972964 on Build GRCh38
rs578077069 Chr.9: 83972792-83972792 on Build GRCh38
rs140053931 Chr.9: 83973065-83973065 on Build GRCh38
rs77240262 Chr.9: 83972793-83972793 on Build GRCh38
rs915731422 Chr.9: 83973173-83973173 on Build GRCh38
rs769430768 Chr.9: 83972799-83972800 on Build GRCh38
rs766162140 Chr.9: 83972994-83972994 on Build GRCh38
rs767474493 Chr.9: 83973156-83973157 on Build GRCh38
rs750091037 Chr.9: 83973207-83973207 on Build GRCh38
rs752241232 Chr.9: 83973004-83973004 on Build GRCh38
rs772290529 Chr.9: 83972976-83972976 on Build GRCh38
rs750253391 Chr.9: 83972851-83972851 on Build GRCh38
rs936011133 Chr.9: 83973222-83973222 on Build GRCh38
rs1017754471 Chr.9: 83973045-83973045 on Build GRCh38
rs759413214 Chr.9: 83972999-83972999 on Build GRCh38
rs1010435128 Chr.9: 83973044-83973044 on Build GRCh38
rs769579932 Chr.9: 83973059-83973062 on Build GRCh38
rs948576487 Chr.9: 83973178-83973178 on Build GRCh38
rs745314545 Chr.9: 83972795-83972796 on Build GRCh38
rs3835289 Chr.9: 83972793-83972793 on Build GRCh38
rs759547988 Chr.9: 83972995-83973000 on Build GRCh38
rs3765560 Chr.9: 83972791-83972791 on Build GRCh38
rs559736471 Chr.9: 83972789-83972789 on Build GRCh38
rs773583886 Chr.9: 83972794-83972794 on Build GRCh38
rs111725016 Chr.9: 83972896-83972896 on Build GRCh38
rs746646470 Chr.9: 83972786-83972788 on Build GRCh38
rs751465909 Chr.9: 83972889-83972889 on Build GRCh38
rs138817029 Chr.9: 83973206-83973206 on Build GRCh38
rs772279373 Chr.9: 83972788-83972788 on Build GRCh38
rs773982891 Chr.9: 83972987-83972988 on Build GRCh38
rs989684215 Chr.9: 83973077-83973077 on Build GRCh38
rs775769520 Chr.9: 83972979-83972979 on Build GRCh38
rs927292110 Chr.9: 83973208-83973209 on Build GRCh38
rs757566555 Chr.9: 83973245-83973245 on Build GRCh38
rs760920425 Chr.9: 83972836-83972836 on Build GRCh38
rs749224444 Chr.9: 83972975-83972977 on Build GRCh38
rs907661630 Chr.9: 83972818-83972818 on Build GRCh38
rs978028038 Chr.9: 83973138-83973138 on Build GRCh38
rs775608588 Chr.9: 83973237-83973237 on Build GRCh38
rs755826067 Chr.9: 83972957-83972957 on Build GRCh38
rs771274321 Chr.9: 83972967-83972967 on Build GRCh38
rs563593756 Chr.9: 83972937-83972937 on Build GRCh38
rs142206310 Chr.9: 83972988-83972988 on Build GRCh38
rs780080785 Chr.9: 83972799-83972802 on Build GRCh38
rs371333102 Chr.9: 83973231-83973232 on Build GRCh38
rs80180335 Chr.9: 83972988-83972988 on Build GRCh38
rs546300415 Chr.9: 83973087-83973087 on Build GRCh38
rs747163849 Chr.9: 83972837-83972837 on Build GRCh38
rs755597232 Chr.9: 83973015-83973015 on Build GRCh38
rs548534808 Chr.9: 83973132-83973132 on Build GRCh38
rs570243748 Chr.9: 83973194-83973194 on Build GRCh38
rs1009082125 Chr.9: 83973020-83973020 on Build GRCh38
rs769042240 Chr.9: 83972985-83972985 on Build GRCh38
rs528129316 Chr.9: 83973068-83973068 on Build GRCh38
rs202203225 Chr.9: 83972997-83972997 on Build GRCh38
rs545288231 Chr.9: 83972795-83972795 on Build GRCh38
rs760834546 Chr.9: 83972982-83972982 on Build GRCh38

View Full Product Details