Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130076331-130076844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR107
Entrez Gene ID
57720
Gene Name
G protein-coupled receptor 107
Gene Aliases
GCDRP, LUSTR1, bA138E2.2
UniGene
Hs.512461
Gene Chromosome Location
Chr.9: 130053706-130140169 on Build GRCh38
Transcripts
NM_001136557.1 NM_001136558.1 NM_001287346.1 NM_020960.4 XM_017014953.1 XM_017014954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1105964 c.256-1G>T p.? Chr.9: 130076411-130076411 on Build GRCh38
5875675 c.274C>T p.R92C Chr.9: 130076430-130076430 on Build GRCh38
1105965 c.280A>C p.K94Q Chr.9: 130076436-130076436 on Build GRCh38
5875674 c.274C>T p.R92C Chr.9: 130076430-130076430 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs948014034 Chr.9: 130076788-130076788 on Build GRCh38
rs774589594 Chr.9: 130076381-130076381 on Build GRCh38
rs533833970 Chr.9: 130076602-130076602 on Build GRCh38
rs778491780 Chr.9: 130076463-130076463 on Build GRCh38
rs540650114 Chr.9: 130076767-130076767 on Build GRCh38
rs879236820 Chr.9: 130076553-130076553 on Build GRCh38
rs922313869 Chr.9: 130076626-130076626 on Build GRCh38
rs774520099 Chr.9: 130076508-130076508 on Build GRCh38
rs774932201 Chr.9: 130076469-130076470 on Build GRCh38
rs752392022 Chr.9: 130076471-130076471 on Build GRCh38
rs762362921 Chr.9: 130076511-130076511 on Build GRCh38
rs1031166977 Chr.9: 130076570-130076570 on Build GRCh38
rs374857228 Chr.9: 130076499-130076499 on Build GRCh38
rs535981986 Chr.9: 130076655-130076655 on Build GRCh38
rs193300635 Chr.9: 130076793-130076793 on Build GRCh38
rs975119610 Chr.9: 130076622-130076622 on Build GRCh38
rs11795316 Chr.9: 130076428-130076428 on Build GRCh38
rs762999461 Chr.9: 130076426-130076426 on Build GRCh38
rs982609649 Chr.9: 130076661-130076661 on Build GRCh38
rs1021942232 Chr.9: 130076612-130076612 on Build GRCh38
rs958349710 Chr.9: 130076557-130076557 on Build GRCh38
rs750773559 Chr.9: 130076440-130076440 on Build GRCh38
rs772667724 Chr.9: 130076809-130076809 on Build GRCh38
rs575437062 Chr.9: 130076674-130076674 on Build GRCh38
rs763165799 Chr.9: 130076505-130076505 on Build GRCh38
rs558312421 Chr.9: 130076627-130076627 on Build GRCh38
rs188689500 Chr.9: 130076578-130076578 on Build GRCh38
rs370049356 Chr.9: 130076482-130076482 on Build GRCh38
rs898544411 Chr.9: 130076756-130076756 on Build GRCh38
rs531579139 Chr.9: 130076817-130076817 on Build GRCh38
rs185428457 Chr.9: 130076404-130076404 on Build GRCh38
rs546259275 Chr.9: 130076800-130076800 on Build GRCh38
rs906633233 Chr.9: 130076568-130076568 on Build GRCh38
rs148958632 Chr.9: 130076782-130076782 on Build GRCh38
rs143387725 Chr.9: 130076671-130076671 on Build GRCh38
rs538008831 Chr.9: 130076731-130076731 on Build GRCh38
rs560681921 Chr.9: 130076407-130076407 on Build GRCh38
rs1023645497 Chr.9: 130076567-130076567 on Build GRCh38
rs776362152 Chr.9: 130076361-130076361 on Build GRCh38
rs757854714 Chr.9: 130076476-130076476 on Build GRCh38
rs746972038 Chr.9: 130076490-130076490 on Build GRCh38
rs759834537 Chr.9: 130076394-130076394 on Build GRCh38
rs562685735 Chr.9: 130076552-130076552 on Build GRCh38
rs547057650 Chr.9: 130076611-130076611 on Build GRCh38
rs762437671 Chr.9: 130076509-130076509 on Build GRCh38
rs889793745 Chr.9: 130076672-130076675 on Build GRCh38
rs141056882 Chr.9: 130076569-130076569 on Build GRCh38
rs975590385 Chr.9: 130076358-130076358 on Build GRCh38
rs775640281 Chr.9: 130076503-130076503 on Build GRCh38
rs201707508 Chr.9: 130076431-130076431 on Build GRCh38
rs941277782 Chr.9: 130076744-130076744 on Build GRCh38
rs933279792 Chr.9: 130076645-130076645 on Build GRCh38
rs981670659 Chr.9: 130076546-130076546 on Build GRCh38
rs578183661 Chr.9: 130076757-130076757 on Build GRCh38
rs571559063 Chr.9: 130076579-130076579 on Build GRCh38
rs770815690 Chr.9: 130076492-130076492 on Build GRCh38
rs549231946 Chr.9: 130076520-130076520 on Build GRCh38
rs1045046486 Chr.9: 130076792-130076792 on Build GRCh38
rs1038717407 Chr.9: 130076777-130076777 on Build GRCh38
rs756482071 Chr.9: 130076460-130076460 on Build GRCh38
rs926984152 Chr.9: 130076552-130076553 on Build GRCh38
rs562920767 Chr.9: 130076804-130076804 on Build GRCh38
rs149964893 Chr.9: 130076519-130076519 on Build GRCh38
rs146712475 Chr.9: 130076740-130076740 on Build GRCh38
rs762349769 Chr.9: 130076430-130076430 on Build GRCh38
rs964025426 Chr.9: 130076618-130076618 on Build GRCh38
rs955110844 Chr.9: 130076654-130076654 on Build GRCh38
rs781075846 Chr.9: 130076495-130076495 on Build GRCh38
rs752113003 Chr.9: 130076429-130076429 on Build GRCh38
rs923092140 Chr.9: 130076624-130076624 on Build GRCh38
rs768868418 Chr.9: 130076506-130076506 on Build GRCh38
rs1007264648 Chr.9: 130076691-130076691 on Build GRCh38
rs567391349 Chr.9: 130076642-130076642 on Build GRCh38
rs1011091766 Chr.9: 130076815-130076815 on Build GRCh38

View Full Product Details