Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 149482813-149483321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDS
Entrez Gene ID
3423
Gene Name
iduronate 2-sulfatase
Gene Aliases
MPS2, SIDS
UniGene
Hs.460960
Gene Chromosome Location
Chr.X: 149476990-149505354 on Build GRCh38
Transcripts
NM_000202.7 NM_001166550.3

SNP Detail

SNP ID Chromosome Location
rs782614727 Chr.X: 149483039-149483039 on Build GRCh38
rs782272275 Chr.X: 149482868-149482868 on Build GRCh38
rs781825818 Chr.X: 149483138-149483138 on Build GRCh38
rs782228221 Chr.X: 149482916-149482916 on Build GRCh38
rs782673868 Chr.X: 149482863-149482863 on Build GRCh38
rs199422231 Chr.X: 149482997-149482997 on Build GRCh38
rs199422229 Chr.X: 149483135-149483135 on Build GRCh38
rs782430567 Chr.X: 149482945-149482945 on Build GRCh38
rs782543505 Chr.X: 149483173-149483173 on Build GRCh38
rs782163574 Chr.X: 149483115-149483115 on Build GRCh38
rs34180481 Chr.X: 149483040-149483041 on Build GRCh38
rs782405504 Chr.X: 149483118-149483118 on Build GRCh38
rs1047741308 Chr.X: 149483071-149483071 on Build GRCh38
rs142248439 Chr.X: 149482909-149482909 on Build GRCh38
rs140633427 Chr.X: 149483109-149483109 on Build GRCh38
rs782190885 Chr.X: 149482922-149482922 on Build GRCh38
rs199982552 Chr.X: 149482979-149482979 on Build GRCh38
rs782443447 Chr.X: 149483050-149483050 on Build GRCh38
rs781951068 Chr.X: 149483007-149483007 on Build GRCh38
rs938593101 Chr.X: 149482910-149482910 on Build GRCh38
rs372205468 Chr.X: 149482900-149482900 on Build GRCh38
rs782657740 Chr.X: 149483177-149483177 on Build GRCh38
rs864622772 Chr.X: 149483006-149483006 on Build GRCh38
rs377093277 Chr.X: 149483269-149483269 on Build GRCh38
rs1803783 Chr.X: 149483140-149483140 on Build GRCh38
rs782724941 Chr.X: 149482839-149482839 on Build GRCh38
rs968443951 Chr.X: 149482892-149482892 on Build GRCh38
rs782793881 Chr.X: 149483030-149483030 on Build GRCh38
rs146819769 Chr.X: 149483029-149483029 on Build GRCh38
rs864622773 Chr.X: 149482966-149482966 on Build GRCh38
rs373373636 Chr.X: 149483253-149483253 on Build GRCh38
rs782347729 Chr.X: 149482921-149482921 on Build GRCh38
rs61736890 Chr.X: 149483130-149483130 on Build GRCh38
rs782203657 Chr.X: 149483106-149483106 on Build GRCh38
rs782107496 Chr.X: 149482961-149482961 on Build GRCh38
rs782066597 Chr.X: 149483142-149483142 on Build GRCh38
rs782590564 Chr.X: 149482924-149482924 on Build GRCh38
rs782433387 Chr.X: 149482851-149482851 on Build GRCh38
rs373820890 Chr.X: 149483129-149483129 on Build GRCh38
rs782690963 Chr.X: 149483052-149483052 on Build GRCh38
rs369603623 Chr.X: 149483047-149483047 on Build GRCh38
rs782340858 Chr.X: 149483027-149483027 on Build GRCh38
rs398123248 Chr.X: 149482891-149482891 on Build GRCh38
rs782022550 Chr.X: 149482955-149482955 on Build GRCh38
rs781831059 Chr.X: 149483206-149483206 on Build GRCh38
rs370074198 Chr.X: 149482903-149482903 on Build GRCh38
rs113993951 Chr.X: 149483005-149483005 on Build GRCh38
rs948618232 Chr.X: 149483002-149483002 on Build GRCh38
rs864622777 Chr.X: 149483219-149483219 on Build GRCh38
rs104894862 Chr.X: 149482935-149482935 on Build GRCh38
rs782665561 Chr.X: 149483248-149483248 on Build GRCh38
rs782375760 Chr.X: 149483267-149483267 on Build GRCh38
rs886044835 Chr.X: 149483134-149483134 on Build GRCh38
rs104894863 Chr.X: 149482933-149482933 on Build GRCh38
rs113993946 Chr.X: 149482996-149482996 on Build GRCh38
rs915730288 Chr.X: 149482990-149482990 on Build GRCh38
rs200950988 Chr.X: 149483232-149483232 on Build GRCh38
rs920711197 Chr.X: 149483046-149483046 on Build GRCh38
rs199422227 Chr.X: 149483072-149483072 on Build GRCh38
rs869025307 Chr.X: 149482936-149482936 on Build GRCh38
rs782711044 Chr.X: 149483163-149483163 on Build GRCh38
rs782475200 Chr.X: 149483215-149483215 on Build GRCh38
rs199965368 Chr.X: 149483233-149483233 on Build GRCh38
rs782371096 Chr.X: 149483065-149483065 on Build GRCh38
rs376811313 Chr.X: 149483036-149483036 on Build GRCh38
rs782189099 Chr.X: 149482941-149482941 on Build GRCh38
rs782739449 Chr.X: 149483137-149483137 on Build GRCh38
rs782063057 Chr.X: 149483019-149483019 on Build GRCh38
rs201905166 Chr.X: 149483172-149483172 on Build GRCh38
rs199422228 Chr.X: 149482894-149482894 on Build GRCh38
rs929376875 Chr.X: 149483066-149483066 on Build GRCh38
rs781916566 Chr.X: 149483114-149483114 on Build GRCh38
rs188486717 Chr.X: 149483231-149483231 on Build GRCh38
rs781803924 Chr.X: 149482847-149482847 on Build GRCh38
rs199422230 Chr.X: 149482974-149482974 on Build GRCh38

View Full Product Details