Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228457619-228458128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST3H2A
Entrez Gene ID
92815
Gene Name
histone cluster 3, H2a
Gene Aliases
-
UniGene
Hs.26331
Gene Chromosome Location
Chr.1: 228457364-228457859 on Build GRCh38
Transcripts
NM_033445.2 NM_175055.2

Gene Symbol
HIST3H2BB
Entrez Gene ID
128312
Gene Name
histone cluster 3, H2bb
Gene Aliases
H2Bb
UniGene
Hs.376691
Gene Chromosome Location
Chr.1: 228458107-228458558 on Build GRCh38
Transcripts
NM_033445.2 NM_175055.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4818057 c.122C>G p.S41W Chr.1: 228457696-228457696 on Build GRCh38
4629931 c.91G>A p.V31M Chr.1: 228457727-228457727 on Build GRCh38
5924826 c.79C>T p.P27S Chr.1: 228457739-228457739 on Build GRCh38
5568012 c.4T>A p.S2T Chr.1: 228457814-228457814 on Build GRCh38
344994 c.65C>T p.A22V Chr.1: 228457753-228457753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776688002 Chr.1: 228457839-228457839 on Build GRCh38
rs773586146 Chr.1: 228457868-228457868 on Build GRCh38
rs1021158394 Chr.1: 228457887-228457887 on Build GRCh38
rs144926526 Chr.1: 228457653-228457653 on Build GRCh38
rs377226278 Chr.1: 228457806-228457806 on Build GRCh38
rs780988555 Chr.1: 228457685-228457685 on Build GRCh38
rs752086533 Chr.1: 228457816-228457816 on Build GRCh38
rs774281987 Chr.1: 228457712-228457712 on Build GRCh38
rs747692221 Chr.1: 228457831-228457831 on Build GRCh38
rs1039109206 Chr.1: 228458073-228458073 on Build GRCh38
rs543171468 Chr.1: 228457877-228457877 on Build GRCh38
rs945894931 Chr.1: 228457898-228457898 on Build GRCh38
rs942855 Chr.1: 228457774-228457774 on Build GRCh38
rs778566807 Chr.1: 228457850-228457850 on Build GRCh38
rs540587079 Chr.1: 228457864-228457864 on Build GRCh38
rs143327597 Chr.1: 228457672-228457672 on Build GRCh38
rs745683569 Chr.1: 228457829-228457829 on Build GRCh38
rs552550547 Chr.1: 228457825-228457825 on Build GRCh38
rs762851936 Chr.1: 228457647-228457647 on Build GRCh38
rs201639497 Chr.1: 228457857-228457857 on Build GRCh38
rs746461177 Chr.1: 228457681-228457681 on Build GRCh38
rs150267399 Chr.1: 228458096-228458096 on Build GRCh38
rs926975441 Chr.1: 228458058-228458058 on Build GRCh38
rs373109004 Chr.1: 228457764-228457764 on Build GRCh38
rs765739478 Chr.1: 228457811-228457811 on Build GRCh38
rs201380350 Chr.1: 228457821-228457821 on Build GRCh38
rs775748103 Chr.1: 228457725-228457725 on Build GRCh38
rs767666170 Chr.1: 228457664-228457664 on Build GRCh38
rs191360468 Chr.1: 228457848-228457848 on Build GRCh38
rs527986027 Chr.1: 228458014-228458014 on Build GRCh38
rs1019782787 Chr.1: 228457994-228457994 on Build GRCh38
rs200984136 Chr.1: 228457680-228457680 on Build GRCh38
rs959052107 Chr.1: 228457934-228457934 on Build GRCh38
rs904439325 Chr.1: 228457904-228457904 on Build GRCh38
rs1008061056 Chr.1: 228457805-228457805 on Build GRCh38
rs747009400 Chr.1: 228457856-228457856 on Build GRCh38
rs891659542 Chr.1: 228457895-228457895 on Build GRCh38
rs749653313 Chr.1: 228457869-228457869 on Build GRCh38
rs537698349 Chr.1: 228458100-228458100 on Build GRCh38
rs970581869 Chr.1: 228458002-228458002 on Build GRCh38
rs980180247 Chr.1: 228457876-228457876 on Build GRCh38
rs780054377 Chr.1: 228457837-228457837 on Build GRCh38
rs1008341408 Chr.1: 228457993-228457993 on Build GRCh38
rs554836275 Chr.1: 228457883-228457883 on Build GRCh38
rs772115546 Chr.1: 228457791-228457791 on Build GRCh38
rs1054926252 Chr.1: 228457700-228457700 on Build GRCh38
rs529372989 Chr.1: 228457873-228457873 on Build GRCh38
rs1042929308 Chr.1: 228457901-228457901 on Build GRCh38
rs552047150 Chr.1: 228457906-228457906 on Build GRCh38
rs780866027 Chr.1: 228457797-228457799 on Build GRCh38
rs777977167 Chr.1: 228457650-228457650 on Build GRCh38
rs949269040 Chr.1: 228457861-228457861 on Build GRCh38
rs200158501 Chr.1: 228457831-228457831 on Build GRCh38
rs755659156 Chr.1: 228457820-228457820 on Build GRCh38
rs909193174 Chr.1: 228458040-228458040 on Build GRCh38
rs750151585 Chr.1: 228457853-228457853 on Build GRCh38
rs533207092 Chr.1: 228457666-228457666 on Build GRCh38
rs756852465 Chr.1: 228457849-228457849 on Build GRCh38
rs1020898228 Chr.1: 228457817-228457817 on Build GRCh38
rs368509155 Chr.1: 228458102-228458102 on Build GRCh38
rs377337442 Chr.1: 228457859-228457859 on Build GRCh38
rs770166768 Chr.1: 228457842-228457842 on Build GRCh38
rs760839318 Chr.1: 228457797-228457797 on Build GRCh38
rs772838473 Chr.1: 228457703-228457703 on Build GRCh38
rs151028786 Chr.1: 228457918-228457918 on Build GRCh38
rs759613154 Chr.1: 228457659-228457659 on Build GRCh38
rs777481655 Chr.1: 228457768-228457768 on Build GRCh38
rs199668562 Chr.1: 228457843-228457843 on Build GRCh38
rs564265762 Chr.1: 228457854-228457854 on Build GRCh38
rs763696925 Chr.1: 228457847-228457847 on Build GRCh38
rs961511824 Chr.1: 228458003-228458003 on Build GRCh38
rs749143766 Chr.1: 228457706-228457706 on Build GRCh38
rs998126525 Chr.1: 228457884-228457884 on Build GRCh38
rs764404411 Chr.1: 228457675-228457675 on Build GRCh38
rs920186932 Chr.1: 228457871-228457871 on Build GRCh38
rs953086593 Chr.1: 228458048-228458048 on Build GRCh38
rs538508693 Chr.1: 228457767-228457767 on Build GRCh38
rs1056874196 Chr.1: 228457890-228457890 on Build GRCh38
rs931185792 Chr.1: 228458024-228458024 on Build GRCh38
rs141105928 Chr.1: 228458092-228458092 on Build GRCh38
rs200407249 Chr.1: 228457683-228457683 on Build GRCh38
rs73101115 Chr.1: 228457824-228457824 on Build GRCh38
rs145889493 Chr.1: 228457686-228457686 on Build GRCh38
rs754510222 Chr.1: 228457762-228457762 on Build GRCh38
rs751264565 Chr.1: 228457830-228457831 on Build GRCh38
rs763508088 Chr.1: 228457815-228457815 on Build GRCh38
rs765827645 Chr.1: 228457752-228457752 on Build GRCh38
rs774687122 Chr.1: 228458083-228458083 on Build GRCh38
rs767929927 Chr.1: 228458087-228458087 on Build GRCh38
rs1028774367 Chr.1: 228457966-228457966 on Build GRCh38
rs770078991 Chr.1: 228457865-228457865 on Build GRCh38
rs560981026 Chr.1: 228458013-228458013 on Build GRCh38
rs887787516 Chr.1: 228457874-228457874 on Build GRCh38
rs369668136 Chr.1: 228457737-228457737 on Build GRCh38
rs372748163 Chr.1: 228457866-228457866 on Build GRCh38
rs749299926 Chr.1: 228457766-228457766 on Build GRCh38
rs569025660 Chr.1: 228458099-228458099 on Build GRCh38
rs749709354 Chr.1: 228457850-228457851 on Build GRCh38
rs775382770 Chr.1: 228457795-228457795 on Build GRCh38
rs186477951 Chr.1: 228457844-228457844 on Build GRCh38
rs546151991 Chr.1: 228458081-228458081 on Build GRCh38
rs779852531 Chr.1: 228457855-228457855 on Build GRCh38
rs919618126 Chr.1: 228457671-228457671 on Build GRCh38
rs773431307 Chr.1: 228457848-228457848 on Build GRCh38
rs143657399 Chr.1: 228457870-228457870 on Build GRCh38
rs929289437 Chr.1: 228457872-228457872 on Build GRCh38
rs1001050817 Chr.1: 228458095-228458095 on Build GRCh38
rs764399724 Chr.1: 228457736-228457736 on Build GRCh38
rs375171103 Chr.1: 228457728-228457728 on Build GRCh38
rs902731382 Chr.1: 228457894-228457894 on Build GRCh38
rs770768289 Chr.1: 228457779-228457779 on Build GRCh38
rs972916661 Chr.1: 228458008-228458008 on Build GRCh38
rs564487637 Chr.1: 228457657-228457657 on Build GRCh38
rs535187855 Chr.1: 228457840-228457840 on Build GRCh38
rs768849083 Chr.1: 228457838-228457838 on Build GRCh38
rs571816531 Chr.1: 228458078-228458078 on Build GRCh38
rs773173464 Chr.1: 228457643-228457643 on Build GRCh38
rs762265732 Chr.1: 228457808-228457808 on Build GRCh38
rs56690462 Chr.1: 228458098-228458098 on Build GRCh38
rs902701729 Chr.1: 228458094-228458094 on Build GRCh38
rs781374851 Chr.1: 228457863-228457863 on Build GRCh38
rs746837643 Chr.1: 228457903-228457903 on Build GRCh38
rs567081351 Chr.1: 228457897-228457898 on Build GRCh38
rs747719816 Chr.1: 228457765-228457765 on Build GRCh38
rs547437583 Chr.1: 228458029-228458029 on Build GRCh38
rs778726446 Chr.1: 228457827-228457827 on Build GRCh38
rs774265600 Chr.1: 228457784-228457784 on Build GRCh38
rs373301076 Chr.1: 228457676-228457676 on Build GRCh38
rs770784546 Chr.1: 228457710-228457710 on Build GRCh38
rs750946591 Chr.1: 228457812-228457812 on Build GRCh38
rs889873146 Chr.1: 228457979-228457979 on Build GRCh38
rs745844717 Chr.1: 228457789-228457789 on Build GRCh38
rs760095350 Chr.1: 228457846-228457846 on Build GRCh38
rs754491870 Chr.1: 228457719-228457720 on Build GRCh38
rs757711200 Chr.1: 228457743-228457743 on Build GRCh38
rs751039844 Chr.1: 228457754-228457754 on Build GRCh38
rs901963872 Chr.1: 228458074-228458074 on Build GRCh38
rs979784812 Chr.1: 228458051-228458051 on Build GRCh38

View Full Product Details