Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 234606164-234606672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF2BP2
Entrez Gene ID
359948
Gene Name
interferon regulatory factor 2 binding protein 2
Gene Aliases
-
UniGene
Hs.350268
Gene Chromosome Location
Chr.1: 234604269-234609525 on Build GRCh38
Transcripts
NM_001077397.1 NM_182972.2

SNP Detail

SNP ID Chromosome Location
rs147260869 Chr.1: 234606613-234606613 on Build GRCh38
rs74147715 Chr.1: 234606267-234606267 on Build GRCh38
rs944352250 Chr.1: 234606289-234606289 on Build GRCh38
rs544271845 Chr.1: 234606508-234606508 on Build GRCh38
rs990891273 Chr.1: 234606440-234606440 on Build GRCh38
rs991264323 Chr.1: 234606299-234606299 on Build GRCh38
rs867708666 Chr.1: 234606554-234606554 on Build GRCh38
rs556030284 Chr.1: 234606568-234606568 on Build GRCh38
rs765325932 Chr.1: 234606622-234606622 on Build GRCh38
rs924175146 Chr.1: 234606459-234606460 on Build GRCh38
rs1053837666 Chr.1: 234606429-234606429 on Build GRCh38
rs1042375039 Chr.1: 234606465-234606465 on Build GRCh38
rs879126016 Chr.1: 234606459-234606459 on Build GRCh38
rs1012750947 Chr.1: 234606524-234606524 on Build GRCh38
rs958809194 Chr.1: 234606510-234606510 on Build GRCh38
rs545827270 Chr.1: 234606616-234606616 on Build GRCh38
rs145600370 Chr.1: 234606455-234606455 on Build GRCh38
rs374807730 Chr.1: 234606216-234606216 on Build GRCh38
rs893884362 Chr.1: 234606430-234606430 on Build GRCh38
rs968125557 Chr.1: 234606617-234606617 on Build GRCh38
rs902796358 Chr.1: 234606281-234606282 on Build GRCh38
rs1003217104 Chr.1: 234606469-234606469 on Build GRCh38
rs1035451270 Chr.1: 234606390-234606390 on Build GRCh38
rs574650661 Chr.1: 234606549-234606549 on Build GRCh38
rs910803392 Chr.1: 234606595-234606595 on Build GRCh38
rs187394782 Chr.1: 234606610-234606610 on Build GRCh38
rs558516780 Chr.1: 234606467-234606467 on Build GRCh38
rs1034301545 Chr.1: 234606597-234606597 on Build GRCh38
rs971767676 Chr.1: 234606547-234606547 on Build GRCh38
rs963077614 Chr.1: 234606342-234606342 on Build GRCh38
rs995894337 Chr.1: 234606359-234606359 on Build GRCh38
rs959458276 Chr.1: 234606426-234606426 on Build GRCh38
rs144730175 Chr.1: 234606471-234606472 on Build GRCh38
rs978230075 Chr.1: 234606619-234606619 on Build GRCh38
rs1004711808 Chr.1: 234606377-234606377 on Build GRCh38
rs946898986 Chr.1: 234606250-234606253 on Build GRCh38
rs766562916 Chr.1: 234606427-234606427 on Build GRCh38
rs971728132 Chr.1: 234606382-234606382 on Build GRCh38
rs753888510 Chr.1: 234606560-234606560 on Build GRCh38
rs77112131 Chr.1: 234606472-234606472 on Build GRCh38
rs931671458 Chr.1: 234606471-234606472 on Build GRCh38
rs183346952 Chr.1: 234606238-234606238 on Build GRCh38
rs374095934 Chr.1: 234606532-234606533 on Build GRCh38
rs80340810 Chr.1: 234606202-234606202 on Build GRCh38
rs887560917 Chr.1: 234606523-234606524 on Build GRCh38
rs767861878 Chr.1: 234606320-234606320 on Build GRCh38
rs1042448847 Chr.1: 234606632-234606632 on Build GRCh38
rs1003218428 Chr.1: 234606562-234606563 on Build GRCh38
rs568626741 Chr.1: 234606301-234606301 on Build GRCh38
rs576809158 Chr.1: 234606483-234606483 on Build GRCh38
rs368649071 Chr.1: 234606458-234606459 on Build GRCh38
rs946867656 Chr.1: 234606454-234606454 on Build GRCh38
rs535670707 Chr.1: 234606338-234606338 on Build GRCh38
rs1035597903 Chr.1: 234606501-234606501 on Build GRCh38
rs140081086 Chr.1: 234606408-234606408 on Build GRCh38
rs879054160 Chr.1: 234606625-234606625 on Build GRCh38
rs1051996511 Chr.1: 234606384-234606384 on Build GRCh38
rs1009575328 Chr.1: 234606459-234606459 on Build GRCh38
rs541899132 Chr.1: 234606562-234606562 on Build GRCh38
rs562478061 Chr.1: 234606525-234606525 on Build GRCh38
rs554016432 Chr.1: 234606375-234606375 on Build GRCh38
rs760943518 Chr.1: 234606374-234606374 on Build GRCh38
rs933570287 Chr.1: 234606383-234606383 on Build GRCh38
rs1012295713 Chr.1: 234606611-234606611 on Build GRCh38
rs375831683 Chr.1: 234606472-234606472 on Build GRCh38
rs915354342 Chr.1: 234606441-234606441 on Build GRCh38
rs527656019 Chr.1: 234606214-234606214 on Build GRCh38
rs773449732 Chr.1: 234606370-234606370 on Build GRCh38
rs887593780 Chr.1: 234606377-234606381 on Build GRCh38
rs570815038 Chr.1: 234606260-234606260 on Build GRCh38
rs574522312 Chr.1: 234606215-234606215 on Build GRCh38
rs967710361 Chr.1: 234606226-234606226 on Build GRCh38
rs918895178 Chr.1: 234606379-234606379 on Build GRCh38
rs755382836 Chr.1: 234606491-234606491 on Build GRCh38
rs573572453 Chr.1: 234606550-234606550 on Build GRCh38
rs918935251 Chr.1: 234606553-234606553 on Build GRCh38
rs550036445 Chr.1: 234606281-234606281 on Build GRCh38
rs963131228 Chr.1: 234606536-234606536 on Build GRCh38
rs542064683 Chr.1: 234606582-234606582 on Build GRCh38
rs900223545 Chr.1: 234606324-234606324 on Build GRCh38
rs140819648 Chr.1: 234606638-234606638 on Build GRCh38

View Full Product Details