Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 20383020-20383563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTATIP2
Entrez Gene ID
10553
Gene Name
HIV-1 Tat interactive protein 2
Gene Aliases
CC3, SDR44U1, TIP30
UniGene
Hs.90753
Gene Chromosome Location
Chr.11: 20363685-20383783 on Build GRCh38
Transcripts
NM_001098520.1 NM_001098521.1 NM_001098522.1 NM_006410.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1585648 c.792C>A p.I264I Chr.11: 20383166-20383166 on Build GRCh38
141331 c.655G>A p.D219N Chr.11: 20383131-20383131 on Build GRCh38
925897 c.690C>A p.I230I Chr.11: 20383166-20383166 on Build GRCh38
4691436 c.718G>A p.V240M Chr.11: 20383092-20383092 on Build GRCh38
4691437 c.616G>A p.V206M Chr.11: 20383092-20383092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187804828 Chr.11: 20383157-20383157 on Build GRCh38
rs995844406 Chr.11: 20383245-20383245 on Build GRCh38
rs764655503 Chr.11: 20383094-20383094 on Build GRCh38
rs373986235 Chr.11: 20383097-20383097 on Build GRCh38
rs1028929036 Chr.11: 20383482-20383482 on Build GRCh38
rs775348386 Chr.11: 20383196-20383196 on Build GRCh38
rs1032275418 Chr.11: 20383399-20383399 on Build GRCh38
rs747027347 Chr.11: 20383145-20383145 on Build GRCh38
rs774484044 Chr.11: 20383065-20383066 on Build GRCh38
rs530341063 Chr.11: 20383452-20383452 on Build GRCh38
rs3824886 Chr.11: 20383067-20383067 on Build GRCh38
rs757620420 Chr.11: 20383101-20383101 on Build GRCh38
rs1056454313 Chr.11: 20383258-20383258 on Build GRCh38
rs375554086 Chr.11: 20383515-20383515 on Build GRCh38
rs749219713 Chr.11: 20383160-20383160 on Build GRCh38
rs3781679 Chr.11: 20383211-20383211 on Build GRCh38
rs765797571 Chr.11: 20383237-20383237 on Build GRCh38
rs183794581 Chr.11: 20383477-20383477 on Build GRCh38
rs372324685 Chr.11: 20383341-20383341 on Build GRCh38
rs575746142 Chr.11: 20383112-20383112 on Build GRCh38
rs1030826980 Chr.11: 20383458-20383458 on Build GRCh38
rs751841952 Chr.11: 20383254-20383254 on Build GRCh38
rs999325258 Chr.11: 20383076-20383076 on Build GRCh38
rs142077207 Chr.11: 20383079-20383079 on Build GRCh38
rs752194360 Chr.11: 20383091-20383091 on Build GRCh38
rs35285540 Chr.11: 20383056-20383057 on Build GRCh38
rs990407224 Chr.11: 20383451-20383451 on Build GRCh38
rs768795404 Chr.11: 20383152-20383152 on Build GRCh38
rs773030202 Chr.11: 20383066-20383066 on Build GRCh38
rs558548051 Chr.11: 20383535-20383535 on Build GRCh38
rs779185313 Chr.11: 20383109-20383109 on Build GRCh38
rs370774686 Chr.11: 20383203-20383203 on Build GRCh38
rs772109156 Chr.11: 20383042-20383042 on Build GRCh38
rs14222 Chr.11: 20383161-20383161 on Build GRCh38
rs769665018 Chr.11: 20383058-20383058 on Build GRCh38
rs755756794 Chr.11: 20383296-20383296 on Build GRCh38
rs774019635 Chr.11: 20383077-20383077 on Build GRCh38
rs747115307 Chr.11: 20383051-20383051 on Build GRCh38
rs760248012 Chr.11: 20383199-20383199 on Build GRCh38
rs780272275 Chr.11: 20383141-20383141 on Build GRCh38
rs903951508 Chr.11: 20383389-20383389 on Build GRCh38
rs997866474 Chr.11: 20383472-20383472 on Build GRCh38
rs901747505 Chr.11: 20383466-20383466 on Build GRCh38
rs202065100 Chr.11: 20383184-20383184 on Build GRCh38
rs780069819 Chr.11: 20383050-20383050 on Build GRCh38
rs1017551648 Chr.11: 20383322-20383322 on Build GRCh38
rs750729301 Chr.11: 20383239-20383239 on Build GRCh38
rs934931376 Chr.11: 20383120-20383121 on Build GRCh38
rs200003351 Chr.11: 20383169-20383169 on Build GRCh38
rs948095544 Chr.11: 20383295-20383295 on Build GRCh38
rs990376038 Chr.11: 20383121-20383121 on Build GRCh38
rs965975054 Chr.11: 20383084-20383084 on Build GRCh38
rs762187614 Chr.11: 20383222-20383222 on Build GRCh38
rs777165692 Chr.11: 20383219-20383219 on Build GRCh38
rs548044158 Chr.11: 20383391-20383391 on Build GRCh38
rs144100738 Chr.11: 20383092-20383092 on Build GRCh38
rs777439517 Chr.11: 20383392-20383392 on Build GRCh38
rs766586797 Chr.11: 20383244-20383244 on Build GRCh38
rs773966588 Chr.11: 20383183-20383183 on Build GRCh38
rs79296772 Chr.11: 20383072-20383072 on Build GRCh38
rs956203587 Chr.11: 20383427-20383427 on Build GRCh38
rs560199282 Chr.11: 20383137-20383137 on Build GRCh38
rs548875479 Chr.11: 20383257-20383257 on Build GRCh38
rs895159200 Chr.11: 20383269-20383269 on Build GRCh38
rs568532494 Chr.11: 20383316-20383316 on Build GRCh38
rs191247504 Chr.11: 20383394-20383394 on Build GRCh38
rs770368315 Chr.11: 20383491-20383491 on Build GRCh38

View Full Product Details