Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65712266-65712773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT5
Entrez Gene ID
10524
Gene Name
lysine acetyltransferase 5
Gene Aliases
ESA1, HTATIP, HTATIP1, PLIP, TIP, TIP60, ZC2HC5, cPLA2
UniGene
Hs.397010
Gene Chromosome Location
Chr.11: 65711996-65719606 on Build GRCh38
Transcripts
NM_001206833.1 NM_006388.3 NM_182709.2 NM_182710.2 XM_006718421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7307922 c.56T>G p.V19G Chr.11: 65712323-65712323 on Build GRCh38
2166333 c.153G>C p.R51R Chr.11: 65712420-65712420 on Build GRCh38
6856436 c.72delC p.P26fs*2 Chr.11: 65712339-65712339 on Build GRCh38
340925 c.181C>G p.L61V Chr.11: 65712768-65712768 on Build GRCh38
5980125 c.9G>C p.E3D Chr.11: 65712276-65712276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770362135 Chr.11: 65712547-65712547 on Build GRCh38
rs149844051 Chr.11: 65712346-65712346 on Build GRCh38
rs779419111 Chr.11: 65712339-65712339 on Build GRCh38
rs757195078 Chr.11: 65712728-65712728 on Build GRCh38
rs756389401 Chr.11: 65712521-65712521 on Build GRCh38
rs754073163 Chr.11: 65712591-65712591 on Build GRCh38
rs764558397 Chr.11: 65712503-65712503 on Build GRCh38
rs138317008 Chr.11: 65712414-65712414 on Build GRCh38
rs747032550 Chr.11: 65712432-65712432 on Build GRCh38
rs777603470 Chr.11: 65712546-65712546 on Build GRCh38
rs780525439 Chr.11: 65712309-65712309 on Build GRCh38
rs751344307 Chr.11: 65712606-65712607 on Build GRCh38
rs4645912 Chr.11: 65712600-65712600 on Build GRCh38
rs34074374 Chr.11: 65712520-65712521 on Build GRCh38
rs771945201 Chr.11: 65712537-65712537 on Build GRCh38
rs1030596555 Chr.11: 65712605-65712605 on Build GRCh38
rs755217033 Chr.11: 65712721-65712721 on Build GRCh38
rs765520391 Chr.11: 65712408-65712408 on Build GRCh38
rs759132850 Chr.11: 65712486-65712486 on Build GRCh38
rs561937939 Chr.11: 65712561-65712561 on Build GRCh38
rs780401552 Chr.11: 65712587-65712587 on Build GRCh38
rs183474864 Chr.11: 65712698-65712698 on Build GRCh38
rs758947166 Chr.11: 65712502-65712502 on Build GRCh38
rs569013298 Chr.11: 65712615-65712615 on Build GRCh38
rs755341837 Chr.11: 65712727-65712727 on Build GRCh38
rs776195524 Chr.11: 65712556-65712556 on Build GRCh38
rs758895785 Chr.11: 65712411-65712411 on Build GRCh38
rs760441688 Chr.11: 65712388-65712388 on Build GRCh38
rs2236682 Chr.11: 65712619-65712619 on Build GRCh38
rs371801645 Chr.11: 65712286-65712286 on Build GRCh38
rs774458759 Chr.11: 65712369-65712369 on Build GRCh38
rs760125975 Chr.11: 65712478-65712478 on Build GRCh38
rs780916526 Chr.11: 65712543-65712543 on Build GRCh38
rs1043659303 Chr.11: 65712686-65712686 on Build GRCh38
rs533327527 Chr.11: 65712642-65712642 on Build GRCh38
rs749972219 Chr.11: 65712732-65712735 on Build GRCh38
rs757310473 Chr.11: 65712510-65712510 on Build GRCh38
rs969631091 Chr.11: 65712354-65712354 on Build GRCh38
rs767147481 Chr.11: 65712373-65712373 on Build GRCh38
rs779478369 Chr.11: 65712743-65712743 on Build GRCh38
rs527338502 Chr.11: 65712464-65712464 on Build GRCh38
rs755661982 Chr.11: 65712297-65712297 on Build GRCh38
rs1046521421 Chr.11: 65712680-65712680 on Build GRCh38
rs145735863 Chr.11: 65712396-65712396 on Build GRCh38
rs189328094 Chr.11: 65712745-65712745 on Build GRCh38
rs754128090 Chr.11: 65712647-65712647 on Build GRCh38
rs985763142 Chr.11: 65712658-65712658 on Build GRCh38
rs757220104 Chr.11: 65712304-65712304 on Build GRCh38
rs762522593 Chr.11: 65712626-65712626 on Build GRCh38
rs922921813 Chr.11: 65712382-65712382 on Build GRCh38
rs762630512 Chr.11: 65712359-65712359 on Build GRCh38
rs769241061 Chr.11: 65712622-65712622 on Build GRCh38
rs763326321 Chr.11: 65712569-65712569 on Build GRCh38
rs772145862 Chr.11: 65712342-65712342 on Build GRCh38
rs768913091 Chr.11: 65712628-65712628 on Build GRCh38
rs746060161 Chr.11: 65712462-65712462 on Build GRCh38
rs954377170 Chr.11: 65712657-65712657 on Build GRCh38
rs762319786 Chr.11: 65712578-65712578 on Build GRCh38
rs750103112 Chr.11: 65712293-65712293 on Build GRCh38
rs780955784 Chr.11: 65712730-65712731 on Build GRCh38
rs746979227 Chr.11: 65712601-65712601 on Build GRCh38
rs960651989 Chr.11: 65712298-65712298 on Build GRCh38
rs781263702 Chr.11: 65712511-65712511 on Build GRCh38
rs923202144 Chr.11: 65712351-65712351 on Build GRCh38
rs372327414 Chr.11: 65712635-65712635 on Build GRCh38
rs754148106 Chr.11: 65712321-65712321 on Build GRCh38
rs984618442 Chr.11: 65712357-65712357 on Build GRCh38
rs11824283 Chr.11: 65712595-65712595 on Build GRCh38
rs775392306 Chr.11: 65712356-65712356 on Build GRCh38
rs762292206 Chr.11: 65712634-65712634 on Build GRCh38
rs374033347 Chr.11: 65712742-65712742 on Build GRCh38
rs759253641 Chr.11: 65712560-65712560 on Build GRCh38
rs755330780 Chr.11: 65712403-65712403 on Build GRCh38
rs376915049 Chr.11: 65712744-65712744 on Build GRCh38
rs528686377 Chr.11: 65712474-65712474 on Build GRCh38
rs767152335 Chr.11: 65712646-65712646 on Build GRCh38
rs776155216 Chr.11: 65712485-65712485 on Build GRCh38
rs563798134 Chr.11: 65712739-65712740 on Build GRCh38
rs761775144 Chr.11: 65712371-65712371 on Build GRCh38
rs949170678 Chr.11: 65712675-65712675 on Build GRCh38
rs547897773 Chr.11: 65712682-65712682 on Build GRCh38
rs747919706 Chr.11: 65712526-65712526 on Build GRCh38
rs748616102 Chr.11: 65712340-65712340 on Build GRCh38
rs193248159 Chr.11: 65712749-65712749 on Build GRCh38
rs992030146 Chr.11: 65712316-65712316 on Build GRCh38
rs965877080 Chr.11: 65712593-65712593 on Build GRCh38
rs1012831415 Chr.11: 65712589-65712589 on Build GRCh38
rs781288835 Chr.11: 65712459-65712459 on Build GRCh38
rs767550971 Chr.11: 65712581-65712581 on Build GRCh38
rs765651272 Chr.11: 65712487-65712487 on Build GRCh38
rs755559349 Chr.11: 65712328-65712328 on Build GRCh38
rs548714821 Chr.11: 65712519-65712519 on Build GRCh38
rs756979869 Chr.11: 65712718-65712719 on Build GRCh38
rs778917918 Chr.11: 65712524-65712524 on Build GRCh38
rs750579504 Chr.11: 65712641-65712641 on Build GRCh38
rs986458897 Chr.11: 65712618-65712618 on Build GRCh38
rs770900061 Chr.11: 65712614-65712614 on Build GRCh38
rs772131194 Chr.11: 65712467-65712467 on Build GRCh38
rs766500643 Chr.11: 65712644-65712644 on Build GRCh38
rs765570662 Chr.11: 65712747-65712747 on Build GRCh38
rs913639558 Chr.11: 65712669-65712669 on Build GRCh38
rs775886362 Chr.11: 65712566-65712566 on Build GRCh38
rs750077392 Chr.11: 65712661-65712661 on Build GRCh38
rs940433483 Chr.11: 65712365-65712365 on Build GRCh38
rs752010869 Chr.11: 65712574-65712574 on Build GRCh38
rs757544883 Chr.11: 65712456-65712456 on Build GRCh38
rs775152111 Chr.11: 65712623-65712623 on Build GRCh38
rs140468859 Chr.11: 65712384-65712384 on Build GRCh38
rs763939081 Chr.11: 65712457-65712458 on Build GRCh38
rs781216718 Chr.11: 65712736-65712736 on Build GRCh38
rs752052309 Chr.11: 65712504-65712504 on Build GRCh38
rs765873081 Chr.11: 65712479-65712479 on Build GRCh38
rs924798141 Chr.11: 65712697-65712697 on Build GRCh38
rs202097241 Chr.11: 65712352-65712352 on Build GRCh38
rs774762553 Chr.11: 65712629-65712629 on Build GRCh38
rs767092355 Chr.11: 65712288-65712288 on Build GRCh38
rs777774982 Chr.11: 65712344-65712344 on Build GRCh38
rs756444430 Chr.11: 65712585-65712585 on Build GRCh38
rs146788368 Chr.11: 65712362-65712362 on Build GRCh38
rs373204283 Chr.11: 65712701-65712701 on Build GRCh38
rs1048625075 Chr.11: 65712720-65712720 on Build GRCh38
rs1013735578 Chr.11: 65712394-65712394 on Build GRCh38
rs1015616110 Chr.11: 65712579-65712579 on Build GRCh38
rs753058652 Chr.11: 65712497-65712497 on Build GRCh38
rs370183972 Chr.11: 65712725-65712725 on Build GRCh38
rs748152989 Chr.11: 65712466-65712466 on Build GRCh38
rs376679947 Chr.11: 65712711-65712711 on Build GRCh38

View Full Product Details