Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26894101-26894603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH3
Entrez Gene ID
63906
Gene Name
G-patch domain containing 3
Gene Aliases
GPATC3
UniGene
Hs.10903
Gene Chromosome Location
Chr.1: 26890488-26900471 on Build GRCh38
Transcripts
NM_022078.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7186364 c.877-1G>A p.? Chr.1: 26894411-26894411 on Build GRCh38
6126228 c.949A>T p.T317S Chr.1: 26894338-26894338 on Build GRCh38
383705 c.887G>A p.R296Q Chr.1: 26894400-26894400 on Build GRCh38
7161547 c.1021G>C p.D341H Chr.1: 26894266-26894266 on Build GRCh38
3804829 c.1021G>A p.D341N Chr.1: 26894266-26894266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749621439 Chr.1: 26894387-26894387 on Build GRCh38
rs1056976490 Chr.1: 26894336-26894336 on Build GRCh38
rs531495529 Chr.1: 26894165-26894165 on Build GRCh38
rs527302467 Chr.1: 26894306-26894306 on Build GRCh38
rs191910281 Chr.1: 26894427-26894427 on Build GRCh38
rs762761245 Chr.1: 26894425-26894425 on Build GRCh38
rs201286459 Chr.1: 26894456-26894456 on Build GRCh38
rs766893371 Chr.1: 26894444-26894445 on Build GRCh38
rs765597899 Chr.1: 26894204-26894204 on Build GRCh38
rs560661349 Chr.1: 26894544-26894544 on Build GRCh38
rs763385280 Chr.1: 26894420-26894421 on Build GRCh38
rs766590048 Chr.1: 26894303-26894303 on Build GRCh38
rs756009262 Chr.1: 26894261-26894261 on Build GRCh38
rs751224352 Chr.1: 26894360-26894360 on Build GRCh38
rs752167533 Chr.1: 26894439-26894439 on Build GRCh38
rs1018218096 Chr.1: 26894574-26894574 on Build GRCh38
rs763231664 Chr.1: 26894219-26894219 on Build GRCh38
rs561329283 Chr.1: 26894254-26894254 on Build GRCh38
rs1022359939 Chr.1: 26894468-26894468 on Build GRCh38
rs574697760 Chr.1: 26894176-26894176 on Build GRCh38
rs541771367 Chr.1: 26894166-26894166 on Build GRCh38
rs766564805 Chr.1: 26894241-26894241 on Build GRCh38
rs200646741 Chr.1: 26894373-26894373 on Build GRCh38
rs754611487 Chr.1: 26894436-26894436 on Build GRCh38
rs140800689 Chr.1: 26894353-26894353 on Build GRCh38
rs111667908 Chr.1: 26894262-26894262 on Build GRCh38
rs374388304 Chr.1: 26894428-26894428 on Build GRCh38
rs777164010 Chr.1: 26894199-26894199 on Build GRCh38
rs910266159 Chr.1: 26894576-26894576 on Build GRCh38
rs779718998 Chr.1: 26894354-26894354 on Build GRCh38
rs769130467 Chr.1: 26894187-26894187 on Build GRCh38
rs750210121 Chr.1: 26894349-26894349 on Build GRCh38
rs773579609 Chr.1: 26894394-26894395 on Build GRCh38
rs532482675 Chr.1: 26894564-26894564 on Build GRCh38
rs773240690 Chr.1: 26894217-26894217 on Build GRCh38
rs753090911 Chr.1: 26894492-26894492 on Build GRCh38
rs570455192 Chr.1: 26894372-26894372 on Build GRCh38
rs750361496 Chr.1: 26894408-26894408 on Build GRCh38
rs376433715 Chr.1: 26894401-26894401 on Build GRCh38
rs924114779 Chr.1: 26894421-26894421 on Build GRCh38
rs939286764 Chr.1: 26894357-26894357 on Build GRCh38
rs558186201 Chr.1: 26894541-26894541 on Build GRCh38
rs867564663 Chr.1: 26894321-26894321 on Build GRCh38
rs201916728 Chr.1: 26894407-26894407 on Build GRCh38
rs751632076 Chr.1: 26894243-26894243 on Build GRCh38
rs1034849149 Chr.1: 26894508-26894508 on Build GRCh38
rs778860374 Chr.1: 26894255-26894255 on Build GRCh38
rs377557354 Chr.1: 26894400-26894400 on Build GRCh38
rs777063753 Chr.1: 26894286-26894286 on Build GRCh38
rs374007151 Chr.1: 26894268-26894268 on Build GRCh38
rs578042270 Chr.1: 26894542-26894542 on Build GRCh38
rs1057050501 Chr.1: 26894473-26894473 on Build GRCh38
rs183238480 Chr.1: 26894448-26894448 on Build GRCh38
rs771262216 Chr.1: 26894455-26894455 on Build GRCh38
rs773609638 Chr.1: 26894293-26894293 on Build GRCh38
rs201761023 Chr.1: 26894343-26894343 on Build GRCh38
rs748658914 Chr.1: 26894290-26894290 on Build GRCh38
rs370948033 Chr.1: 26894201-26894201 on Build GRCh38
rs888527229 Chr.1: 26894558-26894558 on Build GRCh38
rs774578347 Chr.1: 26894397-26894397 on Build GRCh38
rs755608903 Chr.1: 26894440-26894440 on Build GRCh38
rs746498194 Chr.1: 26894282-26894282 on Build GRCh38
rs754416767 Chr.1: 26894364-26894364 on Build GRCh38
rs980562120 Chr.1: 26894487-26894487 on Build GRCh38
rs78128878 Chr.1: 26894134-26894134 on Build GRCh38
rs143131895 Chr.1: 26894382-26894382 on Build GRCh38
rs776885384 Chr.1: 26894355-26894359 on Build GRCh38
rs1006809009 Chr.1: 26894379-26894379 on Build GRCh38
rs938565397 Chr.1: 26894461-26894461 on Build GRCh38
rs746296896 Chr.1: 26894458-26894458 on Build GRCh38
rs780869113 Chr.1: 26894438-26894438 on Build GRCh38
rs960857442 Chr.1: 26894511-26894511 on Build GRCh38
rs768669739 Chr.1: 26894332-26894343 on Build GRCh38
rs753766754 Chr.1: 26894342-26894342 on Build GRCh38
rs772402002 Chr.1: 26894460-26894460 on Build GRCh38
rs755146947 Chr.1: 26894246-26894246 on Build GRCh38
rs771218807 Chr.1: 26894390-26894390 on Build GRCh38
rs901437011 Chr.1: 26894150-26894150 on Build GRCh38
rs763272894 Chr.1: 26894299-26894299 on Build GRCh38
rs534730523 Chr.1: 26894509-26894509 on Build GRCh38
rs947012872 Chr.1: 26894221-26894221 on Build GRCh38
rs770132662 Chr.1: 26894386-26894386 on Build GRCh38
rs561128096 Chr.1: 26894174-26894174 on Build GRCh38
rs770351598 Chr.1: 26894292-26894292 on Build GRCh38
rs762009789 Chr.1: 26894355-26894356 on Build GRCh38
rs747138414 Chr.1: 26894214-26894214 on Build GRCh38
rs914345954 Chr.1: 26894196-26894196 on Build GRCh38
rs765146455 Chr.1: 26894344-26894344 on Build GRCh38
rs867077371 Chr.1: 26894278-26894278 on Build GRCh38
rs753851443 Chr.1: 26894253-26894253 on Build GRCh38
rs768175745 Chr.1: 26894283-26894283 on Build GRCh38

View Full Product Details