Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39761611-39762150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKZF3
Entrez Gene ID
22806
Gene Name
IKAROS family zinc finger 3
Gene Aliases
AIO, AIOLOS, ZNFN1A3
UniGene
Hs.371680
Gene Chromosome Location
Chr.17: 39757715-39864188 on Build GRCh38
Transcripts
NM_001257408.1 NM_001257409.1 NM_001257410.1 NM_001257411.1 NM_001257412.1 NM_001257413.1 NM_001257414.1 NM_001284514.1 NM_001284515.1 NM_001284516.1 NM_012481.4 NM_183228.2 NM_183229.2 NM_183230.2 NM_183231.2 NM_183232.2

SNP Detail

SNP ID Chromosome Location
rs999537966 Chr.17: 39761929-39761929 on Build GRCh38
rs992612026 Chr.17: 39761729-39761729 on Build GRCh38
rs193212176 Chr.17: 39761727-39761727 on Build GRCh38
rs777031119 Chr.17: 39762084-39762084 on Build GRCh38
rs942441410 Chr.17: 39762023-39762023 on Build GRCh38
rs574986209 Chr.17: 39762110-39762110 on Build GRCh38
rs913056579 Chr.17: 39761790-39761790 on Build GRCh38
rs867328045 Chr.17: 39761997-39761997 on Build GRCh38
rs953741956 Chr.17: 39762054-39762054 on Build GRCh38
rs968092696 Chr.17: 39761696-39761697 on Build GRCh38
rs569844747 Chr.17: 39761798-39761798 on Build GRCh38
rs1031139206 Chr.17: 39761976-39761976 on Build GRCh38
rs1047732565 Chr.17: 39762068-39762068 on Build GRCh38
rs532122882 Chr.17: 39761659-39761659 on Build GRCh38
rs930235784 Chr.17: 39762065-39762065 on Build GRCh38
rs751560772 Chr.17: 39761734-39761734 on Build GRCh38
rs185614791 Chr.17: 39761818-39761818 on Build GRCh38
rs547451665 Chr.17: 39761711-39761711 on Build GRCh38
rs552999950 Chr.17: 39761781-39761781 on Build GRCh38
rs988689989 Chr.17: 39761958-39761958 on Build GRCh38
rs940432354 Chr.17: 39762095-39762096 on Build GRCh38
rs867799049 Chr.17: 39761754-39761754 on Build GRCh38
rs111680233 Chr.17: 39762033-39762033 on Build GRCh38
rs777304592 Chr.17: 39761858-39761858 on Build GRCh38
rs1052971564 Chr.17: 39761782-39761782 on Build GRCh38
rs896612283 Chr.17: 39761861-39761861 on Build GRCh38
rs933760673 Chr.17: 39761791-39761791 on Build GRCh38
rs979454150 Chr.17: 39761896-39761896 on Build GRCh38
rs1022235661 Chr.17: 39761874-39761874 on Build GRCh38
rs922244746 Chr.17: 39762034-39762034 on Build GRCh38
rs554422711 Chr.17: 39761773-39761773 on Build GRCh38
rs772618704 Chr.17: 39761895-39761895 on Build GRCh38
rs754749509 Chr.17: 39761747-39761747 on Build GRCh38
rs962869939 Chr.17: 39761654-39761654 on Build GRCh38
rs536571570 Chr.17: 39761738-39761738 on Build GRCh38
rs933791407 Chr.17: 39761995-39761995 on Build GRCh38
rs924089393 Chr.17: 39761698-39761698 on Build GRCh38
rs1025755545 Chr.17: 39761703-39761703 on Build GRCh38
rs147729257 Chr.17: 39761866-39761866 on Build GRCh38
rs1051254253 Chr.17: 39761797-39761797 on Build GRCh38
rs576955757 Chr.17: 39761862-39761862 on Build GRCh38
rs978254673 Chr.17: 39761697-39761700 on Build GRCh38
rs775917490 Chr.17: 39761915-39761915 on Build GRCh38
rs768960999 Chr.17: 39762064-39762064 on Build GRCh38
rs775707551 Chr.17: 39761746-39761746 on Build GRCh38
rs969765149 Chr.17: 39761735-39761735 on Build GRCh38
rs1017113522 Chr.17: 39761643-39761643 on Build GRCh38
rs913074619 Chr.17: 39761984-39761984 on Build GRCh38
rs781161272 Chr.17: 39761777-39761777 on Build GRCh38
rs898780338 Chr.17: 39761851-39761851 on Build GRCh38
rs9909432 Chr.17: 39761853-39761853 on Build GRCh38
rs1003210335 Chr.17: 39761849-39761849 on Build GRCh38
rs957316059 Chr.17: 39761925-39761925 on Build GRCh38
rs572960391 Chr.17: 39761814-39761814 on Build GRCh38
rs991315283 Chr.17: 39761633-39761633 on Build GRCh38
rs552969769 Chr.17: 39762100-39762100 on Build GRCh38
rs1038035120 Chr.17: 39762047-39762047 on Build GRCh38
rs986587306 Chr.17: 39761996-39761996 on Build GRCh38
rs189341742 Chr.17: 39761848-39761848 on Build GRCh38
rs200058262 Chr.17: 39762113-39762113 on Build GRCh38
rs761201699 Chr.17: 39761943-39761943 on Build GRCh38

View Full Product Details