Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39763256-39763755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IKZF3
Entrez Gene ID
22806
Gene Name
IKAROS family zinc finger 3
Gene Aliases
AIO, AIOLOS, ZNFN1A3
UniGene
Hs.371680
Gene Chromosome Location
Chr.17: 39757715-39864188 on Build GRCh38
Transcripts
NM_001257408.1 NM_001257409.1 NM_001257410.1 NM_001257411.1 NM_001257412.1 NM_001257413.1 NM_001257414.1 NM_001284514.1 NM_001284515.1 NM_001284516.1 NM_012481.4 NM_183228.2 NM_183229.2 NM_183230.2 NM_183231.2 NM_183232.2

SNP Detail

SNP ID Chromosome Location
rs188500605 Chr.17: 39763546-39763546 on Build GRCh38
rs758548134 Chr.17: 39763341-39763341 on Build GRCh38
rs549090033 Chr.17: 39763342-39763342 on Build GRCh38
rs943405632 Chr.17: 39763629-39763629 on Build GRCh38
rs879408164 Chr.17: 39763496-39763496 on Build GRCh38
rs1050440836 Chr.17: 39763412-39763412 on Build GRCh38
rs528560806 Chr.17: 39763474-39763474 on Build GRCh38
rs551201402 Chr.17: 39763554-39763554 on Build GRCh38
rs149262106 Chr.17: 39763637-39763637 on Build GRCh38
rs1013027747 Chr.17: 39763663-39763663 on Build GRCh38
rs993768529 Chr.17: 39763505-39763505 on Build GRCh38
rs559447294 Chr.17: 39763364-39763365 on Build GRCh38
rs1057514901 Chr.17: 39763588-39763588 on Build GRCh38
rs559534124 Chr.17: 39763405-39763405 on Build GRCh38
rs899792745 Chr.17: 39763655-39763655 on Build GRCh38
rs113162064 Chr.17: 39763691-39763691 on Build GRCh38
rs978591750 Chr.17: 39763286-39763286 on Build GRCh38
rs188710489 Chr.17: 39763666-39763666 on Build GRCh38
rs530734610 Chr.17: 39763657-39763657 on Build GRCh38
rs923298687 Chr.17: 39763337-39763337 on Build GRCh38
rs1051827096 Chr.17: 39763559-39763559 on Build GRCh38
rs890470815 Chr.17: 39763622-39763622 on Build GRCh38
rs982511703 Chr.17: 39763308-39763308 on Build GRCh38
rs1026775093 Chr.17: 39763680-39763680 on Build GRCh38
rs138745067 Chr.17: 39763433-39763433 on Build GRCh38
rs999871235 Chr.17: 39763705-39763705 on Build GRCh38
rs924421199 Chr.17: 39763298-39763298 on Build GRCh38
rs933354605 Chr.17: 39763392-39763392 on Build GRCh38
rs970393761 Chr.17: 39763316-39763316 on Build GRCh38
rs529249551 Chr.17: 39763312-39763312 on Build GRCh38
rs143460560 Chr.17: 39763667-39763667 on Build GRCh38
rs945455611 Chr.17: 39763534-39763534 on Build GRCh38
rs564055088 Chr.17: 39763684-39763687 on Build GRCh38
rs78848907 Chr.17: 39763566-39763566 on Build GRCh38
rs995379554 Chr.17: 39763675-39763675 on Build GRCh38
rs936894859 Chr.17: 39763449-39763449 on Build GRCh38
rs910668673 Chr.17: 39763427-39763427 on Build GRCh38
rs183734888 Chr.17: 39763329-39763329 on Build GRCh38
rs914070576 Chr.17: 39763496-39763496 on Build GRCh38

View Full Product Details