Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145892165-145891656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA10
Entrez Gene ID
8515
Gene Name
integrin subunit alpha 10
Gene Aliases
PRO827
UniGene
Hs.158237
Gene Chromosome Location
Chr.1: 145891208-145910189 on Build GRCh38
Transcripts
NM_001303040.1 NM_001303041.1 NM_003637.4 XM_005277436.3 XM_011510083.2 XM_011510084.1 XM_017002622.1 XM_017002623.1 XM_017002624.1 XM_017002625.1 XM_017002626.1 XM_017002628.1

View Full Product Details