Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 7048733-7049252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PUDP
Entrez Gene ID
8226
Gene Name
pseudouridine 5'-phosphatase
Gene Aliases
DXF68S1E, FAM16AX, GS1, HDHD1, HDHD1A
UniGene
Hs.185910
Gene Chromosome Location
Chr.X: 6768840-7148190 on Build GRCh38
Transcripts
NM_001135565.1 NM_001178136.1 NM_012080.4

SNP Detail

SNP ID Chromosome Location
rs903227214 Chr.X: 7048902-7048902 on Build GRCh38
rs757202583 Chr.X: 7048856-7048856 on Build GRCh38
rs966442910 Chr.X: 7048887-7048887 on Build GRCh38
rs780880231 Chr.X: 7049032-7049032 on Build GRCh38
rs1023361862 Chr.X: 7049135-7049135 on Build GRCh38
rs916092868 Chr.X: 7049106-7049106 on Build GRCh38
rs893519888 Chr.X: 7048828-7048828 on Build GRCh38
rs1035931198 Chr.X: 7049076-7049076 on Build GRCh38
rs957885811 Chr.X: 7049116-7049116 on Build GRCh38
rs942886119 Chr.X: 7048840-7048840 on Build GRCh38
rs942073048 Chr.X: 7049115-7049115 on Build GRCh38
rs771902571 Chr.X: 7049038-7049038 on Build GRCh38
rs755072308 Chr.X: 7049185-7049185 on Build GRCh38
rs888616466 Chr.X: 7048758-7048758 on Build GRCh38
rs751686739 Chr.X: 7049178-7049178 on Build GRCh38
rs929322201 Chr.X: 7049133-7049133 on Build GRCh38
rs769595530 Chr.X: 7048844-7048845 on Build GRCh38
rs991593917 Chr.X: 7049104-7049104 on Build GRCh38
rs767543514 Chr.X: 7049191-7049191 on Build GRCh38
rs763433493 Chr.X: 7049128-7049128 on Build GRCh38
rs1020250627 Chr.X: 7048880-7048880 on Build GRCh38
rs369641681 Chr.X: 7049108-7049108 on Build GRCh38
rs773116737 Chr.X: 7048875-7048875 on Build GRCh38
rs919246928 Chr.X: 7049126-7049126 on Build GRCh38
rs901265552 Chr.X: 7048861-7048861 on Build GRCh38
rs977635425 Chr.X: 7049002-7049002 on Build GRCh38
rs998117374 Chr.X: 7048878-7048878 on Build GRCh38
rs1017470080 Chr.X: 7048771-7048771 on Build GRCh38
rs1032697372 Chr.X: 7049099-7049099 on Build GRCh38
rs6982 Chr.X: 7049134-7049134 on Build GRCh38

View Full Product Details