Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 7050247-7050749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PUDP
Entrez Gene ID
8226
Gene Name
pseudouridine 5'-phosphatase
Gene Aliases
DXF68S1E, FAM16AX, GS1, HDHD1, HDHD1A
UniGene
Hs.185910
Gene Chromosome Location
Chr.X: 6768840-7148190 on Build GRCh38
Transcripts
NM_001135565.1 NM_001178136.1 NM_012080.4

SNP Detail

SNP ID Chromosome Location
rs201195046 Chr.X: 7050484-7050484 on Build GRCh38
rs755713041 Chr.X: 7050501-7050501 on Build GRCh38
rs758819525 Chr.X: 7050355-7050355 on Build GRCh38
rs764778196 Chr.X: 7050331-7050331 on Build GRCh38
rs755837037 Chr.X: 7050394-7050394 on Build GRCh38
rs1056179405 Chr.X: 7050606-7050606 on Build GRCh38
rs1026448079 Chr.X: 7050688-7050688 on Build GRCh38
rs373339761 Chr.X: 7050275-7050275 on Build GRCh38
rs775472783 Chr.X: 7050609-7050609 on Build GRCh38
rs202174578 Chr.X: 7050303-7050303 on Build GRCh38
rs187299486 Chr.X: 7050694-7050694 on Build GRCh38
rs370293955 Chr.X: 7050311-7050311 on Build GRCh38
rs749169986 Chr.X: 7050408-7050408 on Build GRCh38
rs199601712 Chr.X: 7050453-7050453 on Build GRCh38
rs749155283 Chr.X: 7050294-7050294 on Build GRCh38
rs752120998 Chr.X: 7050367-7050367 on Build GRCh38
rs191631956 Chr.X: 7050726-7050726 on Build GRCh38
rs752293812 Chr.X: 7050343-7050343 on Build GRCh38
rs745826544 Chr.X: 7050419-7050419 on Build GRCh38
rs764035799 Chr.X: 7050665-7050665 on Build GRCh38
rs990618657 Chr.X: 7050582-7050582 on Build GRCh38
rs1047046855 Chr.X: 7050673-7050673 on Build GRCh38
rs964742229 Chr.X: 7050495-7050495 on Build GRCh38
rs752306230 Chr.X: 7050494-7050494 on Build GRCh38
rs369101263 Chr.X: 7050320-7050320 on Build GRCh38
rs886923410 Chr.X: 7050722-7050722 on Build GRCh38
rs200507468 Chr.X: 7050416-7050416 on Build GRCh38
rs778137581 Chr.X: 7050585-7050585 on Build GRCh38
rs1038463916 Chr.X: 7050650-7050650 on Build GRCh38
rs767059729 Chr.X: 7050361-7050361 on Build GRCh38
rs911893079 Chr.X: 7050489-7050489 on Build GRCh38
rs899387458 Chr.X: 7050654-7050654 on Build GRCh38
rs752254426 Chr.X: 7050382-7050382 on Build GRCh38
rs756997311 Chr.X: 7050521-7050521 on Build GRCh38
rs762063343 Chr.X: 7050684-7050684 on Build GRCh38
rs771287708 Chr.X: 7050482-7050482 on Build GRCh38
rs757838790 Chr.X: 7050520-7050521 on Build GRCh38
rs779633399 Chr.X: 7050477-7050477 on Build GRCh38
rs753544905 Chr.X: 7050527-7050527 on Build GRCh38
rs768680877 Chr.X: 7050421-7050421 on Build GRCh38
rs939257883 Chr.X: 7050627-7050627 on Build GRCh38
rs760154085 Chr.X: 7050483-7050483 on Build GRCh38
rs754125234 Chr.X: 7050682-7050682 on Build GRCh38
rs181818576 Chr.X: 7050553-7050553 on Build GRCh38
rs769429949 Chr.X: 7050289-7050289 on Build GRCh38
rs375074314 Chr.X: 7050293-7050293 on Build GRCh38
rs747321685 Chr.X: 7050588-7050588 on Build GRCh38
rs760787954 Chr.X: 7050613-7050613 on Build GRCh38
rs760095547 Chr.X: 7050545-7050545 on Build GRCh38
rs906461056 Chr.X: 7050379-7050379 on Build GRCh38
rs1046933816 Chr.X: 7050400-7050400 on Build GRCh38
rs377699850 Chr.X: 7050395-7050395 on Build GRCh38
rs763314596 Chr.X: 7050454-7050454 on Build GRCh38
rs370631064 Chr.X: 7050507-7050507 on Build GRCh38
rs886823967 Chr.X: 7050409-7050409 on Build GRCh38
rs915184958 Chr.X: 7050542-7050542 on Build GRCh38
rs770636195 Chr.X: 7050413-7050413 on Build GRCh38
rs377646267 Chr.X: 7050502-7050502 on Build GRCh38
rs780874096 Chr.X: 7050372-7050372 on Build GRCh38
rs765490594 Chr.X: 7050325-7050325 on Build GRCh38
rs946643157 Chr.X: 7050543-7050543 on Build GRCh38
rs750235884 Chr.X: 7050723-7050723 on Build GRCh38
rs950900804 Chr.X: 7050695-7050695 on Build GRCh38
rs374553245 Chr.X: 7050326-7050326 on Build GRCh38
rs201388017 Chr.X: 7050371-7050371 on Build GRCh38
rs189402457 Chr.X: 7050434-7050434 on Build GRCh38
rs776734289 Chr.X: 7050431-7050431 on Build GRCh38
rs185423088 Chr.X: 7050306-7050306 on Build GRCh38
rs760680411 Chr.X: 7050308-7050308 on Build GRCh38
rs770113328 Chr.X: 7050450-7050450 on Build GRCh38
rs754524342 Chr.X: 7050554-7050554 on Build GRCh38

View Full Product Details