Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132670458-132670961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C4
Entrez Gene ID
9329
Gene Name
general transcription factor IIIC subunit 4
Gene Aliases
KAT12, TF3C-delta, TFIII90, TFIIIC290, TFIIIC90, TFIIICDELTA
UniGene
Hs.22302
Gene Chromosome Location
Chr.9: 132670035-132694955 on Build GRCh38
Transcripts
NM_012204.3 NR_133925.1 XM_017015317.1 XM_017015318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3413406 c.160C>T p.R54W Chr.9: 132670758-132670758 on Build GRCh38
4881766 c.136G>T p.A46S Chr.9: 132670734-132670734 on Build GRCh38
7174931 c.276C>T p.I92I Chr.9: 132670874-132670874 on Build GRCh38
5547537 c.327_335delGCCCGCACC p.P110_P112delPAP Chr.9: 132670925-132670933 on Build GRCh38
6550563 c.23G>T p.R8L Chr.9: 132670621-132670621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755842655 Chr.9: 132670657-132670658 on Build GRCh38
rs755974065 Chr.9: 132670548-132670548 on Build GRCh38
rs181251276 Chr.9: 132670564-132670564 on Build GRCh38
rs763080138 Chr.9: 132670587-132670587 on Build GRCh38
rs947703253 Chr.9: 132670568-132670568 on Build GRCh38
rs951933277 Chr.9: 132670677-132670677 on Build GRCh38
rs767061987 Chr.9: 132670848-132670848 on Build GRCh38
rs923603068 Chr.9: 132670487-132670487 on Build GRCh38
rs948310457 Chr.9: 132670664-132670664 on Build GRCh38
rs771362964 Chr.9: 132670566-132670566 on Build GRCh38
rs996102613 Chr.9: 132670887-132670887 on Build GRCh38
rs770323812 Chr.9: 132670680-132670680 on Build GRCh38
rs199984441 Chr.9: 132670624-132670624 on Build GRCh38
rs199967299 Chr.9: 132670890-132670890 on Build GRCh38
rs1000996367 Chr.9: 132670494-132670494 on Build GRCh38
rs1022128165 Chr.9: 132670604-132670604 on Build GRCh38
rs747159211 Chr.9: 132670934-132670934 on Build GRCh38
rs916261658 Chr.9: 132670652-132670652 on Build GRCh38
rs1056060390 Chr.9: 132670491-132670491 on Build GRCh38
rs561975228 Chr.9: 132670507-132670507 on Build GRCh38
rs984553277 Chr.9: 132670549-132670549 on Build GRCh38
rs758351457 Chr.9: 132670925-132670925 on Build GRCh38
rs866919383 Chr.9: 132670786-132670786 on Build GRCh38
rs749804068 Chr.9: 132670697-132670697 on Build GRCh38
rs746264187 Chr.9: 132670648-132670648 on Build GRCh38
rs895377489 Chr.9: 132670508-132670508 on Build GRCh38
rs746664824 Chr.9: 132670565-132670565 on Build GRCh38
rs763652907 Chr.9: 132670746-132670746 on Build GRCh38
rs772224377 Chr.9: 132670721-132670721 on Build GRCh38
rs780496217 Chr.9: 132670689-132670689 on Build GRCh38
rs545464325 Chr.9: 132670820-132670820 on Build GRCh38
rs146201597 Chr.9: 132670634-132670634 on Build GRCh38
rs756370178 Chr.9: 132670770-132670770 on Build GRCh38
rs987225385 Chr.9: 132670515-132670515 on Build GRCh38
rs751203468 Chr.9: 132670644-132670644 on Build GRCh38
rs774493926 Chr.9: 132670605-132670605 on Build GRCh38
rs772332932 Chr.9: 132670817-132670817 on Build GRCh38
rs1045759050 Chr.9: 132670523-132670523 on Build GRCh38
rs1047261559 Chr.9: 132670729-132670729 on Build GRCh38
rs762948791 Chr.9: 132670839-132670839 on Build GRCh38
rs768668818 Chr.9: 132670702-132670702 on Build GRCh38
rs565515475 Chr.9: 132670579-132670579 on Build GRCh38
rs756998341 Chr.9: 132670645-132670645 on Build GRCh38
rs758109759 Chr.9: 132670558-132670558 on Build GRCh38
rs964380456 Chr.9: 132670519-132670519 on Build GRCh38
rs769426206 Chr.9: 132670834-132670834 on Build GRCh38
rs960462253 Chr.9: 132670623-132670623 on Build GRCh38
rs771315466 Chr.9: 132670935-132670935 on Build GRCh38
rs930094690 Chr.9: 132670727-132670727 on Build GRCh38
rs368918874 Chr.9: 132670482-132670482 on Build GRCh38
rs1008810586 Chr.9: 132670888-132670888 on Build GRCh38
rs974667368 Chr.9: 132670532-132670532 on Build GRCh38
rs763007124 Chr.9: 132670745-132670745 on Build GRCh38
rs765083568 Chr.9: 132670741-132670741 on Build GRCh38
rs899871735 Chr.9: 132670877-132670877 on Build GRCh38
rs567920424 Chr.9: 132670688-132670688 on Build GRCh38
rs749990071 Chr.9: 132670863-132670863 on Build GRCh38
rs776878483 Chr.9: 132670571-132670571 on Build GRCh38
rs891223754 Chr.9: 132670560-132670560 on Build GRCh38
rs746342503 Chr.9: 132670574-132670574 on Build GRCh38
rs779237127 Chr.9: 132670746-132670746 on Build GRCh38
rs775452875 Chr.9: 132670837-132670837 on Build GRCh38
rs775399601 Chr.9: 132670937-132670937 on Build GRCh38
rs757913856 Chr.9: 132670638-132670638 on Build GRCh38
rs748691331 Chr.9: 132670913-132670913 on Build GRCh38
rs779456083 Chr.9: 132670895-132670895 on Build GRCh38
rs373168131 Chr.9: 132670845-132670845 on Build GRCh38
rs751151981 Chr.9: 132670753-132670753 on Build GRCh38
rs866689535 Chr.9: 132670747-132670747 on Build GRCh38
rs1008328067 Chr.9: 132670849-132670849 on Build GRCh38
rs1001047248 Chr.9: 132670602-132670602 on Build GRCh38
rs756908231 Chr.9: 132670756-132670756 on Build GRCh38
rs867158126 Chr.9: 132670731-132670731 on Build GRCh38
rs780768246 Chr.9: 132670760-132670760 on Build GRCh38
rs529188832 Chr.9: 132670557-132670557 on Build GRCh38
rs1028873172 Chr.9: 132670675-132670675 on Build GRCh38
rs907262396 Chr.9: 132670536-132670536 on Build GRCh38
rs551059877 Chr.9: 132670618-132670618 on Build GRCh38
rs778934641 Chr.9: 132670794-132670794 on Build GRCh38
rs904072463 Chr.9: 132670586-132670586 on Build GRCh38
rs764300564 Chr.9: 132670597-132670597 on Build GRCh38
rs984605635 Chr.9: 132670690-132670690 on Build GRCh38
rs369871890 Chr.9: 132670755-132670755 on Build GRCh38
rs948318268 Chr.9: 132670516-132670516 on Build GRCh38
rs776466561 Chr.9: 132670739-132670739 on Build GRCh38
rs936931407 Chr.9: 132670797-132670797 on Build GRCh38
rs1038524443 Chr.9: 132670521-132670521 on Build GRCh38
rs764172649 Chr.9: 132670844-132670844 on Build GRCh38
rs1044725141 Chr.9: 132670781-132670781 on Build GRCh38
rs938988462 Chr.9: 132670488-132670488 on Build GRCh38
rs956785207 Chr.9: 132670502-132670502 on Build GRCh38
rs534985393 Chr.9: 132670718-132670718 on Build GRCh38
rs537208639 Chr.9: 132670627-132670627 on Build GRCh38
rs773240454 Chr.9: 132670818-132670818 on Build GRCh38
rs144693888 Chr.9: 132670926-132670926 on Build GRCh38
rs200999419 Chr.9: 132670871-132670871 on Build GRCh38
rs553661991 Chr.9: 132670774-132670774 on Build GRCh38
rs140834695 Chr.9: 132670868-132670868 on Build GRCh38
rs577948269 Chr.9: 132670776-132670776 on Build GRCh38
rs752623629 Chr.9: 132670744-132670744 on Build GRCh38
rs1031101014 Chr.9: 132670497-132670497 on Build GRCh38
rs13287671 Chr.9: 132670683-132670683 on Build GRCh38
rs1039064566 Chr.9: 132670703-132670703 on Build GRCh38
rs748278700 Chr.9: 132670804-132670804 on Build GRCh38
rs774437351 Chr.9: 132670709-132670709 on Build GRCh38
rs759314281 Chr.9: 132670740-132670740 on Build GRCh38
rs143172300 Chr.9: 132670608-132670608 on Build GRCh38
rs1020110450 Chr.9: 132670641-132670641 on Build GRCh38
rs375081797 Chr.9: 132670874-132670874 on Build GRCh38
rs781585893 Chr.9: 132670647-132670647 on Build GRCh38
rs370720484 Chr.9: 132670933-132670933 on Build GRCh38
rs753752930 Chr.9: 132670552-132670552 on Build GRCh38
rs750799025 Chr.9: 132670769-132670769 on Build GRCh38
rs769183687 Chr.9: 132670787-132670787 on Build GRCh38
rs754894300 Chr.9: 132670556-132670556 on Build GRCh38
rs765144204 Chr.9: 132670628-132670628 on Build GRCh38
rs867444042 Chr.9: 132670734-132670734 on Build GRCh38
rs977905479 Chr.9: 132670617-132670617 on Build GRCh38
rs775621143 Chr.9: 132670580-132670580 on Build GRCh38

View Full Product Details