Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133030595-133031100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C5
Entrez Gene ID
9328
Gene Name
general transcription factor IIIC subunit 5
Gene Aliases
TFIIIC63, TFIIICepsilon, TFiiiC2-63
UniGene
Hs.495417
Gene Chromosome Location
Chr.9: 133030675-133058503 on Build GRCh38
Transcripts
NM_001122823.1 NM_001286709.1 NM_012087.3 XM_005272234.3 XM_011519201.1 XM_017015315.1 XM_017015316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3905183 c.34G>A p.A12T Chr.9: 133031045-133031045 on Build GRCh38
1460755 c.15G>A p.A5A Chr.9: 133031026-133031026 on Build GRCh38
6499552 c.8G>C p.R3P Chr.9: 133030906-133030906 on Build GRCh38
753303 c.65G>A p.R22Q Chr.9: 133031076-133031076 on Build GRCh38
4611484 c.30delG p.A12fs*7 Chr.9: 133031041-133031041 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1056785701 Chr.9: 133030776-133030776 on Build GRCh38
rs771158991 Chr.9: 133030910-133030910 on Build GRCh38
rs758833918 Chr.9: 133030984-133030984 on Build GRCh38
rs1057167487 Chr.9: 133030831-133030831 on Build GRCh38
rs138716988 Chr.9: 133031062-133031062 on Build GRCh38
rs1048865303 Chr.9: 133030874-133030874 on Build GRCh38
rs532036458 Chr.9: 133031029-133031029 on Build GRCh38
rs1027028269 Chr.9: 133030673-133030673 on Build GRCh38
rs775195499 Chr.9: 133030731-133030731 on Build GRCh38
rs910200753 Chr.9: 133030752-133030752 on Build GRCh38
rs779168832 Chr.9: 133030981-133030981 on Build GRCh38
rs753610409 Chr.9: 133030966-133030966 on Build GRCh38
rs781219648 Chr.9: 133030993-133030993 on Build GRCh38
rs8193015 Chr.9: 133030651-133030651 on Build GRCh38
rs765972421 Chr.9: 133030964-133030964 on Build GRCh38
rs969254841 Chr.9: 133030695-133030695 on Build GRCh38
rs773340595 Chr.9: 133031020-133031020 on Build GRCh38
rs761598447 Chr.9: 133031008-133031008 on Build GRCh38
rs536084010 Chr.9: 133030836-133030836 on Build GRCh38
rs924324065 Chr.9: 133030762-133030762 on Build GRCh38
rs979065411 Chr.9: 133030952-133030952 on Build GRCh38
rs934392248 Chr.9: 133030767-133030767 on Build GRCh38
rs879608212 Chr.9: 133030741-133030741 on Build GRCh38
rs182627267 Chr.9: 133031047-133031047 on Build GRCh38
rs767444539 Chr.9: 133030934-133030934 on Build GRCh38
rs573210444 Chr.9: 133030759-133030759 on Build GRCh38
rs999010180 Chr.9: 133030906-133030906 on Build GRCh38
rs879595108 Chr.9: 133030689-133030689 on Build GRCh38
rs1047581846 Chr.9: 133030810-133030810 on Build GRCh38
rs904715355 Chr.9: 133030892-133030892 on Build GRCh38
rs755858739 Chr.9: 133030851-133030851 on Build GRCh38
rs142991886 Chr.9: 133030813-133030814 on Build GRCh38
rs118099172 Chr.9: 133030884-133030884 on Build GRCh38
rs745917354 Chr.9: 133031055-133031055 on Build GRCh38
rs748357917 Chr.9: 133030640-133030640 on Build GRCh38
rs759144039 Chr.9: 133031034-133031034 on Build GRCh38
rs778109094 Chr.9: 133031049-133031049 on Build GRCh38
rs775479885 Chr.9: 133030988-133030995 on Build GRCh38
rs920458286 Chr.9: 133030768-133030768 on Build GRCh38
rs558926499 Chr.9: 133030802-133030802 on Build GRCh38
rs761674799 Chr.9: 133030932-133030932 on Build GRCh38
rs978831202 Chr.9: 133030749-133030749 on Build GRCh38
rs994791532 Chr.9: 133030868-133030868 on Build GRCh38
rs757737273 Chr.9: 133031051-133031051 on Build GRCh38
rs770006836 Chr.9: 133031003-133031003 on Build GRCh38
rs1057436762 Chr.9: 133030648-133030648 on Build GRCh38
rs969029363 Chr.9: 133030745-133030745 on Build GRCh38
rs574979091 Chr.9: 133030887-133030887 on Build GRCh38
rs899078807 Chr.9: 133030843-133030843 on Build GRCh38
rs561517184 Chr.9: 133030713-133030713 on Build GRCh38
rs759802016 Chr.9: 133030919-133030919 on Build GRCh38
rs144659860 Chr.9: 133031056-133031056 on Build GRCh38
rs773109105 Chr.9: 133031069-133031069 on Build GRCh38
rs939248265 Chr.9: 133030819-133030819 on Build GRCh38
rs371143239 Chr.9: 133030815-133030815 on Build GRCh38
rs534175018 Chr.9: 133030772-133030772 on Build GRCh38
rs964705837 Chr.9: 133031007-133031007 on Build GRCh38
rs929094791 Chr.9: 133030781-133030781 on Build GRCh38
rs544572858 Chr.9: 133030882-133030882 on Build GRCh38
rs397740951 Chr.9: 133030659-133030659 on Build GRCh38
rs140936027 Chr.9: 133031030-133031030 on Build GRCh38
rs1019669375 Chr.9: 133030907-133030907 on Build GRCh38
rs781268552 Chr.9: 133031053-133031053 on Build GRCh38
rs760712648 Chr.9: 133031070-133031070 on Build GRCh38
rs917001166 Chr.9: 133030790-133030790 on Build GRCh38
rs899090663 Chr.9: 133030662-133030662 on Build GRCh38
rs779785987 Chr.9: 133030869-133030869 on Build GRCh38
rs920367013 Chr.9: 133031035-133031035 on Build GRCh38
rs542855608 Chr.9: 133030927-133030927 on Build GRCh38
rs200212860 Chr.9: 133030985-133030985 on Build GRCh38
rs554732040 Chr.9: 133030748-133030748 on Build GRCh38
rs747029308 Chr.9: 133030890-133030890 on Build GRCh38
rs754637922 Chr.9: 133030972-133030972 on Build GRCh38
rs746123269 Chr.9: 133030999-133030999 on Build GRCh38
rs904641279 Chr.9: 133031079-133031079 on Build GRCh38
rs755627393 Chr.9: 133030938-133030938 on Build GRCh38
rs979890294 Chr.9: 133031046-133031046 on Build GRCh38
rs78063416 Chr.9: 133030719-133030719 on Build GRCh38
rs752103957 Chr.9: 133031050-133031050 on Build GRCh38
rs901253166 Chr.9: 133031067-133031067 on Build GRCh38
rs773311831 Chr.9: 133030889-133030889 on Build GRCh38
rs924868993 Chr.9: 133030987-133030987 on Build GRCh38
rs895680953 Chr.9: 133030634-133030634 on Build GRCh38
rs536494166 Chr.9: 133030732-133030732 on Build GRCh38
rs1004330697 Chr.9: 133030710-133030710 on Build GRCh38
rs772583781 Chr.9: 133030888-133030888 on Build GRCh38
rs950429239 Chr.9: 133031046-133031046 on Build GRCh38
rs546878822 Chr.9: 133031012-133031012 on Build GRCh38
rs948511938 Chr.9: 133031038-133031038 on Build GRCh38
rs190910036 Chr.9: 133030636-133030636 on Build GRCh38
rs770251706 Chr.9: 133031010-133031010 on Build GRCh38
rs969358175 Chr.9: 133030921-133030921 on Build GRCh38
rs764878425 Chr.9: 133031041-133031041 on Build GRCh38
rs951529909 Chr.9: 133030700-133030700 on Build GRCh38
rs749957946 Chr.9: 133030936-133030936 on Build GRCh38
rs376349409 Chr.9: 133030989-133030989 on Build GRCh38
rs987136492 Chr.9: 133030727-133030727 on Build GRCh38
rs965459900 Chr.9: 133030909-133030909 on Build GRCh38
rs650627 Chr.9: 133030670-133030670 on Build GRCh38
rs934737049 Chr.9: 133031081-133031081 on Build GRCh38
rs371899002 Chr.9: 133031024-133031024 on Build GRCh38
rs776900278 Chr.9: 133030917-133030917 on Build GRCh38
rs201923823 Chr.9: 133030990-133030990 on Build GRCh38
rs780811165 Chr.9: 133030620-133030620 on Build GRCh38
rs17150121 Chr.9: 133030651-133030651 on Build GRCh38
rs1044254009 Chr.9: 133030641-133030641 on Build GRCh38
rs1019725409 Chr.9: 133030718-133030718 on Build GRCh38

View Full Product Details