Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133055933-133056431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C5
Entrez Gene ID
9328
Gene Name
general transcription factor IIIC subunit 5
Gene Aliases
TFIIIC63, TFIIICepsilon, TFiiiC2-63
UniGene
Hs.495417
Gene Chromosome Location
Chr.9: 133030675-133058503 on Build GRCh38
Transcripts
NM_001122823.1 NM_001286709.1 NM_012087.3 XM_005272234.3 XM_011519201.1 XM_017015315.1 XM_017015316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6418875 c.1240A>G p.N414D Chr.9: 133056084-133056084 on Build GRCh38
6873967 c.1205C>T p.P402L Chr.9: 133056049-133056049 on Build GRCh38
1756016 c.1208A>T p.Y403F Chr.9: 133056052-133056052 on Build GRCh38
4643328 c.1187G>A p.R396Q Chr.9: 133056031-133056031 on Build GRCh38
7370521 c.1225C>A p.Q409K Chr.9: 133056069-133056069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112990533 Chr.9: 133056179-133056179 on Build GRCh38
rs999277748 Chr.9: 133056300-133056300 on Build GRCh38
rs983626648 Chr.9: 133056267-133056267 on Build GRCh38
rs992666378 Chr.9: 133056154-133056154 on Build GRCh38
rs781142288 Chr.9: 133056126-133056126 on Build GRCh38
rs772552845 Chr.9: 133056006-133056006 on Build GRCh38
rs768795851 Chr.9: 133056122-133056123 on Build GRCh38
rs187879215 Chr.9: 133056098-133056098 on Build GRCh38
rs761659472 Chr.9: 133056037-133056037 on Build GRCh38
rs373201428 Chr.9: 133056033-133056033 on Build GRCh38
rs569972673 Chr.9: 133056174-133056174 on Build GRCh38
rs1013531294 Chr.9: 133056313-133056313 on Build GRCh38
rs778796055 Chr.9: 133056124-133056125 on Build GRCh38
rs748917240 Chr.9: 133055998-133055998 on Build GRCh38
rs1027705805 Chr.9: 133056395-133056395 on Build GRCh38
rs771401124 Chr.9: 133056122-133056122 on Build GRCh38
rs970120835 Chr.9: 133056369-133056369 on Build GRCh38
rs545641199 Chr.9: 133056376-133056376 on Build GRCh38
rs750028569 Chr.9: 133056283-133056283 on Build GRCh38
rs750352062 Chr.9: 133055964-133055964 on Build GRCh38
rs1040431209 Chr.9: 133056274-133056274 on Build GRCh38
rs573673713 Chr.9: 133056184-133056184 on Build GRCh38
rs191436953 Chr.9: 133056108-133056108 on Build GRCh38
rs775558352 Chr.9: 133056032-133056032 on Build GRCh38
rs535151124 Chr.9: 133056260-133056260 on Build GRCh38
rs920756849 Chr.9: 133056233-133056233 on Build GRCh38
rs537061033 Chr.9: 133056178-133056178 on Build GRCh38
rs753323854 Chr.9: 133056078-133056078 on Build GRCh38
rs746015258 Chr.9: 133056127-133056127 on Build GRCh38
rs952584526 Chr.9: 133056398-133056398 on Build GRCh38
rs1045155033 Chr.9: 133056195-133056195 on Build GRCh38
rs761528538 Chr.9: 133056182-133056182 on Build GRCh38
rs952111073 Chr.9: 133056404-133056404 on Build GRCh38
rs200715096 Chr.9: 133056092-133056092 on Build GRCh38
rs114135861 Chr.9: 133056130-133056130 on Build GRCh38
rs750413392 Chr.9: 133056047-133056047 on Build GRCh38
rs779391281 Chr.9: 133056075-133056075 on Build GRCh38
rs760208298 Chr.9: 133056309-133056309 on Build GRCh38
rs755664815 Chr.9: 133055981-133055981 on Build GRCh38
rs115755986 Chr.9: 133056155-133056155 on Build GRCh38
rs547161719 Chr.9: 133056014-133056014 on Build GRCh38
rs747548163 Chr.9: 133056116-133056116 on Build GRCh38
rs752394849 Chr.9: 133056099-133056099 on Build GRCh38
rs930861959 Chr.9: 133056030-133056030 on Build GRCh38
rs760559718 Chr.9: 133056054-133056054 on Build GRCh38
rs765840965 Chr.9: 133056077-133056077 on Build GRCh38
rs140832971 Chr.9: 133056024-133056024 on Build GRCh38
rs903737020 Chr.9: 133056165-133056165 on Build GRCh38
rs1040042669 Chr.9: 133056123-133056123 on Build GRCh38
rs952223523 Chr.9: 133056259-133056259 on Build GRCh38
rs903622399 Chr.9: 133056294-133056294 on Build GRCh38
rs1043490487 Chr.9: 133056292-133056292 on Build GRCh38
rs936745751 Chr.9: 133056269-133056269 on Build GRCh38
rs917283744 Chr.9: 133055983-133055983 on Build GRCh38
rs553191268 Chr.9: 133056301-133056301 on Build GRCh38
rs944995500 Chr.9: 133056272-133056272 on Build GRCh38
rs772953630 Chr.9: 133056145-133056145 on Build GRCh38
rs1024706739 Chr.9: 133056340-133056340 on Build GRCh38
rs769998718 Chr.9: 133056031-133056031 on Build GRCh38
rs150101735 Chr.9: 133055994-133055995 on Build GRCh38
rs779598015 Chr.9: 133055996-133055996 on Build GRCh38
rs983595468 Chr.9: 133056406-133056406 on Build GRCh38
rs369309090 Chr.9: 133056134-133056134 on Build GRCh38
rs199790336 Chr.9: 133055999-133055999 on Build GRCh38
rs931730412 Chr.9: 133056286-133056286 on Build GRCh38
rs925341158 Chr.9: 133056205-133056205 on Build GRCh38
rs139171552 Chr.9: 133056378-133056378 on Build GRCh38
rs767169275 Chr.9: 133056208-133056208 on Build GRCh38
rs763143722 Chr.9: 133056133-133056133 on Build GRCh38
rs778234950 Chr.9: 133056109-133056109 on Build GRCh38
rs368959551 Chr.9: 133056023-133056023 on Build GRCh38
rs908080579 Chr.9: 133056270-133056270 on Build GRCh38
rs572211208 Chr.9: 133056363-133056363 on Build GRCh38
rs767273512 Chr.9: 133056039-133056039 on Build GRCh38
rs999392252 Chr.9: 133056172-133056172 on Build GRCh38
rs749457842 Chr.9: 133056061-133056062 on Build GRCh38
rs776731746 Chr.9: 133056026-133056026 on Build GRCh38
rs980204162 Chr.9: 133056232-133056232 on Build GRCh38
rs759678432 Chr.9: 133056029-133056029 on Build GRCh38
rs771156086 Chr.9: 133056025-133056025 on Build GRCh38

View Full Product Details