Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 63189088-63189601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMJD1C
Entrez Gene ID
221037
Gene Name
jumonji domain containing 1C
Gene Aliases
TRIP-8, TRIP8
UniGene
Hs.413416
Gene Chromosome Location
Chr.10: 63167221-63522075 on Build GRCh38
Transcripts
NM_001282948.1 NM_001318153.1 NM_001318154.1 NM_001322252.1 NM_001322254.1 NM_001322258.1 NM_032776.2 NR_134512.1 XM_011539503.1 XM_017015897.1 XM_017015898.1 XM_017015899.1 XM_017015900.1 XM_017015901.1 XM_017015902.1 XM_017015903.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1627569 c.6389A>G p.N2130S Chr.10: 63189349-63189349 on Build GRCh38
465841 c.5643A>G p.E1881E Chr.10: 63189384-63189384 on Build GRCh38
7166739 c.6409G>C p.E2137Q Chr.10: 63189329-63189329 on Build GRCh38
4144780 c.6423A>T p.E2141D Chr.10: 63189315-63189315 on Build GRCh38
5803946 c.5818A>T p.S1940C Chr.10: 63189209-63189209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775570051 Chr.10: 63189401-63189401 on Build GRCh38
rs759721009 Chr.10: 63189269-63189269 on Build GRCh38
rs369068631 Chr.10: 63189328-63189328 on Build GRCh38
rs775597176 Chr.10: 63189494-63189494 on Build GRCh38
rs780606542 Chr.10: 63189146-63189146 on Build GRCh38
rs962426916 Chr.10: 63189242-63189242 on Build GRCh38
rs749634942 Chr.10: 63189350-63189350 on Build GRCh38
rs757976338 Chr.10: 63189121-63189121 on Build GRCh38
rs773164099 Chr.10: 63189425-63189425 on Build GRCh38
rs779329145 Chr.10: 63189473-63189473 on Build GRCh38
rs200368807 Chr.10: 63189311-63189311 on Build GRCh38
rs751191855 Chr.10: 63189329-63189329 on Build GRCh38
rs374161831 Chr.10: 63189446-63189446 on Build GRCh38
rs761256864 Chr.10: 63189477-63189477 on Build GRCh38
rs775330820 Chr.10: 63189468-63189468 on Build GRCh38
rs150004803 Chr.10: 63189500-63189500 on Build GRCh38
rs769512909 Chr.10: 63189465-63189467 on Build GRCh38
rs754646716 Chr.10: 63189334-63189334 on Build GRCh38
rs910809537 Chr.10: 63189364-63189364 on Build GRCh38
rs749794192 Chr.10: 63189459-63189459 on Build GRCh38
rs772246938 Chr.10: 63189397-63189397 on Build GRCh38
rs762916406 Chr.10: 63189433-63189433 on Build GRCh38
rs76935733 Chr.10: 63189165-63189165 on Build GRCh38
rs752775581 Chr.10: 63189279-63189279 on Build GRCh38
rs751378664 Chr.10: 63189441-63189441 on Build GRCh38
rs781226636 Chr.10: 63189369-63189371 on Build GRCh38
rs771077853 Chr.10: 63189143-63189146 on Build GRCh38
rs1023295589 Chr.10: 63189521-63189521 on Build GRCh38
rs1013205488 Chr.10: 63189520-63189520 on Build GRCh38
rs766173063 Chr.10: 63189438-63189438 on Build GRCh38
rs760753884 Chr.10: 63189408-63189408 on Build GRCh38
rs369773366 Chr.10: 63189352-63189352 on Build GRCh38
rs758211591 Chr.10: 63189317-63189317 on Build GRCh38
rs750159796 Chr.10: 63189313-63189313 on Build GRCh38
rs973036563 Chr.10: 63189338-63189338 on Build GRCh38
rs1052202962 Chr.10: 63189490-63189490 on Build GRCh38
rs763428242 Chr.10: 63189169-63189169 on Build GRCh38
rs1050551347 Chr.10: 63189516-63189516 on Build GRCh38
rs767609821 Chr.10: 63189273-63189273 on Build GRCh38
rs909078322 Chr.10: 63189234-63189234 on Build GRCh38
rs2306261 Chr.10: 63189561-63189561 on Build GRCh38
rs747616915 Chr.10: 63189337-63189337 on Build GRCh38
rs531453499 Chr.10: 63189411-63189411 on Build GRCh38
rs996325220 Chr.10: 63189502-63189502 on Build GRCh38
rs199852180 Chr.10: 63189188-63189188 on Build GRCh38
rs774630284 Chr.10: 63189353-63189353 on Build GRCh38
rs767204309 Chr.10: 63189138-63189138 on Build GRCh38
rs750004452 Chr.10: 63189554-63189554 on Build GRCh38
rs562569205 Chr.10: 63189449-63189449 on Build GRCh38
rs371897344 Chr.10: 63189140-63189140 on Build GRCh38
rs746218971 Chr.10: 63189476-63189476 on Build GRCh38
rs544671297 Chr.10: 63189456-63189456 on Build GRCh38
rs756632654 Chr.10: 63189340-63189340 on Build GRCh38
rs752205873 Chr.10: 63189451-63189451 on Build GRCh38
rs946339793 Chr.10: 63189495-63189495 on Build GRCh38
rs768224834 Chr.10: 63189323-63189323 on Build GRCh38
rs746086764 Chr.10: 63189381-63189381 on Build GRCh38
rs1040552447 Chr.10: 63189119-63189119 on Build GRCh38
rs1016977277 Chr.10: 63189299-63189299 on Build GRCh38
rs777467757 Chr.10: 63189458-63189458 on Build GRCh38
rs571334215 Chr.10: 63189470-63189470 on Build GRCh38
rs1006125584 Chr.10: 63189544-63189544 on Build GRCh38
rs889567490 Chr.10: 63189520-63189521 on Build GRCh38
rs200769337 Chr.10: 63189343-63189343 on Build GRCh38
rs935050534 Chr.10: 63189478-63189478 on Build GRCh38
rs528351194 Chr.10: 63189302-63189302 on Build GRCh38
rs542645192 Chr.10: 63189312-63189312 on Build GRCh38
rs748676870 Chr.10: 63189148-63189148 on Build GRCh38
rs1028058539 Chr.10: 63189173-63189173 on Build GRCh38
rs927398945 Chr.10: 63189430-63189430 on Build GRCh38
rs762867188 Chr.10: 63189467-63189468 on Build GRCh38
rs755721303 Chr.10: 63189454-63189454 on Build GRCh38
rs371983340 Chr.10: 63189247-63189247 on Build GRCh38
rs796262244 Chr.10: 63189529-63189529 on Build GRCh38
rs771233853 Chr.10: 63189216-63189216 on Build GRCh38
rs374145101 Chr.10: 63189143-63189143 on Build GRCh38
rs372436233 Chr.10: 63189336-63189336 on Build GRCh38

View Full Product Details