Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45714225-45714753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPP
Entrez Gene ID
3652
Gene Name
intracisternal A particle-promoted polypeptide
Gene Aliases
KLHL27
UniGene
Hs.744252
Gene Chromosome Location
Chr.1: 45694324-45750813 on Build GRCh38
Transcripts
NM_001145349.1 NM_005897.2 XM_005270839.4 XM_006710623.3 XM_011541394.2 XM_017001210.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
71181 c.1411A>G p.M471V Chr.1: 45714365-45714365 on Build GRCh38
3419280 c.1358G>A p.R453H Chr.1: 45714418-45714418 on Build GRCh38
3490252 c.1384T>C p.S462P Chr.1: 45714392-45714392 on Build GRCh38
5485705 c.1521C>A p.S507S Chr.1: 45714255-45714255 on Build GRCh38
5485706 c.1521C>A p.S507S Chr.1: 45714255-45714255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779069362 Chr.1: 45714501-45714501 on Build GRCh38
rs577010046 Chr.1: 45714387-45714387 on Build GRCh38
rs778981327 Chr.1: 45714415-45714415 on Build GRCh38
rs937397954 Chr.1: 45714423-45714423 on Build GRCh38
rs764490795 Chr.1: 45714281-45714281 on Build GRCh38
rs548196857 Chr.1: 45714522-45714522 on Build GRCh38
rs201907976 Chr.1: 45714325-45714325 on Build GRCh38
rs781384084 Chr.1: 45714252-45714252 on Build GRCh38
rs958078297 Chr.1: 45714357-45714357 on Build GRCh38
rs367794190 Chr.1: 45714552-45714552 on Build GRCh38
rs1055433032 Chr.1: 45714397-45714397 on Build GRCh38
rs776248909 Chr.1: 45714435-45714435 on Build GRCh38
rs754016664 Chr.1: 45714492-45714492 on Build GRCh38
rs898069160 Chr.1: 45714554-45714554 on Build GRCh38
rs774336352 Chr.1: 45714312-45714312 on Build GRCh38
rs752071344 Chr.1: 45714451-45714451 on Build GRCh38
rs185256852 Chr.1: 45714629-45714629 on Build GRCh38
rs374800097 Chr.1: 45714293-45714293 on Build GRCh38
rs34724416 Chr.1: 45714371-45714372 on Build GRCh38
rs755319684 Chr.1: 45714385-45714385 on Build GRCh38
rs759966812 Chr.1: 45714455-45714455 on Build GRCh38
rs530056489 Chr.1: 45714405-45714405 on Build GRCh38
rs933011517 Chr.1: 45714646-45714646 on Build GRCh38
rs560178384 Chr.1: 45714528-45714528 on Build GRCh38
rs770722167 Chr.1: 45714299-45714299 on Build GRCh38
rs745742233 Chr.1: 45714416-45714416 on Build GRCh38
rs760180998 Chr.1: 45714349-45714349 on Build GRCh38
rs768118112 Chr.1: 45714658-45714658 on Build GRCh38
rs775295472 Chr.1: 45714326-45714326 on Build GRCh38
rs572992251 Chr.1: 45714365-45714365 on Build GRCh38
rs867500384 Chr.1: 45714544-45714544 on Build GRCh38
rs1007318476 Chr.1: 45714725-45714725 on Build GRCh38
rs552279823 Chr.1: 45714553-45714553 on Build GRCh38
rs755236274 Chr.1: 45714251-45714251 on Build GRCh38
rs759372778 Chr.1: 45714316-45714316 on Build GRCh38
rs994419273 Chr.1: 45714592-45714592 on Build GRCh38
rs780978423 Chr.1: 45714659-45714659 on Build GRCh38
rs544485005 Chr.1: 45714396-45714396 on Build GRCh38
rs766926059 Chr.1: 45714446-45714446 on Build GRCh38
rs745833963 Chr.1: 45714502-45714502 on Build GRCh38
rs751991307 Chr.1: 45714377-45714377 on Build GRCh38
rs746536910 Chr.1: 45714656-45714657 on Build GRCh38
rs1051376542 Chr.1: 45714652-45714652 on Build GRCh38
rs1037327045 Chr.1: 45714498-45714498 on Build GRCh38
rs752953402 Chr.1: 45714470-45714470 on Build GRCh38
rs767897630 Chr.1: 45714467-45714467 on Build GRCh38
rs942169385 Chr.1: 45714510-45714510 on Build GRCh38
rs150261873 Chr.1: 45714366-45714366 on Build GRCh38
rs140212891 Chr.1: 45714270-45714270 on Build GRCh38
rs891272463 Chr.1: 45714728-45714728 on Build GRCh38
rs761229119 Chr.1: 45714438-45714438 on Build GRCh38
rs977765178 Chr.1: 45714373-45714373 on Build GRCh38
rs775387137 Chr.1: 45714419-45714419 on Build GRCh38
rs527760946 Chr.1: 45714542-45714542 on Build GRCh38
rs371580251 Chr.1: 45714427-45714427 on Build GRCh38
rs766844597 Chr.1: 45714376-45714376 on Build GRCh38
rs757426629 Chr.1: 45714493-45714493 on Build GRCh38
rs747929347 Chr.1: 45714340-45714340 on Build GRCh38
rs768175053 Chr.1: 45714430-45714430 on Build GRCh38
rs376914299 Chr.1: 45714487-45714487 on Build GRCh38
rs1036627907 Chr.1: 45714512-45714512 on Build GRCh38
rs562648318 Chr.1: 45714400-45714400 on Build GRCh38
rs756406369 Chr.1: 45714486-45714486 on Build GRCh38
rs779377398 Chr.1: 45714557-45714557 on Build GRCh38
rs749305413 Chr.1: 45714402-45714402 on Build GRCh38
rs771998580 Chr.1: 45714418-45714418 on Build GRCh38
rs3935297 Chr.1: 45714709-45714709 on Build GRCh38
rs201118075 Chr.1: 45714386-45714386 on Build GRCh38
rs769929125 Chr.1: 45714273-45714273 on Build GRCh38

View Full Product Details