Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18068380-18068739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12RB1
Entrez Gene ID
3594
Gene Name
interleukin 12 receptor subunit beta 1
Gene Aliases
CD212, IL-12R-BETA1, IL12RB, IMD30
UniGene
Hs.567294
Gene Chromosome Location
Chr.19: 18058994-18099027 on Build GRCh38
Transcripts
NM_001290023.1 NM_001290024.1 NM_005535.2 XM_006722741.3 XM_011527966.2 XM_011527967.2 XM_011527968.2 XM_011527969.2 XM_011527970.2 XM_011527971.2 XM_011527972.2 XM_011527973.2 XM_011527974.2 XM_011527975.2 XM_011527976.2 XM_017026762.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4075560 c.1217G>T p.G406V Chr.19: 18068499-18068499 on Build GRCh38
4075558 c.1260C>G p.A420A Chr.19: 18068456-18068456 on Build GRCh38
992990 c.1295C>T p.T432M Chr.19: 18068421-18068421 on Build GRCh38
5909717 c.1275G>T p.E425D Chr.19: 18068441-18068441 on Build GRCh38
6684849 c.1268A>C p.H423P Chr.19: 18068448-18068448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775923325 Chr.19: 18068443-18068443 on Build GRCh38
rs913507246 Chr.19: 18068455-18068455 on Build GRCh38
rs754764047 Chr.19: 18068466-18068466 on Build GRCh38
rs752642420 Chr.19: 18068475-18068475 on Build GRCh38
rs758241253 Chr.19: 18068486-18068486 on Build GRCh38
rs759893194 Chr.19: 18068431-18068431 on Build GRCh38
rs774555779 Chr.19: 18068516-18068516 on Build GRCh38
rs1026859419 Chr.19: 18068621-18068621 on Build GRCh38
rs1008128652 Chr.19: 18068552-18068552 on Build GRCh38
rs774271493 Chr.19: 18068450-18068450 on Build GRCh38
rs367647802 Chr.19: 18068447-18068447 on Build GRCh38
rs556121333 Chr.19: 18068639-18068639 on Build GRCh38
rs780762892 Chr.19: 18068494-18068494 on Build GRCh38
rs369102090 Chr.19: 18068575-18068575 on Build GRCh38
rs757004204 Chr.19: 18068493-18068493 on Build GRCh38
rs376271 Chr.19: 18068474-18068474 on Build GRCh38
rs771562593 Chr.19: 18068416-18068416 on Build GRCh38
rs12608554 Chr.19: 18068603-18068603 on Build GRCh38
rs767846981 Chr.19: 18068451-18068451 on Build GRCh38
rs777395626 Chr.19: 18068487-18068487 on Build GRCh38
rs376538454 Chr.19: 18068551-18068551 on Build GRCh38
rs1043211799 Chr.19: 18068492-18068492 on Build GRCh38
rs1019518857 Chr.19: 18068594-18068594 on Build GRCh38
rs375654921 Chr.19: 18068404-18068404 on Build GRCh38
rs763936009 Chr.19: 18068565-18068565 on Build GRCh38
rs747676268 Chr.19: 18068410-18068410 on Build GRCh38
rs999005899 Chr.19: 18068625-18068625 on Build GRCh38
rs181568874 Chr.19: 18068608-18068608 on Build GRCh38
rs867396174 Chr.19: 18068433-18068433 on Build GRCh38
rs952562732 Chr.19: 18068626-18068626 on Build GRCh38
rs559106838 Chr.19: 18068420-18068420 on Build GRCh38
rs1022617834 Chr.19: 18068713-18068713 on Build GRCh38
rs778663995 Chr.19: 18068538-18068538 on Build GRCh38
rs899825991 Chr.19: 18068555-18068555 on Build GRCh38
rs972579837 Chr.19: 18068432-18068432 on Build GRCh38
rs767867372 Chr.19: 18068523-18068523 on Build GRCh38
rs777322101 Chr.19: 18068417-18068417 on Build GRCh38
rs201548803 Chr.19: 18068454-18068454 on Build GRCh38
rs771616497 Chr.19: 18068532-18068533 on Build GRCh38
rs770296365 Chr.19: 18068437-18068437 on Build GRCh38
rs960243454 Chr.19: 18068627-18068627 on Build GRCh38
rs777041534 Chr.19: 18068423-18068423 on Build GRCh38
rs759435925 Chr.19: 18068534-18068534 on Build GRCh38
rs761731702 Chr.19: 18068519-18068519 on Build GRCh38
rs367838016 Chr.19: 18068661-18068661 on Build GRCh38
rs993035904 Chr.19: 18068691-18068691 on Build GRCh38
rs775796804 Chr.19: 18068506-18068506 on Build GRCh38
rs199504990 Chr.19: 18068499-18068499 on Build GRCh38
rs746490724 Chr.19: 18068489-18068489 on Build GRCh38
rs575013704 Chr.19: 18068650-18068650 on Build GRCh38
rs199755161 Chr.19: 18068444-18068444 on Build GRCh38
rs202048215 Chr.19: 18068508-18068508 on Build GRCh38
rs539168120 Chr.19: 18068407-18068407 on Build GRCh38
rs371881148 Chr.19: 18068539-18068539 on Build GRCh38
rs979906261 Chr.19: 18068665-18068665 on Build GRCh38
rs747584562 Chr.19: 18068645-18068645 on Build GRCh38
rs200875188 Chr.19: 18068421-18068421 on Build GRCh38

View Full Product Details