Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83310339-83310906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPSE
Entrez Gene ID
10855
Gene Name
heparanase
Gene Aliases
HPA, HPA1, HPR1, HPSE1, HSE1
UniGene
Hs.44227
Gene Chromosome Location
Chr.4: 83292461-83335153 on Build GRCh38
Transcripts
NM_001098540.2 NM_001166498.2 NM_001199830.1 NM_006665.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7352534 c.707T>G p.F236C Chr.4: 83310857-83310857 on Build GRCh38
4950926 c.779delA p.N260fs*20 Chr.4: 83310785-83310785 on Build GRCh38
74945 c.802G>A p.V268I Chr.4: 83310762-83310762 on Build GRCh38
3606596 c.815G>A p.R272Q Chr.4: 83310749-83310749 on Build GRCh38
5947830 c.820A>T p.K274* Chr.4: 83310744-83310744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781076238 Chr.4: 83310773-83310773 on Build GRCh38
rs150778862 Chr.4: 83310844-83310844 on Build GRCh38
rs575499964 Chr.4: 83310845-83310845 on Build GRCh38
rs993602541 Chr.4: 83310372-83310373 on Build GRCh38
rs756585984 Chr.4: 83310863-83310863 on Build GRCh38
rs771952831 Chr.4: 83310788-83310788 on Build GRCh38
rs901161666 Chr.4: 83310407-83310407 on Build GRCh38
rs924690520 Chr.4: 83310533-83310533 on Build GRCh38
rs140349032 Chr.4: 83310876-83310876 on Build GRCh38
rs770418887 Chr.4: 83310825-83310825 on Build GRCh38
rs537053894 Chr.4: 83310514-83310514 on Build GRCh38
rs768082904 Chr.4: 83310621-83310621 on Build GRCh38
rs759055125 Chr.4: 83310830-83310830 on Build GRCh38
rs138490774 Chr.4: 83310671-83310671 on Build GRCh38
rs749390915 Chr.4: 83310764-83310764 on Build GRCh38
rs776064636 Chr.4: 83310829-83310829 on Build GRCh38
rs750976278 Chr.4: 83310740-83310740 on Build GRCh38
rs954452040 Chr.4: 83310481-83310481 on Build GRCh38
rs1042208586 Chr.4: 83310531-83310531 on Build GRCh38
rs115487484 Chr.4: 83310513-83310513 on Build GRCh38
rs780103654 Chr.4: 83310646-83310646 on Build GRCh38
rs759247069 Chr.4: 83310720-83310720 on Build GRCh38
rs749143509 Chr.4: 83310490-83310490 on Build GRCh38
rs1037935314 Chr.4: 83310604-83310604 on Build GRCh38
rs369496444 Chr.4: 83310573-83310573 on Build GRCh38
rs534508715 Chr.4: 83310558-83310558 on Build GRCh38
rs779306396 Chr.4: 83310781-83310781 on Build GRCh38
rs552779301 Chr.4: 83310575-83310575 on Build GRCh38
rs61755719 Chr.4: 83310857-83310857 on Build GRCh38
rs760082862 Chr.4: 83310705-83310705 on Build GRCh38
rs148701049 Chr.4: 83310751-83310751 on Build GRCh38
rs142218633 Chr.4: 83310798-83310798 on Build GRCh38
rs757891984 Chr.4: 83310494-83310494 on Build GRCh38
rs570255676 Chr.4: 83310538-83310538 on Build GRCh38
rs970342581 Chr.4: 83310526-83310526 on Build GRCh38
rs764885481 Chr.4: 83310722-83310722 on Build GRCh38
rs982197001 Chr.4: 83310527-83310527 on Build GRCh38
rs557241980 Chr.4: 83310835-83310835 on Build GRCh38
rs139137504 Chr.4: 83310854-83310854 on Build GRCh38
rs145910527 Chr.4: 83310505-83310505 on Build GRCh38
rs757661122 Chr.4: 83310736-83310736 on Build GRCh38
rs776265748 Chr.4: 83310717-83310717 on Build GRCh38
rs777448793 Chr.4: 83310565-83310565 on Build GRCh38
rs987546504 Chr.4: 83310484-83310484 on Build GRCh38
rs753927046 Chr.4: 83310868-83310868 on Build GRCh38
rs370635319 Chr.4: 83310802-83310802 on Build GRCh38
rs747851387 Chr.4: 83310782-83310782 on Build GRCh38
rs903280683 Chr.4: 83310454-83310454 on Build GRCh38
rs770872092 Chr.4: 83310505-83310506 on Build GRCh38
rs765915260 Chr.4: 83310716-83310716 on Build GRCh38
rs6857500 Chr.4: 83310622-83310622 on Build GRCh38
rs756139166 Chr.4: 83310749-83310749 on Build GRCh38
rs200209715 Chr.4: 83310373-83310373 on Build GRCh38
rs1042119970 Chr.4: 83310672-83310672 on Build GRCh38
rs998211022 Chr.4: 83310435-83310435 on Build GRCh38
rs910218075 Chr.4: 83310488-83310488 on Build GRCh38
rs989385401 Chr.4: 83310512-83310512 on Build GRCh38
rs1020345201 Chr.4: 83310442-83310442 on Build GRCh38
rs12512551 Chr.4: 83310866-83310866 on Build GRCh38
rs770835401 Chr.4: 83310619-83310619 on Build GRCh38
rs1033093262 Chr.4: 83310750-83310750 on Build GRCh38
rs1026443784 Chr.4: 83310645-83310645 on Build GRCh38
rs11556201 Chr.4: 83310827-83310827 on Build GRCh38
rs746724240 Chr.4: 83310595-83310595 on Build GRCh38
rs779808746 Chr.4: 83310688-83310692 on Build GRCh38
rs942875159 Chr.4: 83310605-83310605 on Build GRCh38
rs6852183 Chr.4: 83310520-83310520 on Build GRCh38
rs751858151 Chr.4: 83310730-83310730 on Build GRCh38
rs780338532 Chr.4: 83310762-83310762 on Build GRCh38
rs140765573 Chr.4: 83310847-83310847 on Build GRCh38
rs200786477 Chr.4: 83310815-83310815 on Build GRCh38
rs767885091 Chr.4: 83310739-83310739 on Build GRCh38

View Full Product Details