Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33655521-33656018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPR3
Entrez Gene ID
3710
Gene Name
inositol 1,4,5-trisphosphate receptor type 3
Gene Aliases
IP3R, IP3R3
UniGene
Hs.65758
Gene Chromosome Location
Chr.6: 33621333-33696574 on Build GRCh38
Transcripts
NM_002224.3 XM_011514577.2 XM_017010832.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6253832 c.199G>T p.A67S Chr.6: 33655804-33655804 on Build GRCh38
1443833 c.189C>T p.N63N Chr.6: 33655794-33655794 on Build GRCh38
3873226 c.225G>A p.K75K Chr.6: 33655830-33655830 on Build GRCh38
1312093 c.231T>C p.T77T Chr.6: 33655836-33655836 on Build GRCh38
5726324 c.166C>T p.L56F Chr.6: 33655771-33655771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191542110 Chr.6: 33655907-33655907 on Build GRCh38
rs781556323 Chr.6: 33655905-33655905 on Build GRCh38
rs567027723 Chr.6: 33655951-33655951 on Build GRCh38
rs762654309 Chr.6: 33655822-33655822 on Build GRCh38
rs756494812 Chr.6: 33655761-33655762 on Build GRCh38
rs757724767 Chr.6: 33655904-33655904 on Build GRCh38
rs768228122 Chr.6: 33655921-33655921 on Build GRCh38
rs748171412 Chr.6: 33655737-33655737 on Build GRCh38
rs894660590 Chr.6: 33655648-33655648 on Build GRCh38
rs755200273 Chr.6: 33655724-33655724 on Build GRCh38
rs747804133 Chr.6: 33655850-33655852 on Build GRCh38
rs773776907 Chr.6: 33655825-33655825 on Build GRCh38
rs904632803 Chr.6: 33655919-33655919 on Build GRCh38
rs866335984 Chr.6: 33655783-33655783 on Build GRCh38
rs147411876 Chr.6: 33655741-33655743 on Build GRCh38
rs761766269 Chr.6: 33655832-33655832 on Build GRCh38
rs528193999 Chr.6: 33655582-33655582 on Build GRCh38
rs550908549 Chr.6: 33655870-33655870 on Build GRCh38
rs1018261239 Chr.6: 33655931-33655931 on Build GRCh38
rs374283783 Chr.6: 33655797-33655797 on Build GRCh38
rs142231270 Chr.6: 33655720-33655720 on Build GRCh38
rs747081724 Chr.6: 33655894-33655894 on Build GRCh38
rs148251650 Chr.6: 33655846-33655846 on Build GRCh38
rs778532227 Chr.6: 33655847-33655849 on Build GRCh38
rs1049671284 Chr.6: 33655640-33655641 on Build GRCh38
rs142283170 Chr.6: 33655803-33655803 on Build GRCh38
rs1018521596 Chr.6: 33655656-33655656 on Build GRCh38
rs1045967189 Chr.6: 33655655-33655655 on Build GRCh38
rs368973185 Chr.6: 33655902-33655903 on Build GRCh38
rs143158380 Chr.6: 33655845-33655845 on Build GRCh38
rs751853134 Chr.6: 33655892-33655893 on Build GRCh38
rs746247025 Chr.6: 33655807-33655807 on Build GRCh38
rs953865444 Chr.6: 33655740-33655740 on Build GRCh38
rs529007788 Chr.6: 33655574-33655574 on Build GRCh38
rs763641665 Chr.6: 33655824-33655824 on Build GRCh38
rs367604676 Chr.6: 33655926-33655926 on Build GRCh38
rs951631060 Chr.6: 33655721-33655721 on Build GRCh38
rs575008243 Chr.6: 33655890-33655891 on Build GRCh38
rs141238136 Chr.6: 33655857-33655857 on Build GRCh38
rs919658611 Chr.6: 33655602-33655602 on Build GRCh38
rs149835704 Chr.6: 33655891-33655892 on Build GRCh38
rs747527908 Chr.6: 33655796-33655796 on Build GRCh38
rs1028129760 Chr.6: 33655738-33655738 on Build GRCh38
rs775884452 Chr.6: 33655912-33655912 on Build GRCh38
rs941685419 Chr.6: 33655755-33655755 on Build GRCh38
rs752952768 Chr.6: 33655867-33655867 on Build GRCh38
rs964401502 Chr.6: 33655953-33655953 on Build GRCh38
rs777771192 Chr.6: 33655748-33655748 on Build GRCh38
rs373782032 Chr.6: 33655908-33655908 on Build GRCh38
rs998411882 Chr.6: 33655985-33655985 on Build GRCh38
rs771373957 Chr.6: 33655802-33655802 on Build GRCh38
rs766342194 Chr.6: 33655858-33655858 on Build GRCh38
rs774812114 Chr.6: 33655611-33655612 on Build GRCh38
rs200123519 Chr.6: 33655771-33655771 on Build GRCh38
rs533634577 Chr.6: 33655920-33655920 on Build GRCh38
rs200966677 Chr.6: 33655818-33655818 on Build GRCh38
rs775123437 Chr.6: 33655821-33655821 on Build GRCh38
rs78064999 Chr.6: 33655732-33655732 on Build GRCh38
rs139463901 Chr.6: 33655750-33655751 on Build GRCh38
rs200898330 Chr.6: 33655928-33655928 on Build GRCh38
rs147568121 Chr.6: 33655649-33655649 on Build GRCh38
rs977423539 Chr.6: 33655697-33655697 on Build GRCh38
rs968698670 Chr.6: 33655633-33655633 on Build GRCh38
rs931104730 Chr.6: 33655612-33655612 on Build GRCh38
rs964306320 Chr.6: 33655665-33655665 on Build GRCh38
rs772274152 Chr.6: 33655747-33655747 on Build GRCh38

View Full Product Details