Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33657725-33658230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPR3
Entrez Gene ID
3710
Gene Name
inositol 1,4,5-trisphosphate receptor type 3
Gene Aliases
IP3R, IP3R3
UniGene
Hs.65758
Gene Chromosome Location
Chr.6: 33621333-33696574 on Build GRCh38
Transcripts
NM_002224.3 XM_011514577.2 XM_017010832.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1316887 c.284A>T p.H95L Chr.6: 33657933-33657933 on Build GRCh38
1316889 c.286G>A p.A96T Chr.6: 33657935-33657935 on Build GRCh38
1078094 c.287C>T p.A96V Chr.6: 33657936-33657936 on Build GRCh38
6236306 c.289G>T p.A97S Chr.6: 33657938-33657938 on Build GRCh38
5405047 c.303G>A p.Q101Q Chr.6: 33657952-33657952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369332167 Chr.6: 33658055-33658055 on Build GRCh38
rs373557507 Chr.6: 33658067-33658067 on Build GRCh38
rs113761020 Chr.6: 33657833-33657834 on Build GRCh38
rs980830471 Chr.6: 33658175-33658176 on Build GRCh38
rs748014012 Chr.6: 33657823-33657824 on Build GRCh38
rs932411510 Chr.6: 33658129-33658129 on Build GRCh38
rs773275283 Chr.6: 33657896-33657896 on Build GRCh38
rs768293062 Chr.6: 33657995-33657995 on Build GRCh38
rs531922810 Chr.6: 33658095-33658095 on Build GRCh38
rs2274200 Chr.6: 33657919-33657919 on Build GRCh38
rs374380070 Chr.6: 33657831-33657834 on Build GRCh38
rs181352034 Chr.6: 33658169-33658169 on Build GRCh38
rs543316860 Chr.6: 33657994-33657994 on Build GRCh38
rs565380886 Chr.6: 33658185-33658185 on Build GRCh38
rs772896075 Chr.6: 33658044-33658044 on Build GRCh38
rs917932098 Chr.6: 33657940-33657940 on Build GRCh38
rs989512552 Chr.6: 33658127-33658127 on Build GRCh38
rs890793101 Chr.6: 33658202-33658202 on Build GRCh38
rs1015346235 Chr.6: 33657843-33657843 on Build GRCh38
rs3831079 Chr.6: 33657824-33657824 on Build GRCh38
rs780360454 Chr.6: 33657984-33657984 on Build GRCh38
rs372169367 Chr.6: 33657783-33657783 on Build GRCh38
rs753725957 Chr.6: 33657913-33657913 on Build GRCh38
rs746539714 Chr.6: 33657999-33658006 on Build GRCh38
rs138371134 Chr.6: 33657939-33657939 on Build GRCh38
rs927816060 Chr.6: 33657816-33657821 on Build GRCh38
rs996560402 Chr.6: 33657749-33657749 on Build GRCh38
rs528211739 Chr.6: 33657816-33657819 on Build GRCh38
rs924068343 Chr.6: 33657979-33657979 on Build GRCh38
rs766573065 Chr.6: 33658094-33658094 on Build GRCh38
rs577614233 Chr.6: 33657814-33657814 on Build GRCh38
rs1054198146 Chr.6: 33658068-33658068 on Build GRCh38
rs760750869 Chr.6: 33657900-33657900 on Build GRCh38
rs530714835 Chr.6: 33657818-33657819 on Build GRCh38
rs758598534 Chr.6: 33657917-33657917 on Build GRCh38
rs1008574794 Chr.6: 33657835-33657835 on Build GRCh38
rs372392252 Chr.6: 33657857-33657857 on Build GRCh38
rs142845743 Chr.6: 33657958-33657958 on Build GRCh38
rs766254344 Chr.6: 33657901-33657901 on Build GRCh38
rs201717246 Chr.6: 33658043-33658043 on Build GRCh38
rs959621125 Chr.6: 33657882-33657882 on Build GRCh38
rs1027058856 Chr.6: 33657856-33657856 on Build GRCh38
rs3831080 Chr.6: 33657816-33657817 on Build GRCh38
rs376628421 Chr.6: 33657936-33657936 on Build GRCh38
rs544995573 Chr.6: 33657851-33657851 on Build GRCh38
rs553764730 Chr.6: 33657887-33657887 on Build GRCh38
rs778432469 Chr.6: 33658041-33658041 on Build GRCh38
rs200213977 Chr.6: 33657966-33657966 on Build GRCh38
rs969473158 Chr.6: 33657815-33657816 on Build GRCh38
rs372548498 Chr.6: 33658042-33658042 on Build GRCh38
rs767419764 Chr.6: 33657895-33657895 on Build GRCh38
rs913861471 Chr.6: 33657879-33657879 on Build GRCh38
rs556464598 Chr.6: 33657850-33657850 on Build GRCh38
rs1028644969 Chr.6: 33657766-33657766 on Build GRCh38

View Full Product Details