Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150950831-150951331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH2
Entrez Gene ID
3757
Gene Name
potassium voltage-gated channel subfamily H member 2
Gene Aliases
ERG-1, ERG1, H-ERG, HERG, HERG1, Kv11.1, LQT2, SQT1
UniGene
Hs.647099
Gene Chromosome Location
Chr.7: 150944956-150978314 on Build GRCh38
Transcripts
NM_000238.3 NM_001204798.1 NM_172056.2 NM_172057.2 XM_011516185.2 XM_011516186.2 XM_017012195.1 XM_017012196.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6690865 c.2087G>A p.R696H Chr.7: 150950979-150950979 on Build GRCh38
5649010 c.2110G>A p.A704T Chr.7: 150950956-150950956 on Build GRCh38
1241320 c.1959T>C p.A653A Chr.7: 150951107-150951107 on Build GRCh38
6177233 c.2143G>C p.A715P Chr.7: 150950923-150950923 on Build GRCh38
4694325 c.2019C>T p.Y673Y Chr.7: 150951047-150951047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750045237 Chr.7: 150951144-150951144 on Build GRCh38
rs199472985 Chr.7: 150950949-150950949 on Build GRCh38
rs371749379 Chr.7: 150950912-150950912 on Build GRCh38
rs745370146 Chr.7: 150951051-150951051 on Build GRCh38
rs794728382 Chr.7: 150950962-150950962 on Build GRCh38
rs794728501 Chr.7: 150950982-150950982 on Build GRCh38
rs540263080 Chr.7: 150951257-150951257 on Build GRCh38
rs1039561682 Chr.7: 150951222-150951222 on Build GRCh38
rs755537090 Chr.7: 150951234-150951234 on Build GRCh38
rs556108319 Chr.7: 150951293-150951293 on Build GRCh38
rs550680502 Chr.7: 150950969-150950969 on Build GRCh38
rs765717858 Chr.7: 150950986-150950986 on Build GRCh38
rs778135438 Chr.7: 150951013-150951013 on Build GRCh38
rs1026484955 Chr.7: 150951205-150951205 on Build GRCh38
rs199473532 Chr.7: 150950935-150950935 on Build GRCh38
rs369388440 Chr.7: 150950881-150950882 on Build GRCh38
rs199472983 Chr.7: 150950988-150950988 on Build GRCh38
rs777341700 Chr.7: 150950910-150950910 on Build GRCh38
rs576727639 Chr.7: 150951290-150951290 on Build GRCh38
rs41314369 Chr.7: 150950905-150950905 on Build GRCh38
rs794728383 Chr.7: 150950985-150950985 on Build GRCh38
rs756870695 Chr.7: 150951036-150951036 on Build GRCh38
rs904695533 Chr.7: 150951273-150951273 on Build GRCh38
rs887593798 Chr.7: 150951072-150951072 on Build GRCh38
rs371820756 Chr.7: 150951134-150951134 on Build GRCh38
rs918568445 Chr.7: 150951241-150951241 on Build GRCh38
rs567628192 Chr.7: 150951030-150951030 on Build GRCh38
rs779504703 Chr.7: 150951121-150951122 on Build GRCh38
rs769197005 Chr.7: 150951052-150951052 on Build GRCh38
rs199852343 Chr.7: 150950932-150950932 on Build GRCh38
rs41314372 Chr.7: 150950906-150950906 on Build GRCh38
rs2072413 Chr.7: 150950881-150950881 on Build GRCh38
rs753149816 Chr.7: 150951011-150951011 on Build GRCh38
rs1001618150 Chr.7: 150951280-150951280 on Build GRCh38
rs748872880 Chr.7: 150951172-150951172 on Build GRCh38
rs755482792 Chr.7: 150950890-150950890 on Build GRCh38
rs199472976 Chr.7: 150951120-150951120 on Build GRCh38
rs794728443 Chr.7: 150950951-150950951 on Build GRCh38
rs199472981 Chr.7: 150951033-150951033 on Build GRCh38
rs574433640 Chr.7: 150951237-150951237 on Build GRCh38
rs930085189 Chr.7: 150951243-150951243 on Build GRCh38
rs562349092 Chr.7: 150950933-150950933 on Build GRCh38
rs1048480835 Chr.7: 150951254-150951254 on Build GRCh38
rs554733186 Chr.7: 150951225-150951225 on Build GRCh38
rs774258761 Chr.7: 150951086-150951086 on Build GRCh38
rs758751607 Chr.7: 150951012-150951012 on Build GRCh38
rs200905835 Chr.7: 150951141-150951141 on Build GRCh38
rs1032073874 Chr.7: 150951185-150951185 on Build GRCh38
rs780887954 Chr.7: 150951046-150951046 on Build GRCh38
rs376848355 Chr.7: 150951136-150951136 on Build GRCh38
rs199473531 Chr.7: 150950979-150950979 on Build GRCh38
rs933916948 Chr.7: 150951236-150951236 on Build GRCh38
rs538413875 Chr.7: 150951197-150951197 on Build GRCh38
rs543261098 Chr.7: 150950874-150950874 on Build GRCh38
rs767761418 Chr.7: 150950872-150950872 on Build GRCh38
rs553107049 Chr.7: 150951092-150951092 on Build GRCh38
rs199472984 Chr.7: 150950980-150950980 on Build GRCh38
rs794728490 Chr.7: 150951110-150951112 on Build GRCh38
rs794728375 Chr.7: 150951121-150951121 on Build GRCh38
rs776736691 Chr.7: 150951125-150951125 on Build GRCh38
rs769765942 Chr.7: 150950929-150950929 on Build GRCh38
rs751915754 Chr.7: 150951140-150951140 on Build GRCh38
rs768572372 Chr.7: 150950939-150950939 on Build GRCh38
rs199472980 Chr.7: 150951081-150951081 on Build GRCh38
rs201021019 Chr.7: 150950904-150950904 on Build GRCh38
rs373097096 Chr.7: 150951223-150951223 on Build GRCh38
rs772508319 Chr.7: 150951259-150951259 on Build GRCh38
rs1137617 Chr.7: 150951110-150951110 on Build GRCh38
rs968886132 Chr.7: 150950975-150950975 on Build GRCh38
rs755822548 Chr.7: 150950888-150950898 on Build GRCh38
rs763430080 Chr.7: 150951137-150951137 on Build GRCh38
rs756193223 Chr.7: 150950878-150950878 on Build GRCh38
rs767094055 Chr.7: 150951143-150951143 on Build GRCh38
rs199472978 Chr.7: 150951097-150951097 on Build GRCh38
rs531272738 Chr.7: 150950863-150950863 on Build GRCh38
rs147739629 Chr.7: 150950864-150950864 on Build GRCh38
rs794728488 Chr.7: 150951122-150951122 on Build GRCh38
rs181086490 Chr.7: 150951305-150951305 on Build GRCh38
rs768519164 Chr.7: 150951084-150951084 on Build GRCh38
rs774672362 Chr.7: 150951101-150951101 on Build GRCh38
rs746696683 Chr.7: 150950911-150950911 on Build GRCh38
rs990955022 Chr.7: 150951229-150951229 on Build GRCh38
rs569487823 Chr.7: 150951190-150951190 on Build GRCh38
rs794728381 Chr.7: 150951040-150951040 on Build GRCh38
rs375864407 Chr.7: 150951067-150951068 on Build GRCh38
rs779970711 Chr.7: 150951054-150951054 on Build GRCh38
rs553486233 Chr.7: 150951258-150951258 on Build GRCh38
rs199472977 Chr.7: 150951099-150951099 on Build GRCh38
rs794728384 Chr.7: 150950921-150950921 on Build GRCh38
rs199472979 Chr.7: 150951087-150951087 on Build GRCh38
rs753966347 Chr.7: 150950974-150950974 on Build GRCh38
rs369192064 Chr.7: 150951139-150951139 on Build GRCh38
rs367997739 Chr.7: 150951129-150951129 on Build GRCh38
rs886039385 Chr.7: 150950920-150950920 on Build GRCh38
rs755607860 Chr.7: 150951145-150951145 on Build GRCh38
rs202214833 Chr.7: 150950957-150950957 on Build GRCh38
rs779449553 Chr.7: 150951158-150951158 on Build GRCh38
rs79624542 Chr.7: 150951031-150951031 on Build GRCh38
rs2072412 Chr.7: 150950882-150950882 on Build GRCh38
rs942551390 Chr.7: 150951196-150951196 on Build GRCh38
rs794728500 Chr.7: 150951013-150951013 on Build GRCh38
rs909710218 Chr.7: 150951188-150951188 on Build GRCh38
rs199472982 Chr.7: 150951007-150951007 on Build GRCh38
rs780656919 Chr.7: 150950922-150950922 on Build GRCh38
rs552000706 Chr.7: 150950924-150950924 on Build GRCh38
rs749211387 Chr.7: 150951063-150951063 on Build GRCh38
rs773524985 Chr.7: 150950956-150950956 on Build GRCh38

View Full Product Details