Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 88270627-88271225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK10
Entrez Gene ID
54207
Gene Name
potassium two pore domain channel subfamily K member 10
Gene Aliases
K2p10.1, PPP1R97, TREK-2, TREK2
UniGene
Hs.560255
Gene Chromosome Location
Chr.14: 88180108-88326912 on Build GRCh38
Transcripts
NM_021161.4 NM_138317.2 NM_138318.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1323353 c.13T>C p.F5L Chr.14: 88270777-88270777 on Build GRCh38
5773710 c.47A>T p.D16V Chr.14: 88270743-88270743 on Build GRCh38
2250285 c.41G>A p.R14H Chr.14: 88270749-88270749 on Build GRCh38
3498523 c.26G>A p.R9K Chr.14: 88270764-88270764 on Build GRCh38
3498524 c.18G>A p.K6K Chr.14: 88270772-88270772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762030307 Chr.14: 88270812-88270812 on Build GRCh38
rs537251609 Chr.14: 88270807-88270807 on Build GRCh38
rs769877176 Chr.14: 88270650-88270650 on Build GRCh38
rs373698918 Chr.14: 88270896-88270896 on Build GRCh38
rs531065600 Chr.14: 88270981-88270981 on Build GRCh38
rs527539054 Chr.14: 88270783-88270783 on Build GRCh38
rs769558527 Chr.14: 88270940-88270940 on Build GRCh38
rs764706419 Chr.14: 88271023-88271023 on Build GRCh38
rs780260867 Chr.14: 88270832-88270832 on Build GRCh38
rs1034284512 Chr.14: 88270667-88270667 on Build GRCh38
rs966470057 Chr.14: 88271080-88271080 on Build GRCh38
rs1055825512 Chr.14: 88271173-88271173 on Build GRCh38
rs75962207 Chr.14: 88270973-88270973 on Build GRCh38
rs1029164337 Chr.14: 88270976-88270976 on Build GRCh38
rs994203469 Chr.14: 88271006-88271006 on Build GRCh38
rs751401506 Chr.14: 88271198-88271198 on Build GRCh38
rs373648441 Chr.14: 88270752-88270752 on Build GRCh38
rs762567599 Chr.14: 88270800-88270800 on Build GRCh38
rs546315252 Chr.14: 88270871-88270871 on Build GRCh38
rs754013514 Chr.14: 88270834-88270834 on Build GRCh38
rs565399104 Chr.14: 88271126-88271126 on Build GRCh38
rs1043015669 Chr.14: 88270880-88270880 on Build GRCh38
rs377549070 Chr.14: 88270939-88270939 on Build GRCh38
rs770576335 Chr.14: 88270892-88270892 on Build GRCh38
rs534631702 Chr.14: 88271127-88271127 on Build GRCh38
rs776373706 Chr.14: 88270760-88270760 on Build GRCh38
rs1040530683 Chr.14: 88270962-88270962 on Build GRCh38
rs1026258498 Chr.14: 88270804-88270804 on Build GRCh38
rs775036016 Chr.14: 88270791-88270791 on Build GRCh38
rs4904441 Chr.14: 88271054-88271054 on Build GRCh38
rs751249403 Chr.14: 88270809-88270809 on Build GRCh38
rs895202647 Chr.14: 88270708-88270708 on Build GRCh38
rs759215898 Chr.14: 88271022-88271022 on Build GRCh38
rs991996210 Chr.14: 88270732-88270732 on Build GRCh38
rs777859574 Chr.14: 88270816-88270819 on Build GRCh38
rs914904414 Chr.14: 88270872-88270872 on Build GRCh38
rs1298237 Chr.14: 88270849-88270849 on Build GRCh38
rs78415523 Chr.14: 88271088-88271088 on Build GRCh38
rs917770350 Chr.14: 88270748-88270748 on Build GRCh38
rs182406892 Chr.14: 88270749-88270749 on Build GRCh38
rs774720928 Chr.14: 88270855-88270855 on Build GRCh38
rs769285872 Chr.14: 88270778-88270778 on Build GRCh38
rs780348688 Chr.14: 88270737-88270737 on Build GRCh38
rs759470354 Chr.14: 88270769-88270769 on Build GRCh38
rs200578491 Chr.14: 88270750-88270750 on Build GRCh38
rs750962866 Chr.14: 88270826-88270826 on Build GRCh38
rs570282212 Chr.14: 88270893-88270893 on Build GRCh38
rs373233223 Chr.14: 88270653-88270653 on Build GRCh38
rs1005600703 Chr.14: 88270722-88270722 on Build GRCh38
rs987296094 Chr.14: 88270846-88270846 on Build GRCh38
rs989218338 Chr.14: 88271066-88271066 on Build GRCh38
rs574614791 Chr.14: 88270756-88270756 on Build GRCh38
rs370283141 Chr.14: 88270870-88270870 on Build GRCh38
rs547033979 Chr.14: 88271024-88271024 on Build GRCh38
rs376872265 Chr.14: 88270830-88270830 on Build GRCh38
rs916738394 Chr.14: 88271177-88271177 on Build GRCh38
rs915018703 Chr.14: 88271073-88271073 on Build GRCh38
rs936608579 Chr.14: 88271182-88271182 on Build GRCh38
rs150467519 Chr.14: 88270974-88270974 on Build GRCh38
rs557150896 Chr.14: 88270821-88270821 on Build GRCh38
rs767721168 Chr.14: 88270755-88270756 on Build GRCh38
rs939581235 Chr.14: 88271172-88271172 on Build GRCh38
rs755114982 Chr.14: 88270838-88270838 on Build GRCh38
rs750984923 Chr.14: 88270709-88270709 on Build GRCh38
rs551165171 Chr.14: 88271015-88271015 on Build GRCh38
rs756341926 Chr.14: 88270759-88270760 on Build GRCh38
rs957397764 Chr.14: 88270842-88270842 on Build GRCh38
rs945054721 Chr.14: 88270875-88270875 on Build GRCh38
rs756197292 Chr.14: 88270730-88270730 on Build GRCh38
rs746740042 Chr.14: 88270753-88270753 on Build GRCh38
rs897148173 Chr.14: 88271005-88271005 on Build GRCh38
rs936282789 Chr.14: 88270816-88270817 on Build GRCh38
rs752848098 Chr.14: 88270758-88270759 on Build GRCh38
rs1017396888 Chr.14: 88271034-88271034 on Build GRCh38

View Full Product Details