Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32444573-32445076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DRA
Entrez Gene ID
3122
Gene Name
major histocompatibility complex, class II, DR alpha
Gene Aliases
HLA-DRA1, MLRW
UniGene
Hs.520048
Gene Chromosome Location
Chr.6: 32439842-32445046 on Build GRCh38
Transcripts
NM_019111.4

SNP Detail

SNP ID Chromosome Location
rs557028220 Chr.6: 32444875-32444875 on Build GRCh38
rs777586636 Chr.6: 32444761-32444761 on Build GRCh38
rs1041885 Chr.6: 32445032-32445032 on Build GRCh38
rs1051015 Chr.6: 32444755-32444755 on Build GRCh38
rs550585478 Chr.6: 32444628-32444628 on Build GRCh38
rs781366068 Chr.6: 32444954-32444954 on Build GRCh38
rs950105583 Chr.6: 32444880-32444880 on Build GRCh38
rs7197 Chr.6: 32444803-32444803 on Build GRCh38
rs903165614 Chr.6: 32444915-32444915 on Build GRCh38
rs564163628 Chr.6: 32444610-32444610 on Build GRCh38
rs1052916 Chr.6: 32445009-32445009 on Build GRCh38
rs935964121 Chr.6: 32444939-32444939 on Build GRCh38
rs7196 Chr.6: 32444794-32444794 on Build GRCh38
rs4935356 Chr.6: 32444611-32444611 on Build GRCh38
rs567006522 Chr.6: 32444774-32444774 on Build GRCh38
rs996497312 Chr.6: 32445000-32445000 on Build GRCh38
rs111471704 Chr.6: 32444889-32444889 on Build GRCh38
rs893688624 Chr.6: 32444970-32444970 on Build GRCh38
rs4935354 Chr.6: 32444621-32444621 on Build GRCh38
rs976725617 Chr.6: 32444817-32444817 on Build GRCh38
rs7194 Chr.6: 32444703-32444703 on Build GRCh38
rs1131541 Chr.6: 32444789-32444789 on Build GRCh38
rs188003551 Chr.6: 32444897-32444897 on Build GRCh38
rs953354820 Chr.6: 32444940-32444940 on Build GRCh38
rs3135390 Chr.6: 32444618-32444618 on Build GRCh38
rs987219934 Chr.6: 32444788-32444788 on Build GRCh38
rs1063590 Chr.6: 32444966-32444966 on Build GRCh38
rs1051336 Chr.6: 32444815-32444815 on Build GRCh38
rs1053043 Chr.6: 32445015-32445015 on Build GRCh38
rs1012655688 Chr.6: 32444656-32444656 on Build GRCh38
rs115857426 Chr.6: 32444603-32444603 on Build GRCh38
rs3177928 Chr.6: 32444658-32444658 on Build GRCh38
rs986215091 Chr.6: 32444974-32444974 on Build GRCh38
rs1052531 Chr.6: 32444964-32444964 on Build GRCh38
rs937599809 Chr.6: 32444835-32444835 on Build GRCh38
rs866495193 Chr.6: 32444986-32444986 on Build GRCh38
rs1008585710 Chr.6: 32445047-32445047 on Build GRCh38
rs3191043 Chr.6: 32444679-32444679 on Build GRCh38
rs7195 Chr.6: 32444762-32444762 on Build GRCh38
rs112637929 Chr.6: 32445046-32445046 on Build GRCh38
rs890318115 Chr.6: 32445035-32445035 on Build GRCh38
rs991632602 Chr.6: 32444847-32444847 on Build GRCh38

View Full Product Details