Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33068941-33069445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DPA1
Entrez Gene ID
3113
Gene Name
major histocompatibility complex, class II, DP alpha 1
Gene Aliases
DP(W3), DP(W4), HLA-DP1A, HLADP, HLASB, PLT1
UniGene
Hs.347270
Gene Chromosome Location
Chr.6: 33064569-33080778 on Build GRCh38
Transcripts
NM_001242524.1 NM_001242525.1 NM_033554.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5003198 c.413G>T p.C138F Chr.6: 33069234-33069234 on Build GRCh38
3349257 c.353C>A p.P118H Chr.6: 33069294-33069294 on Build GRCh38
7171746 c.616C>G p.L206V Chr.6: 33069031-33069031 on Build GRCh38
4690654 c.353_354insC p.E119fs*1 Chr.6: 33069293-33069294 on Build GRCh38
7273253 c.627G>T p.W209C Chr.6: 33069020-33069020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773990109 Chr.6: 33069061-33069061 on Build GRCh38
rs754800174 Chr.6: 33069298-33069298 on Build GRCh38
rs2308927 Chr.6: 33069200-33069200 on Build GRCh38
rs34950776 Chr.6: 33069327-33069327 on Build GRCh38
rs780344579 Chr.6: 33069255-33069255 on Build GRCh38
rs779689086 Chr.6: 33069086-33069086 on Build GRCh38
rs150225191 Chr.6: 33069094-33069094 on Build GRCh38
rs775560319 Chr.6: 33069000-33069000 on Build GRCh38
rs770665831 Chr.6: 33068976-33068976 on Build GRCh38
rs571128277 Chr.6: 33069047-33069047 on Build GRCh38
rs765581858 Chr.6: 33069042-33069042 on Build GRCh38
rs763690407 Chr.6: 33069061-33069061 on Build GRCh38
rs775670754 Chr.6: 33069157-33069157 on Build GRCh38
rs41552628 Chr.6: 33069092-33069092 on Build GRCh38
rs566020840 Chr.6: 33069395-33069395 on Build GRCh38
rs774804146 Chr.6: 33069332-33069332 on Build GRCh38
rs749909092 Chr.6: 33069288-33069288 on Build GRCh38
rs150537907 Chr.6: 33069238-33069238 on Build GRCh38
rs370530697 Chr.6: 33069035-33069035 on Build GRCh38
rs573850653 Chr.6: 33069320-33069320 on Build GRCh38
rs1126544 Chr.6: 33069284-33069284 on Build GRCh38
rs148775831 Chr.6: 33069213-33069213 on Build GRCh38
rs375332822 Chr.6: 33069241-33069241 on Build GRCh38
rs538643313 Chr.6: 33069072-33069072 on Build GRCh38
rs111756066 Chr.6: 33069304-33069304 on Build GRCh38
rs11551422 Chr.6: 33069275-33069275 on Build GRCh38
rs538199867 Chr.6: 33069369-33069369 on Build GRCh38
rs377649060 Chr.6: 33069033-33069033 on Build GRCh38
rs753520006 Chr.6: 33069293-33069293 on Build GRCh38
rs760411464 Chr.6: 33069290-33069290 on Build GRCh38
rs2308929 Chr.6: 33069182-33069182 on Build GRCh38
rs745889672 Chr.6: 33069308-33069308 on Build GRCh38
rs924004206 Chr.6: 33069259-33069259 on Build GRCh38
rs201076982 Chr.6: 33069180-33069190 on Build GRCh38
rs752680650 Chr.6: 33069301-33069301 on Build GRCh38
rs41562016 Chr.6: 33069268-33069268 on Build GRCh38
rs776906534 Chr.6: 33069130-33069130 on Build GRCh38
rs773349772 Chr.6: 33069113-33069113 on Build GRCh38
rs753289265 Chr.6: 33069044-33069044 on Build GRCh38
rs143266461 Chr.6: 33069274-33069274 on Build GRCh38
rs781047915 Chr.6: 33069077-33069077 on Build GRCh38
rs765891990 Chr.6: 33069292-33069292 on Build GRCh38
rs900678785 Chr.6: 33069088-33069088 on Build GRCh38
rs746112212 Chr.6: 33069228-33069228 on Build GRCh38
rs535481190 Chr.6: 33069203-33069203 on Build GRCh38
rs3830066 Chr.6: 33069410-33069410 on Build GRCh38
rs763519010 Chr.6: 33069158-33069158 on Build GRCh38
rs374569171 Chr.6: 33069326-33069326 on Build GRCh38
rs386699856 Chr.6: 33069303-33069305 on Build GRCh38
rs916743471 Chr.6: 33069199-33069199 on Build GRCh38
rs762113154 Chr.6: 33069151-33069151 on Build GRCh38
rs115722167 Chr.6: 33069283-33069283 on Build GRCh38
rs769698699 Chr.6: 33068989-33068989 on Build GRCh38
rs759723756 Chr.6: 33069146-33069146 on Build GRCh38
rs3830065 Chr.6: 33069422-33069422 on Build GRCh38
rs1042231 Chr.6: 33069148-33069148 on Build GRCh38
rs199854107 Chr.6: 33069299-33069299 on Build GRCh38
rs748074351 Chr.6: 33069220-33069220 on Build GRCh38
rs772240925 Chr.6: 33069015-33069015 on Build GRCh38
rs139998266 Chr.6: 33069377-33069377 on Build GRCh38
rs67201607 Chr.6: 33069365-33069365 on Build GRCh38
rs1042190 Chr.6: 33069222-33069222 on Build GRCh38
rs941802691 Chr.6: 33069361-33069361 on Build GRCh38
rs868179663 Chr.6: 33069090-33069090 on Build GRCh38
rs73739671 Chr.6: 33069303-33069303 on Build GRCh38
rs757189367 Chr.6: 33069104-33069104 on Build GRCh38
rs781137504 Chr.6: 33068980-33068980 on Build GRCh38
rs775934301 Chr.6: 33069246-33069246 on Build GRCh38
rs190604997 Chr.6: 33069172-33069172 on Build GRCh38
rs773120072 Chr.6: 33069018-33069018 on Build GRCh38
rs762437711 Chr.6: 33069348-33069348 on Build GRCh38
rs138758542 Chr.6: 33069128-33069128 on Build GRCh38
rs1016021044 Chr.6: 33069186-33069186 on Build GRCh38
rs372242232 Chr.6: 33069328-33069328 on Build GRCh38
rs886666238 Chr.6: 33069351-33069351 on Build GRCh38
rs556101955 Chr.6: 33069013-33069013 on Build GRCh38
rs776677427 Chr.6: 33069115-33069115 on Build GRCh38
rs34140193 Chr.6: 33069305-33069305 on Build GRCh38
rs2308930 Chr.6: 33069174-33069174 on Build GRCh38
rs386699855 Chr.6: 33069173-33069174 on Build GRCh38
rs779927534 Chr.6: 33069188-33069188 on Build GRCh38
rs537578968 Chr.6: 33068987-33068987 on Build GRCh38
rs759952653 Chr.6: 33069036-33069036 on Build GRCh38
rs760991360 Chr.6: 33069022-33069022 on Build GRCh38
rs1001726438 Chr.6: 33069405-33069405 on Build GRCh38
rs1035291194 Chr.6: 33069150-33069150 on Build GRCh38
rs370268630 Chr.6: 33069303-33069305 on Build GRCh38
rs2308931 Chr.6: 33069173-33069173 on Build GRCh38
rs750145849 Chr.6: 33069181-33069181 on Build GRCh38
rs764514281 Chr.6: 33069277-33069277 on Build GRCh38
rs764449302 Chr.6: 33069169-33069169 on Build GRCh38
rs745507259 Chr.6: 33069080-33069080 on Build GRCh38
rs2308928 Chr.6: 33069194-33069194 on Build GRCh38
rs541442614 Chr.6: 33069230-33069230 on Build GRCh38
rs113113454 Chr.6: 33069221-33069221 on Build GRCh38
rs1042308 Chr.6: 33069076-33069076 on Build GRCh38
rs756010042 Chr.6: 33069083-33069083 on Build GRCh38
rs1042211 Chr.6: 33069178-33069178 on Build GRCh38
rs576529566 Chr.6: 33069424-33069424 on Build GRCh38

View Full Product Details