Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33086459-33086970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DPB1
Entrez Gene ID
3115
Gene Name
major histocompatibility complex, class II, DP beta 1
Gene Aliases
DPB1, HLA-DP, HLA-DP1B, HLA-DPB
UniGene
Hs.485130
Gene Chromosome Location
Chr.6: 33075926-33089696 on Build GRCh38
Transcripts
NM_002121.5

SNP Detail

SNP ID Chromosome Location
rs1042634 Chr.6: 33086782-33086782 on Build GRCh38
rs1042511 Chr.6: 33086636-33086636 on Build GRCh38
rs796725249 Chr.6: 33086636-33086637 on Build GRCh38
rs931 Chr.6: 33086773-33086773 on Build GRCh38
rs928 Chr.6: 33086775-33086775 on Build GRCh38
rs9277532 Chr.6: 33086934-33086934 on Build GRCh38
rs1042508 Chr.6: 33086633-33086633 on Build GRCh38
rs1126723 Chr.6: 33086554-33086554 on Build GRCh38
rs553488718 Chr.6: 33086522-33086522 on Build GRCh38
rs386699893 Chr.6: 33086576-33086578 on Build GRCh38
rs1042507 Chr.6: 33086627-33086627 on Build GRCh38
rs3749984 Chr.6: 33086597-33086597 on Build GRCh38
rs909180587 Chr.6: 33086579-33086579 on Build GRCh38
rs1014 Chr.6: 33086832-33086832 on Build GRCh38
rs935 Chr.6: 33086802-33086802 on Build GRCh38
rs186748908 Chr.6: 33086778-33086778 on Build GRCh38
rs930 Chr.6: 33086879-33086879 on Build GRCh38
rs1053222732 Chr.6: 33086754-33086754 on Build GRCh38
rs1042502 Chr.6: 33086623-33086623 on Build GRCh38
rs532241225 Chr.6: 33086583-33086583 on Build GRCh38
rs771322584 Chr.6: 33086503-33086505 on Build GRCh38
rs6760 Chr.6: 33086601-33086601 on Build GRCh38
rs143935385 Chr.6: 33086899-33086899 on Build GRCh38
rs564159821 Chr.6: 33086581-33086581 on Build GRCh38
rs386699891 Chr.6: 33086503-33086504 on Build GRCh38
rs114068468 Chr.6: 33086686-33086686 on Build GRCh38
rs1016898607 Chr.6: 33086781-33086781 on Build GRCh38
rs1005852121 Chr.6: 33086734-33086734 on Build GRCh38
rs9277518 Chr.6: 33086525-33086525 on Build GRCh38
rs144401610 Chr.6: 33086823-33086827 on Build GRCh38
rs3749985 Chr.6: 33086656-33086656 on Build GRCh38
rs933 Chr.6: 33086818-33086818 on Build GRCh38
rs9277517 Chr.6: 33086504-33086504 on Build GRCh38
rs1009941953 Chr.6: 33086511-33086511 on Build GRCh38
rs1042488 Chr.6: 33086604-33086604 on Build GRCh38
rs938818013 Chr.6: 33086501-33086501 on Build GRCh38
rs756813500 Chr.6: 33086531-33086531 on Build GRCh38
rs1126719 Chr.6: 33086548-33086548 on Build GRCh38
rs9277533 Chr.6: 33086944-33086944 on Build GRCh38
rs201032790 Chr.6: 33086491-33086494 on Build GRCh38
rs1007029397 Chr.6: 33086534-33086534 on Build GRCh38
rs759169647 Chr.6: 33086696-33086696 on Build GRCh38
rs1042448 Chr.6: 33086565-33086565 on Build GRCh38
rs927395705 Chr.6: 33086776-33086779 on Build GRCh38
rs9277530 Chr.6: 33086906-33086906 on Build GRCh38
rs998251307 Chr.6: 33086765-33086765 on Build GRCh38
rs545910027 Chr.6: 33086771-33086771 on Build GRCh38
rs1043734780 Chr.6: 33086817-33086817 on Build GRCh38
rs946360312 Chr.6: 33086536-33086536 on Build GRCh38
rs3833672 Chr.6: 33086824-33086828 on Build GRCh38
rs9277522 Chr.6: 33086576-33086576 on Build GRCh38
rs11551420 Chr.6: 33086705-33086705 on Build GRCh38
rs34087328 Chr.6: 33086644-33086644 on Build GRCh38
rs1042544 Chr.6: 33086680-33086680 on Build GRCh38
rs901318338 Chr.6: 33086757-33086757 on Build GRCh38
rs1042497 Chr.6: 33086609-33086609 on Build GRCh38
rs9277523 Chr.6: 33086578-33086578 on Build GRCh38
rs386699892 Chr.6: 33086548-33086578 on Build GRCh38
rs929 Chr.6: 33086842-33086842 on Build GRCh38
rs386699894 Chr.6: 33086596-33086637 on Build GRCh38
rs1042644 Chr.6: 33086742-33086742 on Build GRCh38
rs367833675 Chr.6: 33086850-33086850 on Build GRCh38
rs1042467 Chr.6: 33086596-33086596 on Build GRCh38
rs9277531 Chr.6: 33086910-33086910 on Build GRCh38
rs9277516 Chr.6: 33086503-33086503 on Build GRCh38
rs9277515 Chr.6: 33086491-33086491 on Build GRCh38
rs934 Chr.6: 33086882-33086882 on Build GRCh38
rs932 Chr.6: 33086809-33086809 on Build GRCh38
rs529885638 Chr.6: 33086800-33086800 on Build GRCh38
rs568686979 Chr.6: 33086893-33086893 on Build GRCh38
rs386699895 Chr.6: 33086773-33086775 on Build GRCh38
rs559452923 Chr.6: 33086600-33086600 on Build GRCh38
rs980622169 Chr.6: 33086595-33086595 on Build GRCh38
rs1042516 Chr.6: 33086637-33086637 on Build GRCh38
rs386699896 Chr.6: 33086879-33086944 on Build GRCh38
rs9277529 Chr.6: 33086898-33086898 on Build GRCh38

View Full Product Details