Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33087194-33087692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DPB1
Entrez Gene ID
3115
Gene Name
major histocompatibility complex, class II, DP beta 1
Gene Aliases
DPB1, HLA-DP, HLA-DP1B, HLA-DPB
UniGene
Hs.485130
Gene Chromosome Location
Chr.6: 33075926-33089696 on Build GRCh38
Transcripts
NM_002121.5

SNP Detail

SNP ID Chromosome Location
rs1029746008 Chr.6: 33087615-33087615 on Build GRCh38
rs386699897 Chr.6: 33087270-33087302 on Build GRCh38
rs551251831 Chr.6: 33087658-33087658 on Build GRCh38
rs67703005 Chr.6: 33087409-33087410 on Build GRCh38
rs527319816 Chr.6: 33087607-33087607 on Build GRCh38
rs9277539 Chr.6: 33087302-33087302 on Build GRCh38
rs997143281 Chr.6: 33087652-33087652 on Build GRCh38
rs1020242108 Chr.6: 33087385-33087385 on Build GRCh38
rs903282438 Chr.6: 33087391-33087391 on Build GRCh38
rs908297401 Chr.6: 33087286-33087286 on Build GRCh38
rs9277541 Chr.6: 33087381-33087381 on Build GRCh38
rs986041352 Chr.6: 33087487-33087487 on Build GRCh38
rs908180254 Chr.6: 33087494-33087494 on Build GRCh38
rs3097678 Chr.6: 33087335-33087335 on Build GRCh38
rs9277542 Chr.6: 33087470-33087470 on Build GRCh38
rs534325583 Chr.6: 33087428-33087428 on Build GRCh38
rs1000254634 Chr.6: 33087415-33087415 on Build GRCh38
rs3097679 Chr.6: 33087533-33087533 on Build GRCh38
rs962791087 Chr.6: 33087502-33087502 on Build GRCh38
rs896404855 Chr.6: 33087586-33087586 on Build GRCh38
rs1023899754 Chr.6: 33087609-33087609 on Build GRCh38
rs73740309 Chr.6: 33087511-33087511 on Build GRCh38
rs929952325 Chr.6: 33087572-33087572 on Build GRCh38
rs996967609 Chr.6: 33087435-33087436 on Build GRCh38
rs375342014 Chr.6: 33087450-33087450 on Build GRCh38
rs1038630373 Chr.6: 33087319-33087319 on Build GRCh38
rs933697811 Chr.6: 33087248-33087248 on Build GRCh38
rs747535022 Chr.6: 33087285-33087285 on Build GRCh38
rs897188938 Chr.6: 33087556-33087556 on Build GRCh38
rs765293588 Chr.6: 33087612-33087612 on Build GRCh38
rs772533259 Chr.6: 33087222-33087222 on Build GRCh38
rs9277540 Chr.6: 33087346-33087346 on Build GRCh38
rs1014301467 Chr.6: 33087384-33087384 on Build GRCh38
rs568432283 Chr.6: 33087627-33087627 on Build GRCh38
rs9501257 Chr.6: 33087578-33087578 on Build GRCh38
rs551656344 Chr.6: 33087618-33087618 on Build GRCh38
rs922838819 Chr.6: 33087246-33087246 on Build GRCh38
rs9501258 Chr.6: 33087583-33087583 on Build GRCh38
rs557330813 Chr.6: 33087512-33087512 on Build GRCh38
rs975664303 Chr.6: 33087245-33087245 on Build GRCh38
rs1033497505 Chr.6: 33087419-33087420 on Build GRCh38
rs775933556 Chr.6: 33087269-33087313 on Build GRCh38
rs934206700 Chr.6: 33087486-33087486 on Build GRCh38
rs9280312 Chr.6: 33087420-33087420 on Build GRCh38
rs9277549 Chr.6: 33087642-33087642 on Build GRCh38
rs964266931 Chr.6: 33087432-33087432 on Build GRCh38
rs539592780 Chr.6: 33087350-33087350 on Build GRCh38
rs540086241 Chr.6: 33087505-33087505 on Build GRCh38
rs541802787 Chr.6: 33087510-33087511 on Build GRCh38
rs9501255 Chr.6: 33087321-33087321 on Build GRCh38
rs9277537 Chr.6: 33087232-33087232 on Build GRCh38
rs9280313 Chr.6: 33087505-33087506 on Build GRCh38
rs367734650 Chr.6: 33087505-33087505 on Build GRCh38
rs956413893 Chr.6: 33087644-33087644 on Build GRCh38
rs1054584402 Chr.6: 33087587-33087587 on Build GRCh38
rs9277538 Chr.6: 33087270-33087270 on Build GRCh38
rs9277544 Chr.6: 33087531-33087531 on Build GRCh38
rs1012083285 Chr.6: 33087296-33087296 on Build GRCh38
rs72500564 Chr.6: 33087420-33087420 on Build GRCh38
rs979173499 Chr.6: 33087329-33087329 on Build GRCh38
rs9277545 Chr.6: 33087546-33087546 on Build GRCh38
rs9277543 Chr.6: 33087517-33087517 on Build GRCh38
rs962430970 Chr.6: 33087662-33087662 on Build GRCh38
rs9277548 Chr.6: 33087613-33087613 on Build GRCh38
rs540330554 Chr.6: 33087555-33087555 on Build GRCh38
rs1022186492 Chr.6: 33087251-33087251 on Build GRCh38
rs967909549 Chr.6: 33087259-33087259 on Build GRCh38
rs9277546 Chr.6: 33087569-33087569 on Build GRCh38
rs139610109 Chr.6: 33087503-33087505 on Build GRCh38
rs372571124 Chr.6: 33087257-33087257 on Build GRCh38
rs200599840 Chr.6: 33087663-33087663 on Build GRCh38
rs9277547 Chr.6: 33087590-33087590 on Build GRCh38

View Full Product Details