Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33087591-33088100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DPB1
Entrez Gene ID
3115
Gene Name
major histocompatibility complex, class II, DP beta 1
Gene Aliases
DPB1, HLA-DP, HLA-DP1B, HLA-DPB
UniGene
Hs.485130
Gene Chromosome Location
Chr.6: 33075926-33089696 on Build GRCh38
Transcripts
NM_002121.5

SNP Detail

SNP ID Chromosome Location
rs1029746008 Chr.6: 33087615-33087615 on Build GRCh38
rs1029972087 Chr.6: 33088042-33088042 on Build GRCh38
rs899929817 Chr.6: 33087812-33087812 on Build GRCh38
rs551251831 Chr.6: 33087658-33087658 on Build GRCh38
rs386699898 Chr.6: 33087761-33087774 on Build GRCh38
rs921235795 Chr.6: 33088064-33088064 on Build GRCh38
rs9277550 Chr.6: 33087710-33087710 on Build GRCh38
rs959079666 Chr.6: 33087716-33087716 on Build GRCh38
rs997143281 Chr.6: 33087652-33087652 on Build GRCh38
rs9277555 Chr.6: 33087828-33087828 on Build GRCh38
rs947981553 Chr.6: 33087934-33087934 on Build GRCh38
rs978321035 Chr.6: 33088026-33088026 on Build GRCh38
rs539758150 Chr.6: 33087725-33087725 on Build GRCh38
rs995192912 Chr.6: 33087926-33087926 on Build GRCh38
rs369472644 Chr.6: 33087946-33087947 on Build GRCh38
rs58245331 Chr.6: 33088068-33088071 on Build GRCh38
rs146520390 Chr.6: 33087694-33087694 on Build GRCh38
rs9277551 Chr.6: 33087717-33087717 on Build GRCh38
rs1056484716 Chr.6: 33087898-33087898 on Build GRCh38
rs1051218897 Chr.6: 33087829-33087829 on Build GRCh38
rs1000705128 Chr.6: 33087974-33087974 on Build GRCh38
rs3128963 Chr.6: 33088003-33088003 on Build GRCh38
rs966571925 Chr.6: 33088023-33088023 on Build GRCh38
rs1044969105 Chr.6: 33087938-33087938 on Build GRCh38
rs192449683 Chr.6: 33087948-33087948 on Build GRCh38
rs202073495 Chr.6: 33088065-33088068 on Build GRCh38
rs765293588 Chr.6: 33087612-33087612 on Build GRCh38
rs9277552 Chr.6: 33087724-33087724 on Build GRCh38
rs533414261 Chr.6: 33088008-33088008 on Build GRCh38
rs112119417 Chr.6: 33087722-33087722 on Build GRCh38
rs777184976 Chr.6: 33087877-33087877 on Build GRCh38
rs112170964 Chr.6: 33087943-33087944 on Build GRCh38
rs148969528 Chr.6: 33087888-33087890 on Build GRCh38
rs568432283 Chr.6: 33087627-33087627 on Build GRCh38
rs551656344 Chr.6: 33087618-33087618 on Build GRCh38
rs892224229 Chr.6: 33088040-33088040 on Build GRCh38
rs201709182 Chr.6: 33088067-33088068 on Build GRCh38
rs376017737 Chr.6: 33087892-33087894 on Build GRCh38
rs9277549 Chr.6: 33087642-33087642 on Build GRCh38
rs917894146 Chr.6: 33087913-33087913 on Build GRCh38
rs9277553 Chr.6: 33087739-33087739 on Build GRCh38
rs974596808 Chr.6: 33087786-33087786 on Build GRCh38
rs925231231 Chr.6: 33088035-33088035 on Build GRCh38
rs956413893 Chr.6: 33087644-33087644 on Build GRCh38
rs898737111 Chr.6: 33087919-33087919 on Build GRCh38
rs9501259 Chr.6: 33087774-33087774 on Build GRCh38
rs567910044 Chr.6: 33087951-33087951 on Build GRCh38
rs5875436 Chr.6: 33088067-33088070 on Build GRCh38
rs936049650 Chr.6: 33087752-33087752 on Build GRCh38
rs3128964 Chr.6: 33088041-33088041 on Build GRCh38
rs77339491 Chr.6: 33088068-33088068 on Build GRCh38
rs962430970 Chr.6: 33087662-33087662 on Build GRCh38
rs9277548 Chr.6: 33087613-33087613 on Build GRCh38
rs991935488 Chr.6: 33088001-33088001 on Build GRCh38
rs9277554 Chr.6: 33087761-33087761 on Build GRCh38
rs60460211 Chr.6: 33087944-33087945 on Build GRCh38
rs990185661 Chr.6: 33088043-33088043 on Build GRCh38
rs200599840 Chr.6: 33087663-33087663 on Build GRCh38

View Full Product Details