Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33204468-33204978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B8
Entrez Gene ID
7923
Gene Name
hydroxysteroid 17-beta dehydrogenase 8
Gene Aliases
D6S2245E, FABG, FABGL, H2-KE6, HKE6, KE6, RING2, SDR30C1, dJ1033B10.9
UniGene
Hs.415058
Gene Chromosome Location
Chr.6: 33204637-33206831 on Build GRCh38
Transcripts
NM_014234.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6425884 c.128T>C p.L43P Chr.6: 33204977-33204977 on Build GRCh38
7130804 c.10C>G p.Q4E Chr.6: 33204678-33204678 on Build GRCh38
6038919 c.110C>A p.T37N Chr.6: 33204959-33204959 on Build GRCh38
4645569 c.102G>T p.E34D Chr.6: 33204951-33204951 on Build GRCh38
1621508 c.51A>G p.T17T Chr.6: 33204719-33204719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748042744 Chr.6: 33204942-33204942 on Build GRCh38
rs759984516 Chr.6: 33204705-33204705 on Build GRCh38
rs926002424 Chr.6: 33204828-33204828 on Build GRCh38
rs1036429353 Chr.6: 33204528-33204528 on Build GRCh38
rs975528528 Chr.6: 33204847-33204847 on Build GRCh38
rs1036578513 Chr.6: 33204736-33204736 on Build GRCh38
rs754302023 Chr.6: 33204667-33204667 on Build GRCh38
rs956937028 Chr.6: 33204909-33204909 on Build GRCh38
rs759296612 Chr.6: 33204648-33204648 on Build GRCh38
rs769773247 Chr.6: 33204654-33204654 on Build GRCh38
rs759784341 Chr.6: 33204771-33204771 on Build GRCh38
rs574075966 Chr.6: 33204697-33204697 on Build GRCh38
rs953455532 Chr.6: 33204937-33204937 on Build GRCh38
rs376172570 Chr.6: 33204657-33204657 on Build GRCh38
rs1051344765 Chr.6: 33204863-33204863 on Build GRCh38
rs368394952 Chr.6: 33204960-33204960 on Build GRCh38
rs1008591978 Chr.6: 33204509-33204509 on Build GRCh38
rs776370698 Chr.6: 33204646-33204646 on Build GRCh38
rs756749998 Chr.6: 33204737-33204737 on Build GRCh38
rs142093440 Chr.6: 33204669-33204669 on Build GRCh38
rs772470105 Chr.6: 33204637-33204637 on Build GRCh38
rs771070307 Chr.6: 33204763-33204763 on Build GRCh38
rs768158059 Chr.6: 33204682-33204682 on Build GRCh38
rs531064075 Chr.6: 33204752-33204752 on Build GRCh38
rs769508877 Chr.6: 33204680-33204680 on Build GRCh38
rs372381285 Chr.6: 33204495-33204496 on Build GRCh38
rs940331753 Chr.6: 33204883-33204883 on Build GRCh38
rs926068012 Chr.6: 33204652-33204652 on Build GRCh38
rs370135457 Chr.6: 33204580-33204580 on Build GRCh38
rs557942898 Chr.6: 33204621-33204622 on Build GRCh38
rs754724398 Chr.6: 33204938-33204938 on Build GRCh38
rs912179628 Chr.6: 33204715-33204715 on Build GRCh38
rs45621635 Chr.6: 33204859-33204859 on Build GRCh38
rs750104417 Chr.6: 33204880-33204880 on Build GRCh38
rs994999284 Chr.6: 33204544-33204544 on Build GRCh38
rs898887165 Chr.6: 33204926-33204926 on Build GRCh38
rs915354788 Chr.6: 33204932-33204932 on Build GRCh38
rs150495100 Chr.6: 33204665-33204665 on Build GRCh38
rs776306585 Chr.6: 33204681-33204688 on Build GRCh38
rs753167802 Chr.6: 33204719-33204719 on Build GRCh38
rs569282496 Chr.6: 33204939-33204939 on Build GRCh38
rs761801597 Chr.6: 33204659-33204659 on Build GRCh38
rs749005101 Chr.6: 33204746-33204746 on Build GRCh38
rs138620921 Chr.6: 33204702-33204702 on Build GRCh38
rs946064697 Chr.6: 33204610-33204610 on Build GRCh38
rs112105996 Chr.6: 33204741-33204741 on Build GRCh38
rs372709810 Chr.6: 33204497-33204497 on Build GRCh38
rs992801004 Chr.6: 33204574-33204574 on Build GRCh38
rs765641312 Chr.6: 33204709-33204709 on Build GRCh38
rs1048861982 Chr.6: 33204791-33204791 on Build GRCh38
rs890689189 Chr.6: 33204503-33204503 on Build GRCh38
rs764481891 Chr.6: 33204729-33204729 on Build GRCh38
rs777007739 Chr.6: 33204768-33204768 on Build GRCh38
rs547035463 Chr.6: 33204609-33204609 on Build GRCh38
rs558676405 Chr.6: 33204664-33204664 on Build GRCh38
rs371052687 Chr.6: 33204853-33204853 on Build GRCh38
rs894213651 Chr.6: 33204809-33204809 on Build GRCh38
rs775400099 Chr.6: 33204865-33204865 on Build GRCh38
rs759026575 Chr.6: 33204689-33204694 on Build GRCh38
rs771158268 Chr.6: 33204945-33204945 on Build GRCh38
rs751132851 Chr.6: 33204730-33204730 on Build GRCh38
rs932120773 Chr.6: 33204492-33204492 on Build GRCh38
rs1014155326 Chr.6: 33204720-33204720 on Build GRCh38
rs963200127 Chr.6: 33204494-33204494 on Build GRCh38
rs554888772 Chr.6: 33204556-33204556 on Build GRCh38
rs749196565 Chr.6: 33204686-33204686 on Build GRCh38
rs770521610 Chr.6: 33204680-33204696 on Build GRCh38
rs370208017 Chr.6: 33204499-33204500 on Build GRCh38
rs114699053 Chr.6: 33204529-33204529 on Build GRCh38
rs1034199255 Chr.6: 33204703-33204703 on Build GRCh38
rs774140723 Chr.6: 33204755-33204755 on Build GRCh38
rs1034228632 Chr.6: 33204524-33204524 on Build GRCh38
rs772071122 Chr.6: 33204690-33204690 on Build GRCh38
rs563634522 Chr.6: 33204738-33204738 on Build GRCh38
rs773380800 Chr.6: 33204691-33204691 on Build GRCh38
rs761774157 Chr.6: 33204796-33204796 on Build GRCh38
rs752007306 Chr.6: 33204866-33204866 on Build GRCh38
rs540783107 Chr.6: 33204817-33204817 on Build GRCh38
rs747151419 Chr.6: 33204758-33204758 on Build GRCh38
rs767575152 Chr.6: 33204660-33204660 on Build GRCh38
rs958591184 Chr.6: 33204708-33204708 on Build GRCh38
rs1012786864 Chr.6: 33204817-33204817 on Build GRCh38
rs552817372 Chr.6: 33204670-33204670 on Build GRCh38
rs201389008 Chr.6: 33204636-33204636 on Build GRCh38
rs371077739 Chr.6: 33204672-33204672 on Build GRCh38
rs564671066 Chr.6: 33204858-33204858 on Build GRCh38
rs187291206 Chr.6: 33204840-33204840 on Build GRCh38
rs1036445906 Chr.6: 33204906-33204906 on Build GRCh38
rs145971755 Chr.6: 33204675-33204675 on Build GRCh38
rs758903333 Chr.6: 33204728-33204728 on Build GRCh38
rs766764231 Chr.6: 33204700-33204700 on Build GRCh38
rs779854220 Chr.6: 33204744-33204744 on Build GRCh38
rs766962775 Chr.6: 33204875-33204875 on Build GRCh38
rs373903352 Chr.6: 33204673-33204673 on Build GRCh38
rs971923248 Chr.6: 33204851-33204866 on Build GRCh38
rs546797834 Chr.6: 33204493-33204493 on Build GRCh38
rs202140618 Chr.6: 33204928-33204928 on Build GRCh38
rs547049883 Chr.6: 33204862-33204862 on Build GRCh38
rs746618614 Chr.6: 33204634-33204634 on Build GRCh38
rs773332438 Chr.6: 33204870-33204870 on Build GRCh38
rs370759993 Chr.6: 33204921-33204921 on Build GRCh38
rs764787302 Chr.6: 33204725-33204726 on Build GRCh38
rs894301835 Chr.6: 33204956-33204956 on Build GRCh38
rs780803866 Chr.6: 33204677-33204677 on Build GRCh38
rs568602675 Chr.6: 33204902-33204902 on Build GRCh38

View Full Product Details