Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59642005-59642555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2A2
Entrez Gene ID
2958
Gene Name
general transcription factor IIA subunit 2
Gene Aliases
HsT18745, T18745, TF2A2, TFIIA, TFIIA-12, TFIIA-gamma, TFIIAS
UniGene
Hs.512934
Gene Chromosome Location
Chr.15: 59638062-59657541 on Build GRCh38
Transcripts
NM_001320929.1 NM_001320930.1 NM_004492.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6463988 c.238G>A p.E80K Chr.15: 59642202-59642202 on Build GRCh38
4531458 c.178G>A p.G60S Chr.15: 59642262-59642262 on Build GRCh38
470900 c.270G>T p.V90V Chr.15: 59642170-59642170 on Build GRCh38
88048 c.191C>T p.T64M Chr.15: 59642249-59642249 on Build GRCh38
6255901 c.191C>A p.T64K Chr.15: 59642249-59642249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1802673 Chr.15: 59642215-59642215 on Build GRCh38
rs184691175 Chr.15: 59642058-59642058 on Build GRCh38
rs771681416 Chr.15: 59642290-59642290 on Build GRCh38
rs540777806 Chr.15: 59642288-59642288 on Build GRCh38
rs758978144 Chr.15: 59642087-59642087 on Build GRCh38
rs1020159364 Chr.15: 59642062-59642062 on Build GRCh38
rs897053039 Chr.15: 59642344-59642344 on Build GRCh38
rs754038295 Chr.15: 59642354-59642354 on Build GRCh38
rs775396492 Chr.15: 59642309-59642312 on Build GRCh38
rs780232285 Chr.15: 59642114-59642114 on Build GRCh38
rs149519150 Chr.15: 59642177-59642177 on Build GRCh38
rs373021830 Chr.15: 59642289-59642289 on Build GRCh38
rs756690646 Chr.15: 59642272-59642273 on Build GRCh38
rs760199278 Chr.15: 59642299-59642299 on Build GRCh38
rs897105744 Chr.15: 59642038-59642038 on Build GRCh38
rs755834900 Chr.15: 59642184-59642184 on Build GRCh38
rs138437737 Chr.15: 59642312-59642312 on Build GRCh38
rs1010392577 Chr.15: 59642468-59642468 on Build GRCh38
rs143067220 Chr.15: 59642127-59642127 on Build GRCh38
rs1036848545 Chr.15: 59642325-59642325 on Build GRCh38
rs1020483435 Chr.15: 59642470-59642470 on Build GRCh38
rs539151913 Chr.15: 59642093-59642093 on Build GRCh38
rs748569475 Chr.15: 59642129-59642130 on Build GRCh38
rs748840894 Chr.15: 59642036-59642036 on Build GRCh38
rs536931482 Chr.15: 59642358-59642358 on Build GRCh38
rs565160928 Chr.15: 59642296-59642296 on Build GRCh38
rs745576490 Chr.15: 59642306-59642307 on Build GRCh38
rs546788797 Chr.15: 59642491-59642491 on Build GRCh38
rs773933529 Chr.15: 59642146-59642146 on Build GRCh38
rs377176564 Chr.15: 59642258-59642258 on Build GRCh38
rs780063380 Chr.15: 59642088-59642088 on Build GRCh38
rs968931599 Chr.15: 59642480-59642480 on Build GRCh38
rs1802671 Chr.15: 59642191-59642191 on Build GRCh38
rs370188854 Chr.15: 59642248-59642248 on Build GRCh38
rs1007009495 Chr.15: 59642444-59642444 on Build GRCh38
rs989014579 Chr.15: 59642106-59642106 on Build GRCh38
rs748659438 Chr.15: 59642301-59642301 on Build GRCh38
rs766440312 Chr.15: 59642128-59642128 on Build GRCh38
rs748287278 Chr.15: 59642249-59642249 on Build GRCh38
rs184976775 Chr.15: 59642417-59642417 on Build GRCh38
rs774088158 Chr.15: 59642303-59642304 on Build GRCh38
rs148247017 Chr.15: 59642046-59642046 on Build GRCh38
rs1013876363 Chr.15: 59642036-59642036 on Build GRCh38
rs372745095 Chr.15: 59642256-59642256 on Build GRCh38
rs750825411 Chr.15: 59642199-59642199 on Build GRCh38
rs189450115 Chr.15: 59642122-59642122 on Build GRCh38
rs748766489 Chr.15: 59642298-59642298 on Build GRCh38
rs532826446 Chr.15: 59642297-59642297 on Build GRCh38
rs771760455 Chr.15: 59642307-59642308 on Build GRCh38
rs1019354592 Chr.15: 59642453-59642453 on Build GRCh38
rs953898297 Chr.15: 59642056-59642056 on Build GRCh38
rs778160675 Chr.15: 59642297-59642298 on Build GRCh38
rs567509913 Chr.15: 59642439-59642439 on Build GRCh38
rs770569483 Chr.15: 59642303-59642305 on Build GRCh38
rs949233799 Chr.15: 59642320-59642320 on Build GRCh38
rs765482793 Chr.15: 59642196-59642196 on Build GRCh38
rs759114302 Chr.15: 59642156-59642156 on Build GRCh38
rs34972010 Chr.15: 59642438-59642439 on Build GRCh38
rs548742979 Chr.15: 59642435-59642435 on Build GRCh38
rs1048013567 Chr.15: 59642372-59642372 on Build GRCh38
rs561619087 Chr.15: 59642236-59642236 on Build GRCh38
rs1026624441 Chr.15: 59642047-59642047 on Build GRCh38
rs780540387 Chr.15: 59642217-59642217 on Build GRCh38
rs767194568 Chr.15: 59642174-59642174 on Build GRCh38
rs192601744 Chr.15: 59642286-59642286 on Build GRCh38
rs749115140 Chr.15: 59642284-59642284 on Build GRCh38
rs768797460 Chr.15: 59642096-59642096 on Build GRCh38
rs563069475 Chr.15: 59642369-59642369 on Build GRCh38
rs113175335 Chr.15: 59642109-59642109 on Build GRCh38
rs763824338 Chr.15: 59642311-59642311 on Build GRCh38
rs979017076 Chr.15: 59642504-59642504 on Build GRCh38
rs776824491 Chr.15: 59642105-59642105 on Build GRCh38
rs112440432 Chr.15: 59642098-59642098 on Build GRCh38
rs3736104 Chr.15: 59642467-59642467 on Build GRCh38
rs755033629 Chr.15: 59642239-59642239 on Build GRCh38
rs143116097 Chr.15: 59642037-59642037 on Build GRCh38

View Full Product Details