Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123659398-123659886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2H3
Entrez Gene ID
2967
Gene Name
general transcription factor IIH subunit 3
Gene Aliases
BTF2, P34, TFB4, TFIIH
UniGene
Hs.355348
Gene Chromosome Location
Chr.12: 123633739-123662604 on Build GRCh38
Transcripts
NM_001271866.1 NM_001271867.1 NM_001271868.1 NM_001516.4 XM_017019228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1677455 c.644T>C p.L215P Chr.12: 123659544-123659544 on Build GRCh38
430621 c.685T>A p.W229R Chr.12: 123659795-123659795 on Build GRCh38
5840443 c.713G>A p.R238K Chr.12: 123659823-123659823 on Build GRCh38
6577797 c.686G>A p.W229* Chr.12: 123659796-123659796 on Build GRCh38
6000363 c.776G>A p.R259Q Chr.12: 123659886-123659886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769919689 Chr.12: 123659430-123659430 on Build GRCh38
rs114301796 Chr.12: 123659606-123659606 on Build GRCh38
rs532345098 Chr.12: 123659722-123659722 on Build GRCh38
rs546441397 Chr.12: 123659711-123659711 on Build GRCh38
rs768120496 Chr.12: 123659849-123659849 on Build GRCh38
rs766161754 Chr.12: 123659810-123659810 on Build GRCh38
rs749680653 Chr.12: 123659433-123659433 on Build GRCh38
rs534462205 Chr.12: 123659434-123659434 on Build GRCh38
rs568985227 Chr.12: 123659752-123659752 on Build GRCh38
rs772765644 Chr.12: 123659571-123659571 on Build GRCh38
rs762429375 Chr.12: 123659438-123659438 on Build GRCh38
rs773853949 Chr.12: 123659574-123659574 on Build GRCh38
rs756213897 Chr.12: 123659501-123659501 on Build GRCh38
rs375205709 Chr.12: 123659777-123659777 on Build GRCh38
rs369722963 Chr.12: 123659860-123659860 on Build GRCh38
rs752367380 Chr.12: 123659473-123659473 on Build GRCh38
rs779216405 Chr.12: 123659621-123659621 on Build GRCh38
rs748592594 Chr.12: 123659623-123659623 on Build GRCh38
rs775766482 Chr.12: 123659431-123659431 on Build GRCh38
rs370185510 Chr.12: 123659675-123659675 on Build GRCh38
rs11573003 Chr.12: 123659636-123659636 on Build GRCh38
rs1051793 Chr.12: 123659848-123659848 on Build GRCh38
rs776283199 Chr.12: 123659453-123659453 on Build GRCh38
rs748453785 Chr.12: 123659564-123659564 on Build GRCh38
rs768357583 Chr.12: 123659764-123659764 on Build GRCh38
rs953200588 Chr.12: 123659697-123659697 on Build GRCh38
rs763670748 Chr.12: 123659600-123659600 on Build GRCh38
rs764868061 Chr.12: 123659471-123659471 on Build GRCh38
rs755253319 Chr.12: 123659615-123659615 on Build GRCh38
rs139176876 Chr.12: 123659537-123659537 on Build GRCh38
rs559255846 Chr.12: 123659858-123659858 on Build GRCh38
rs761632902 Chr.12: 123659773-123659773 on Build GRCh38
rs548323779 Chr.12: 123659839-123659839 on Build GRCh38
rs146276537 Chr.12: 123659529-123659529 on Build GRCh38
rs778197124 Chr.12: 123659750-123659750 on Build GRCh38
rs759076180 Chr.12: 123659457-123659457 on Build GRCh38
rs566450540 Chr.12: 123659846-123659846 on Build GRCh38
rs772030945 Chr.12: 123659775-123659778 on Build GRCh38
rs753817946 Chr.12: 123659613-123659613 on Build GRCh38
rs765597746 Chr.12: 123659820-123659820 on Build GRCh38
rs780200564 Chr.12: 123659612-123659612 on Build GRCh38
rs751371333 Chr.12: 123659601-123659601 on Build GRCh38
rs567907024 Chr.12: 123659809-123659809 on Build GRCh38
rs772830165 Chr.12: 123659791-123659791 on Build GRCh38
rs747273795 Chr.12: 123659843-123659843 on Build GRCh38
rs768094485 Chr.12: 123659446-123659446 on Build GRCh38
rs776674162 Chr.12: 123659437-123659437 on Build GRCh38
rs11573002 Chr.12: 123659603-123659603 on Build GRCh38
rs11573005 Chr.12: 123659732-123659732 on Build GRCh38
rs762314613 Chr.12: 123659761-123659761 on Build GRCh38
rs750524688 Chr.12: 123659500-123659500 on Build GRCh38
rs749504129 Chr.12: 123659527-123659527 on Build GRCh38
rs903426090 Chr.12: 123659553-123659553 on Build GRCh38
rs182585779 Chr.12: 123659429-123659429 on Build GRCh38
rs778236454 Chr.12: 123659840-123659840 on Build GRCh38
rs894759559 Chr.12: 123659825-123659825 on Build GRCh38
rs771584402 Chr.12: 123659856-123659856 on Build GRCh38
rs202070887 Chr.12: 123659592-123659592 on Build GRCh38
rs11573004 Chr.12: 123659642-123659642 on Build GRCh38
rs762710324 Chr.12: 123659478-123659478 on Build GRCh38
rs1027259740 Chr.12: 123659619-123659619 on Build GRCh38
rs1042077070 Chr.12: 123659733-123659733 on Build GRCh38
rs781051455 Chr.12: 123659498-123659498 on Build GRCh38
rs936228264 Chr.12: 123659808-123659808 on Build GRCh38
rs559316478 Chr.12: 123659482-123659482 on Build GRCh38
rs774036529 Chr.12: 123659853-123659853 on Build GRCh38
rs371920712 Chr.12: 123659756-123659756 on Build GRCh38
rs57744548 Chr.12: 123659787-123659787 on Build GRCh38
rs971215496 Chr.12: 123659704-123659704 on Build GRCh38
rs374696426 Chr.12: 123659483-123659483 on Build GRCh38
rs564843649 Chr.12: 123659712-123659712 on Build GRCh38
rs149957130 Chr.12: 123659582-123659582 on Build GRCh38
rs757191863 Chr.12: 123659492-123659492 on Build GRCh38
rs527701892 Chr.12: 123659686-123659686 on Build GRCh38
rs773986112 Chr.12: 123659769-123659769 on Build GRCh38
rs752946645 Chr.12: 123659829-123659829 on Build GRCh38
rs367723135 Chr.12: 123659585-123659585 on Build GRCh38
rs11573006 Chr.12: 123659812-123659812 on Build GRCh38
rs781539845 Chr.12: 123659847-123659847 on Build GRCh38
rs920393015 Chr.12: 123659670-123659670 on Build GRCh38
rs780029936 Chr.12: 123659774-123659775 on Build GRCh38
rs973360090 Chr.12: 123659652-123659652 on Build GRCh38
rs1024491445 Chr.12: 123659698-123659698 on Build GRCh38
rs760697637 Chr.12: 123659800-123659800 on Build GRCh38
rs780178599 Chr.12: 123659524-123659524 on Build GRCh38
rs375224225 Chr.12: 123659604-123659604 on Build GRCh38
rs771302619 Chr.12: 123659754-123659754 on Build GRCh38
rs759011429 Chr.12: 123659577-123659577 on Build GRCh38
rs1014628082 Chr.12: 123659616-123659616 on Build GRCh38
rs755106348 Chr.12: 123659813-123659813 on Build GRCh38

View Full Product Details