Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75634015-75634575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KARS
Entrez Gene ID
3735
Gene Name
lysyl-tRNA synthetase
Gene Aliases
CMTRIB, DFNB89, KARS1, KARS2, KRS
UniGene
Hs.3100
Gene Chromosome Location
Chr.16: 75627724-75647687 on Build GRCh38
Transcripts
NM_001130089.1 NM_005548.2 XM_017023217.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1379831 c.941A>G p.Y314C Chr.16: 75634231-75634231 on Build GRCh38
5828451 c.880-3delT p.? Chr.16: 75634295-75634295 on Build GRCh38
6690114 c.974G>A p.R325K Chr.16: 75634198-75634198 on Build GRCh38
1379832 c.857A>G p.Y286C Chr.16: 75634231-75634231 on Build GRCh38
2691519 c.934A>G p.I312V Chr.16: 75634238-75634238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141524539 Chr.16: 75634320-75634320 on Build GRCh38
rs557354682 Chr.16: 75634135-75634135 on Build GRCh38
rs1014934540 Chr.16: 75634243-75634243 on Build GRCh38
rs911194115 Chr.16: 75634505-75634505 on Build GRCh38
rs950178168 Chr.16: 75634394-75634394 on Build GRCh38
rs749868218 Chr.16: 75634112-75634112 on Build GRCh38
rs751162820 Chr.16: 75634543-75634543 on Build GRCh38
rs188266903 Chr.16: 75634542-75634542 on Build GRCh38
rs972789417 Chr.16: 75634274-75634274 on Build GRCh38
rs761777225 Chr.16: 75634109-75634109 on Build GRCh38
rs565287634 Chr.16: 75634461-75634461 on Build GRCh38
rs755039049 Chr.16: 75634339-75634339 on Build GRCh38
rs531015931 Chr.16: 75634465-75634465 on Build GRCh38
rs143117090 Chr.16: 75634214-75634214 on Build GRCh38
rs1056213032 Chr.16: 75634194-75634194 on Build GRCh38
rs749546625 Chr.16: 75634159-75634160 on Build GRCh38
rs112834496 Chr.16: 75634367-75634367 on Build GRCh38
rs6564270 Chr.16: 75634056-75634056 on Build GRCh38
rs767658376 Chr.16: 75634086-75634086 on Build GRCh38
rs534578327 Chr.16: 75634119-75634119 on Build GRCh38
rs770535491 Chr.16: 75634253-75634253 on Build GRCh38
rs753714622 Chr.16: 75634083-75634083 on Build GRCh38
rs146955132 Chr.16: 75634266-75634266 on Build GRCh38
rs769847503 Chr.16: 75634105-75634105 on Build GRCh38
rs766658174 Chr.16: 75634330-75634330 on Build GRCh38
rs966580606 Chr.16: 75634472-75634472 on Build GRCh38
rs967809910 Chr.16: 75634508-75634508 on Build GRCh38
rs758174482 Chr.16: 75634182-75634182 on Build GRCh38
rs1027933590 Chr.16: 75634272-75634272 on Build GRCh38
rs961467752 Chr.16: 75634263-75634263 on Build GRCh38
rs749922851 Chr.16: 75634064-75634064 on Build GRCh38
rs760113619 Chr.16: 75634148-75634148 on Build GRCh38
rs780032185 Chr.16: 75634139-75634139 on Build GRCh38
rs1021146171 Chr.16: 75634494-75634494 on Build GRCh38
rs147818487 Chr.16: 75634224-75634224 on Build GRCh38
rs995278439 Chr.16: 75634445-75634445 on Build GRCh38
rs765929610 Chr.16: 75634152-75634152 on Build GRCh38
rs1054708195 Chr.16: 75634346-75634346 on Build GRCh38
rs545723783 Chr.16: 75634233-75634233 on Build GRCh38
rs780786808 Chr.16: 75634118-75634118 on Build GRCh38
rs141370499 Chr.16: 75634245-75634245 on Build GRCh38
rs752461013 Chr.16: 75634178-75634178 on Build GRCh38
rs985433156 Chr.16: 75634313-75634313 on Build GRCh38
rs979100397 Chr.16: 75634510-75634510 on Build GRCh38
rs750601552 Chr.16: 75634206-75634206 on Build GRCh38
rs778742573 Chr.16: 75634089-75634089 on Build GRCh38
rs891524825 Chr.16: 75634101-75634101 on Build GRCh38
rs571244623 Chr.16: 75634460-75634460 on Build GRCh38
rs765146915 Chr.16: 75634373-75634373 on Build GRCh38
rs542932941 Chr.16: 75634352-75634352 on Build GRCh38
rs4888442 Chr.16: 75634427-75634427 on Build GRCh38
rs569905559 Chr.16: 75634079-75634079 on Build GRCh38
rs765194793 Chr.16: 75634277-75634277 on Build GRCh38
rs769283347 Chr.16: 75634118-75634118 on Build GRCh38
rs756393472 Chr.16: 75634212-75634212 on Build GRCh38
rs573478004 Chr.16: 75634383-75634383 on Build GRCh38
rs557125923 Chr.16: 75634343-75634343 on Build GRCh38
rs574224894 Chr.16: 75634351-75634351 on Build GRCh38
rs913579962 Chr.16: 75634347-75634347 on Build GRCh38
rs867760892 Chr.16: 75634048-75634048 on Build GRCh38
rs780722583 Chr.16: 75634111-75634111 on Build GRCh38
rs549286799 Chr.16: 75634103-75634103 on Build GRCh38
rs1023941782 Chr.16: 75634110-75634110 on Build GRCh38
rs781430538 Chr.16: 75634100-75634100 on Build GRCh38
rs757474004 Chr.16: 75634099-75634099 on Build GRCh38
rs759723301 Chr.16: 75634082-75634082 on Build GRCh38
rs987925704 Chr.16: 75634482-75634482 on Build GRCh38
rs374546258 Chr.16: 75634309-75634309 on Build GRCh38
rs765141659 Chr.16: 75634174-75634174 on Build GRCh38
rs753337428 Chr.16: 75634159-75634159 on Build GRCh38
rs773433097 Chr.16: 75634239-75634239 on Build GRCh38
rs1055405418 Chr.16: 75634384-75634384 on Build GRCh38
rs200641085 Chr.16: 75634333-75634333 on Build GRCh38
rs368330279 Chr.16: 75634223-75634223 on Build GRCh38
rs372373278 Chr.16: 75634318-75634318 on Build GRCh38
rs368734032 Chr.16: 75634316-75634316 on Build GRCh38
rs922605828 Chr.16: 75634285-75634285 on Build GRCh38
rs1017538134 Chr.16: 75634422-75634422 on Build GRCh38
rs544762350 Chr.16: 75634511-75634511 on Build GRCh38
rs529170677 Chr.16: 75634091-75634091 on Build GRCh38
rs772853728 Chr.16: 75634146-75634146 on Build GRCh38
rs962326605 Chr.16: 75634251-75634251 on Build GRCh38
rs772521991 Chr.16: 75634414-75634414 on Build GRCh38
rs1019130204 Chr.16: 75634471-75634471 on Build GRCh38
rs563450588 Chr.16: 75634528-75634528 on Build GRCh38
rs1017048975 Chr.16: 75634235-75634235 on Build GRCh38
rs945876939 Chr.16: 75634549-75634549 on Build GRCh38
rs762673443 Chr.16: 75634291-75634291 on Build GRCh38
rs953600279 Chr.16: 75634280-75634280 on Build GRCh38
rs565530256 Chr.16: 75634408-75634408 on Build GRCh38
rs933914367 Chr.16: 75634075-75634078 on Build GRCh38
rs751308806 Chr.16: 75634317-75634317 on Build GRCh38
rs1036832955 Chr.16: 75634404-75634404 on Build GRCh38
rs749348050 Chr.16: 75634216-75634216 on Build GRCh38
rs750161496 Chr.16: 75634374-75634374 on Build GRCh38
rs926253245 Chr.16: 75634538-75634538 on Build GRCh38
rs371292797 Chr.16: 75634254-75634254 on Build GRCh38
rs771095432 Chr.16: 75634245-75634246 on Build GRCh38
rs374114920 Chr.16: 75634202-75634202 on Build GRCh38
rs113846451 Chr.16: 75634388-75634388 on Build GRCh38
rs937922429 Chr.16: 75634372-75634372 on Build GRCh38
rs139177840 Chr.16: 75634117-75634117 on Build GRCh38
rs922569032 Chr.16: 75634060-75634060 on Build GRCh38
rs759506917 Chr.16: 75634165-75634165 on Build GRCh38
rs752616530 Chr.16: 75634281-75634281 on Build GRCh38
rs1007773505 Chr.16: 75634462-75634462 on Build GRCh38

View Full Product Details