Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45193470-45194060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2F2
Entrez Gene ID
2963
Gene Name
general transcription factor IIF subunit 2
Gene Aliases
BTF4, RAP30, TF2F2, TFIIF
UniGene
Hs.654582
Gene Chromosome Location
Chr.13: 45119830-45284105 on Build GRCh38
Transcripts
NM_004128.2 XM_011535052.2 XM_017020551.1 NM_198404.2

Gene Symbol
KCTD4
Entrez Gene ID
386618
Gene Name
potassium channel tetramerization domain containing 4
Gene Aliases
bA321C24.3
UniGene
Hs.23406
Gene Chromosome Location
Chr.13: 45192853-45201040 on Build GRCh38
Transcripts
NM_004128.2 XM_011535052.2 XM_017020551.1 NM_198404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3384851 c.542A>C p.D181A Chr.13: 45194026-45194026 on Build GRCh38
321149 c.556G>A p.E186K Chr.13: 45194012-45194012 on Build GRCh38
6250638 c.597C>A p.F199L Chr.13: 45193971-45193971 on Build GRCh38
212339 c.574A>C p.N192H Chr.13: 45193994-45193994 on Build GRCh38
947636 c.684C>T p.I228I Chr.13: 45193884-45193884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024498772 Chr.13: 45193505-45193505 on Build GRCh38
rs774662198 Chr.13: 45193806-45193806 on Build GRCh38
rs764800319 Chr.13: 45193851-45193851 on Build GRCh38
rs757957755 Chr.13: 45193761-45193761 on Build GRCh38
rs754320190 Chr.13: 45193738-45193738 on Build GRCh38
rs748258708 Chr.13: 45194006-45194006 on Build GRCh38
rs940553707 Chr.13: 45193694-45193694 on Build GRCh38
rs540154257 Chr.13: 45193622-45193622 on Build GRCh38
rs771518022 Chr.13: 45193999-45193999 on Build GRCh38
rs777328955 Chr.13: 45193926-45193926 on Build GRCh38
rs550541067 Chr.13: 45193681-45193681 on Build GRCh38
rs769543508 Chr.13: 45193969-45193969 on Build GRCh38
rs757122143 Chr.13: 45193951-45193951 on Build GRCh38
rs372074622 Chr.13: 45193723-45193723 on Build GRCh38
rs781127025 Chr.13: 45193961-45193961 on Build GRCh38
rs1052518259 Chr.13: 45193601-45193601 on Build GRCh38
rs747563889 Chr.13: 45193740-45193740 on Build GRCh38
rs931911942 Chr.13: 45193655-45193655 on Build GRCh38
rs1046431135 Chr.13: 45193973-45193973 on Build GRCh38
rs764757723 Chr.13: 45193845-45193845 on Build GRCh38
rs761346913 Chr.13: 45193828-45193828 on Build GRCh38
rs529145408 Chr.13: 45193680-45193680 on Build GRCh38
rs557380190 Chr.13: 45193673-45193673 on Build GRCh38
rs773222427 Chr.13: 45193812-45193812 on Build GRCh38
rs903340778 Chr.13: 45193993-45193993 on Build GRCh38
rs781332329 Chr.13: 45193780-45193789 on Build GRCh38
rs147828648 Chr.13: 45193575-45193575 on Build GRCh38
rs745619832 Chr.13: 45193968-45193968 on Build GRCh38
rs911717082 Chr.13: 45193682-45193682 on Build GRCh38
rs561599305 Chr.13: 45193633-45193633 on Build GRCh38
rs200819000 Chr.13: 45193775-45193775 on Build GRCh38
rs577265945 Chr.13: 45193700-45193700 on Build GRCh38
rs551823576 Chr.13: 45193990-45193990 on Build GRCh38
rs977519007 Chr.13: 45193547-45193547 on Build GRCh38
rs987373780 Chr.13: 45193667-45193667 on Build GRCh38
rs752365096 Chr.13: 45193858-45193858 on Build GRCh38
rs756751126 Chr.13: 45193886-45193886 on Build GRCh38
rs768815718 Chr.13: 45193786-45193786 on Build GRCh38
rs766996295 Chr.13: 45193822-45193822 on Build GRCh38
rs754195705 Chr.13: 45193827-45193827 on Build GRCh38
rs770917508 Chr.13: 45193773-45193773 on Build GRCh38
rs761258409 Chr.13: 45193813-45193813 on Build GRCh38
rs777188983 Chr.13: 45193769-45193769 on Build GRCh38
rs767608044 Chr.13: 45193811-45193811 on Build GRCh38
rs572480858 Chr.13: 45193603-45193603 on Build GRCh38
rs556713717 Chr.13: 45193525-45193525 on Build GRCh38
rs367728341 Chr.13: 45193839-45193839 on Build GRCh38
rs1047098349 Chr.13: 45193648-45193648 on Build GRCh38
rs370282435 Chr.13: 45193629-45193629 on Build GRCh38
rs751100423 Chr.13: 45193949-45193949 on Build GRCh38
rs975386674 Chr.13: 45193632-45193632 on Build GRCh38
rs754608643 Chr.13: 45193739-45193739 on Build GRCh38
rs575286288 Chr.13: 45193549-45193549 on Build GRCh38
rs771409844 Chr.13: 45193494-45193494 on Build GRCh38
rs563956377 Chr.13: 45193595-45193595 on Build GRCh38
rs17851609 Chr.13: 45193984-45193984 on Build GRCh38
rs776409810 Chr.13: 45193774-45193774 on Build GRCh38
rs747097419 Chr.13: 45193750-45193750 on Build GRCh38
rs150359535 Chr.13: 45193987-45193987 on Build GRCh38
rs533478138 Chr.13: 45193923-45193923 on Build GRCh38
rs772248786 Chr.13: 45193988-45193988 on Build GRCh38
rs765613388 Chr.13: 45194038-45194038 on Build GRCh38
rs1002637628 Chr.13: 45193686-45193686 on Build GRCh38
rs574459832 Chr.13: 45193516-45193519 on Build GRCh38
rs760161994 Chr.13: 45194017-45194017 on Build GRCh38
rs768681870 Chr.13: 45193760-45193762 on Build GRCh38
rs753309309 Chr.13: 45193809-45193809 on Build GRCh38
rs779758432 Chr.13: 45193983-45193983 on Build GRCh38
rs201765984 Chr.13: 45193807-45193807 on Build GRCh38
rs936833390 Chr.13: 45193598-45193598 on Build GRCh38
rs371466603 Chr.13: 45193882-45193882 on Build GRCh38
rs777310266 Chr.13: 45194003-45194003 on Build GRCh38
rs376571272 Chr.13: 45193998-45193998 on Build GRCh38
rs894709061 Chr.13: 45193918-45193918 on Build GRCh38

View Full Product Details