Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134498239-134498816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPRT1
Entrez Gene ID
3251
Gene Name
hypoxanthine phosphoribosyltransferase 1
Gene Aliases
HGPRT, HPRT
UniGene
Hs.412707
Gene Chromosome Location
Chr.X: 134460145-134500668 on Build GRCh38
Transcripts
NM_000194.2

SNP Detail

SNP ID Chromosome Location
rs137852493 Chr.X: 134498431-134498431 on Build GRCh38
rs759140165 Chr.X: 134498357-134498357 on Build GRCh38
rs962490410 Chr.X: 134498272-134498272 on Build GRCh38
rs727503964 Chr.X: 134498438-134498438 on Build GRCh38
rs973594871 Chr.X: 134498282-134498282 on Build GRCh38
rs946040539 Chr.X: 134498585-134498585 on Build GRCh38
rs137852497 Chr.X: 134498412-134498412 on Build GRCh38
rs776417613 Chr.X: 134498343-134498343 on Build GRCh38
rs746922210 Chr.X: 134498598-134498598 on Build GRCh38
rs775260163 Chr.X: 134498379-134498379 on Build GRCh38
rs770859486 Chr.X: 134498646-134498646 on Build GRCh38
rs756174482 Chr.X: 134498450-134498453 on Build GRCh38
rs137852492 Chr.X: 134498433-134498433 on Build GRCh38
rs1052743076 Chr.X: 134498737-134498737 on Build GRCh38
rs201069506 Chr.X: 134498465-134498465 on Build GRCh38
rs775350369 Chr.X: 134498722-134498722 on Build GRCh38
rs769520593 Chr.X: 134498367-134498367 on Build GRCh38
rs767780010 Chr.X: 134498413-134498413 on Build GRCh38
rs777742917 Chr.X: 134498350-134498351 on Build GRCh38
rs745705483 Chr.X: 134498712-134498712 on Build GRCh38
rs769680528 Chr.X: 134498719-134498719 on Build GRCh38
rs780870313 Chr.X: 134498704-134498704 on Build GRCh38
rs113467921 Chr.X: 134498481-134498481 on Build GRCh38
rs750762293 Chr.X: 134498426-134498426 on Build GRCh38
rs762592264 Chr.X: 134498381-134498381 on Build GRCh38
rs762682297 Chr.X: 134498723-134498723 on Build GRCh38
rs764117649 Chr.X: 134498769-134498769 on Build GRCh38
rs746151223 Chr.X: 134498293-134498293 on Build GRCh38
rs759719818 Chr.X: 134498362-134498362 on Build GRCh38
rs187024559 Chr.X: 134498291-134498291 on Build GRCh38
rs917474055 Chr.X: 134498555-134498555 on Build GRCh38
rs756354323 Chr.X: 134498460-134498460 on Build GRCh38
rs137852486 Chr.X: 134498670-134498670 on Build GRCh38
rs267606863 Chr.X: 134498655-134498655 on Build GRCh38
rs1000189373 Chr.X: 134498698-134498698 on Build GRCh38
rs137852504 Chr.X: 134498657-134498657 on Build GRCh38
rs747017210 Chr.X: 134498359-134498363 on Build GRCh38
rs111845471 Chr.X: 134498284-134498284 on Build GRCh38
rs762261130 Chr.X: 134498732-134498732 on Build GRCh38
rs906750848 Chr.X: 134498547-134498547 on Build GRCh38
rs977446460 Chr.X: 134498694-134498694 on Build GRCh38
rs137852498 Chr.X: 134498407-134498407 on Build GRCh38
rs763693519 Chr.X: 134498383-134498383 on Build GRCh38
rs773846679 Chr.X: 134498726-134498726 on Build GRCh38
rs137852479 Chr.X: 134498677-134498677 on Build GRCh38
rs1039536233 Chr.X: 134498755-134498755 on Build GRCh38
rs780307417 Chr.X: 134498470-134498470 on Build GRCh38
rs778990771 Chr.X: 134498569-134498569 on Build GRCh38
rs398123242 Chr.X: 134498400-134498400 on Build GRCh38
rs398123241 Chr.X: 134498389-134498389 on Build GRCh38
rs1046614776 Chr.X: 134498516-134498516 on Build GRCh38
rs575446030 Chr.X: 134498781-134498781 on Build GRCh38
rs534390401 Chr.X: 134498409-134498409 on Build GRCh38
rs768066647 Chr.X: 134498725-134498725 on Build GRCh38

View Full Product Details