Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138178483-138179031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD8
Entrez Gene ID
10902
Gene Name
bromodomain containing 8
Gene Aliases
SMAP, SMAP2, p120
UniGene
Hs.519337
Gene Chromosome Location
Chr.5: 138139766-138178669 on Build GRCh38
Transcripts
NM_001164326.1 NM_001300961.1 NM_001300962.1 NM_001300966.1 NM_006696.3 NM_139199.1 XM_005271855.2 XM_005271856.2 XM_005271857.2 XM_005271859.3 XM_005271860.3 XM_005271861.3 XM_011543109.1 XM_011543110.1 XM_017008969.1 XM_017008970.1 XM_017008971.1 XM_017008972.1 XM_017008973.1 XM_017008974.1 XM_017008975.1 XM_017008976.1 NM_005733.2

Gene Symbol
KIF20A
Entrez Gene ID
10112
Gene Name
kinesin family member 20A
Gene Aliases
MKLP2, RAB6KIFL
UniGene
Hs.718626
Gene Chromosome Location
Chr.5: 138178728-138187715 on Build GRCh38
Transcripts
NM_001164326.1 NM_001300961.1 NM_001300962.1 NM_001300966.1 NM_006696.3 NM_139199.1 XM_005271855.2 XM_005271856.2 XM_005271857.2 XM_005271859.3 XM_005271860.3 XM_005271861.3 XM_011543109.1 XM_011543110.1 XM_017008969.1 XM_017008970.1 XM_017008971.1 XM_017008972.1 XM_017008973.1 XM_017008974.1 XM_017008975.1 XM_017008976.1 NM_005733.2

SNP Detail

SNP ID Chromosome Location
rs12109653 Chr.5: 138178994-138178994 on Build GRCh38
rs1001623027 Chr.5: 138178892-138178892 on Build GRCh38
rs904898791 Chr.5: 138178965-138178965 on Build GRCh38
rs1027308581 Chr.5: 138178719-138178719 on Build GRCh38
rs1048694534 Chr.5: 138178964-138178964 on Build GRCh38
rs891219015 Chr.5: 138178508-138178508 on Build GRCh38
rs375431282 Chr.5: 138178545-138178545 on Build GRCh38
rs972844794 Chr.5: 138178982-138178982 on Build GRCh38
rs777407632 Chr.5: 138178685-138178685 on Build GRCh38
rs767542577 Chr.5: 138178570-138178570 on Build GRCh38
rs183712991 Chr.5: 138178873-138178873 on Build GRCh38
rs897178509 Chr.5: 138178850-138178850 on Build GRCh38
rs376401689 Chr.5: 138178509-138178509 on Build GRCh38
rs925138454 Chr.5: 138178693-138178693 on Build GRCh38
rs771468142 Chr.5: 138178604-138178604 on Build GRCh38
rs766305294 Chr.5: 138178658-138178658 on Build GRCh38
rs187858303 Chr.5: 138178768-138178768 on Build GRCh38
rs774815156 Chr.5: 138178897-138178897 on Build GRCh38
rs1013311549 Chr.5: 138178915-138178915 on Build GRCh38
rs965442154 Chr.5: 138178974-138178974 on Build GRCh38
rs918591119 Chr.5: 138178990-138178990 on Build GRCh38
rs966817695 Chr.5: 138178929-138178929 on Build GRCh38
rs551346132 Chr.5: 138178524-138178524 on Build GRCh38
rs199741689 Chr.5: 138178600-138178600 on Build GRCh38
rs985300184 Chr.5: 138178760-138178760 on Build GRCh38
rs912425794 Chr.5: 138178959-138178959 on Build GRCh38
rs771305924 Chr.5: 138178855-138178855 on Build GRCh38
rs749466497 Chr.5: 138178663-138178663 on Build GRCh38
rs969607784 Chr.5: 138178722-138178722 on Build GRCh38
rs781755711 Chr.5: 138178614-138178614 on Build GRCh38
rs753563541 Chr.5: 138178589-138178589 on Build GRCh38
rs985269136 Chr.5: 138178956-138178956 on Build GRCh38
rs988608692 Chr.5: 138178777-138178777 on Build GRCh38
rs1049158803 Chr.5: 138178880-138178880 on Build GRCh38
rs866492656 Chr.5: 138178705-138178705 on Build GRCh38
rs751621993 Chr.5: 138178565-138178565 on Build GRCh38
rs906116116 Chr.5: 138178881-138178881 on Build GRCh38
rs755939657 Chr.5: 138178583-138178583 on Build GRCh38
rs1032640450 Chr.5: 138178941-138178941 on Build GRCh38
rs944556897 Chr.5: 138178907-138178907 on Build GRCh38
rs192986534 Chr.5: 138178840-138178840 on Build GRCh38
rs372349827 Chr.5: 138178644-138178644 on Build GRCh38
rs761770953 Chr.5: 138178569-138178569 on Build GRCh38
rs966634427 Chr.5: 138178667-138178667 on Build GRCh38
rs931873001 Chr.5: 138178879-138178879 on Build GRCh38
rs767493772 Chr.5: 138178654-138178654 on Build GRCh38
rs574020246 Chr.5: 138178977-138178977 on Build GRCh38
rs911690324 Chr.5: 138178902-138178902 on Build GRCh38
rs932437868 Chr.5: 138178726-138178726 on Build GRCh38
rs569333270 Chr.5: 138178781-138178781 on Build GRCh38
rs558094474 Chr.5: 138178922-138178922 on Build GRCh38
rs773212596 Chr.5: 138178655-138178655 on Build GRCh38
rs779933454 Chr.5: 138178587-138178587 on Build GRCh38
rs896215362 Chr.5: 138178914-138178914 on Build GRCh38
rs749544382 Chr.5: 138178639-138178639 on Build GRCh38
rs567434905 Chr.5: 138178755-138178755 on Build GRCh38
rs996186624 Chr.5: 138178526-138178526 on Build GRCh38
rs776906101 Chr.5: 138178784-138178785 on Build GRCh38
rs1035586799 Chr.5: 138178733-138178733 on Build GRCh38
rs201418867 Chr.5: 138178623-138178623 on Build GRCh38
rs747742263 Chr.5: 138178603-138178603 on Build GRCh38
rs746428643 Chr.5: 138178936-138178936 on Build GRCh38
rs986077121 Chr.5: 138178885-138178885 on Build GRCh38
rs558909217 Chr.5: 138178594-138178594 on Build GRCh38
rs7729926 Chr.5: 138178952-138178952 on Build GRCh38
rs748477206 Chr.5: 138178654-138178654 on Build GRCh38
rs540489038 Chr.5: 138178930-138178930 on Build GRCh38
rs944124495 Chr.5: 138178804-138178804 on Build GRCh38
rs532930353 Chr.5: 138178523-138178523 on Build GRCh38
rs367956159 Chr.5: 138178555-138178555 on Build GRCh38
rs371786405 Chr.5: 138178573-138178573 on Build GRCh38
rs199798642 Chr.5: 138178631-138178631 on Build GRCh38
rs775784362 Chr.5: 138178634-138178634 on Build GRCh38
rs113669474 Chr.5: 138178649-138178649 on Build GRCh38
rs1037049114 Chr.5: 138178519-138178519 on Build GRCh38
rs556115115 Chr.5: 138178770-138178770 on Build GRCh38
rs572920523 Chr.5: 138178928-138178928 on Build GRCh38
rs994468203 Chr.5: 138178671-138178671 on Build GRCh38

View Full Product Details