Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 27824203-27824759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H4J
Entrez Gene ID
8363
Gene Name
histone cluster 1, H4j
Gene Aliases
H4/e, H4F2iv, H4FE, dJ160A22.2
UniGene
Hs.278483
Gene Chromosome Location
Chr.6: 27824125-27824480 on Build GRCh38
Transcripts
NM_021968.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1442760 c.115_118delGCTC p.R40fs*4 Chr.6: 27824239-27824242 on Build GRCh38
1662144 c.235C>T p.R79C Chr.6: 27824359-27824359 on Build GRCh38
1076962 c.223G>A p.E75K Chr.6: 27824347-27824347 on Build GRCh38
4950657 c.175C>A p.L59M Chr.6: 27824299-27824299 on Build GRCh38
6862217 c.297C>A p.Y99* Chr.6: 27824421-27824421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778000351 Chr.6: 27824422-27824422 on Build GRCh38
rs984384976 Chr.6: 27824379-27824379 on Build GRCh38
rs1009055384 Chr.6: 27824718-27824718 on Build GRCh38
rs749213131 Chr.6: 27824358-27824358 on Build GRCh38
rs200336073 Chr.6: 27824730-27824740 on Build GRCh38
rs558036557 Chr.6: 27824539-27824539 on Build GRCh38
rs1005880844 Chr.6: 27824498-27824498 on Build GRCh38
rs376029911 Chr.6: 27824480-27824480 on Build GRCh38
rs969860853 Chr.6: 27824502-27824502 on Build GRCh38
rs762031886 Chr.6: 27824324-27824324 on Build GRCh38
rs1021569378 Chr.6: 27824611-27824611 on Build GRCh38
rs760576808 Chr.6: 27824309-27824310 on Build GRCh38
rs761825882 Chr.6: 27824251-27824251 on Build GRCh38
rs796926826 Chr.6: 27824316-27824316 on Build GRCh38
rs200142548 Chr.6: 27824469-27824469 on Build GRCh38
rs966916462 Chr.6: 27824614-27824614 on Build GRCh38
rs980240865 Chr.6: 27824506-27824506 on Build GRCh38
rs780912629 Chr.6: 27824349-27824349 on Build GRCh38
rs927227017 Chr.6: 27824665-27824665 on Build GRCh38
rs768450749 Chr.6: 27824318-27824318 on Build GRCh38
rs200499 Chr.6: 27824443-27824443 on Build GRCh38
rs368679902 Chr.6: 27824223-27824223 on Build GRCh38
rs534137698 Chr.6: 27824559-27824559 on Build GRCh38
rs567597862 Chr.6: 27824652-27824652 on Build GRCh38
rs764195698 Chr.6: 27824242-27824242 on Build GRCh38
rs771121560 Chr.6: 27824695-27824695 on Build GRCh38
rs368683667 Chr.6: 27824291-27824291 on Build GRCh38
rs756724260 Chr.6: 27824290-27824290 on Build GRCh38
rs201388893 Chr.6: 27824277-27824277 on Build GRCh38
rs141884356 Chr.6: 27824522-27824522 on Build GRCh38
rs768344636 Chr.6: 27824361-27824361 on Build GRCh38
rs572587792 Chr.6: 27824317-27824317 on Build GRCh38
rs905737285 Chr.6: 27824585-27824585 on Build GRCh38
rs1007061623 Chr.6: 27824311-27824311 on Build GRCh38
rs753735996 Chr.6: 27824414-27824414 on Build GRCh38
rs200498 Chr.6: 27824450-27824450 on Build GRCh38
rs374087494 Chr.6: 27824393-27824393 on Build GRCh38
rs1025867088 Chr.6: 27824487-27824487 on Build GRCh38
rs758191325 Chr.6: 27824459-27824459 on Build GRCh38
rs775407783 Chr.6: 27824226-27824226 on Build GRCh38
rs760765881 Chr.6: 27824381-27824381 on Build GRCh38
rs888658273 Chr.6: 27824492-27824492 on Build GRCh38
rs964406571 Chr.6: 27824655-27824655 on Build GRCh38
rs879105861 Chr.6: 27824304-27824304 on Build GRCh38
rs775764850 Chr.6: 27824441-27824441 on Build GRCh38
rs756918047 Chr.6: 27824346-27824346 on Build GRCh38
rs746418656 Chr.6: 27824481-27824481 on Build GRCh38
rs571748474 Chr.6: 27824699-27824699 on Build GRCh38
rs780164912 Chr.6: 27824355-27824355 on Build GRCh38
rs905410244 Chr.6: 27824708-27824708 on Build GRCh38
rs749747655 Chr.6: 27824476-27824476 on Build GRCh38
rs560575579 Chr.6: 27824622-27824622 on Build GRCh38
rs754930996 Chr.6: 27824415-27824415 on Build GRCh38
rs528147725 Chr.6: 27824605-27824605 on Build GRCh38
rs578139145 Chr.6: 27824591-27824591 on Build GRCh38
rs920245106 Chr.6: 27824637-27824637 on Build GRCh38
rs529279651 Chr.6: 27824658-27824658 on Build GRCh38
rs201673306 Chr.6: 27824447-27824447 on Build GRCh38
rs566078638 Chr.6: 27824670-27824670 on Build GRCh38
rs578132001 Chr.6: 27824546-27824546 on Build GRCh38
rs950407247 Chr.6: 27824368-27824368 on Build GRCh38
rs964923581 Chr.6: 27824624-27824624 on Build GRCh38
rs1009978044 Chr.6: 27824734-27824734 on Build GRCh38
rs942824669 Chr.6: 27824557-27824557 on Build GRCh38
rs1010972814 Chr.6: 27824604-27824604 on Build GRCh38
rs371683547 Chr.6: 27824301-27824301 on Build GRCh38
rs369854319 Chr.6: 27824391-27824391 on Build GRCh38
rs760028958 Chr.6: 27824383-27824383 on Build GRCh38
rs976747767 Chr.6: 27824656-27824656 on Build GRCh38
rs767016473 Chr.6: 27824552-27824553 on Build GRCh38
rs74619448 Chr.6: 27824661-27824661 on Build GRCh38
rs756556409 Chr.6: 27824265-27824265 on Build GRCh38
rs751184309 Chr.6: 27824343-27824343 on Build GRCh38
rs376723488 Chr.6: 27824407-27824407 on Build GRCh38
rs777486485 Chr.6: 27824463-27824463 on Build GRCh38
rs780031529 Chr.6: 27824310-27824310 on Build GRCh38
rs777009121 Chr.6: 27824382-27824382 on Build GRCh38
rs1025961714 Chr.6: 27824354-27824354 on Build GRCh38
rs951919209 Chr.6: 27824639-27824639 on Build GRCh38
rs576512350 Chr.6: 27824268-27824268 on Build GRCh38
rs930409984 Chr.6: 27824564-27824564 on Build GRCh38
rs910033855 Chr.6: 27824686-27824686 on Build GRCh38
rs757161572 Chr.6: 27824708-27824709 on Build GRCh38
rs1021120441 Chr.6: 27824505-27824505 on Build GRCh38
rs930052366 Chr.6: 27824666-27824666 on Build GRCh38
rs558237222 Chr.6: 27824685-27824685 on Build GRCh38
rs774622497 Chr.6: 27824724-27824724 on Build GRCh38
rs754198461 Chr.6: 27824274-27824274 on Build GRCh38
rs772949804 Chr.6: 27824243-27824244 on Build GRCh38
rs537290511 Chr.6: 27824701-27824701 on Build GRCh38
rs541294957 Chr.6: 27824332-27824332 on Build GRCh38
rs974420746 Chr.6: 27824629-27824629 on Build GRCh38
rs745935948 Chr.6: 27824472-27824472 on Build GRCh38
rs36108923 Chr.6: 27824385-27824385 on Build GRCh38
rs375380321 Chr.6: 27824732-27824732 on Build GRCh38
rs749919181 Chr.6: 27824350-27824350 on Build GRCh38
rs769710695 Chr.6: 27824473-27824473 on Build GRCh38
rs772154105 Chr.6: 27824325-27824325 on Build GRCh38
rs753860349 Chr.6: 27824454-27824454 on Build GRCh38
rs752588777 Chr.6: 27824452-27824452 on Build GRCh38
rs117048465 Chr.6: 27824584-27824584 on Build GRCh38
rs913155559 Chr.6: 27824526-27824526 on Build GRCh38
rs765570238 Chr.6: 27824340-27824340 on Build GRCh38
rs535888933 Chr.6: 27824510-27824510 on Build GRCh38
rs893793646 Chr.6: 27824684-27824684 on Build GRCh38
rs187852525 Chr.6: 27824418-27824418 on Build GRCh38
rs979070063 Chr.6: 27824650-27824650 on Build GRCh38
rs368673524 Chr.6: 27824331-27824331 on Build GRCh38
rs1002211115 Chr.6: 27824586-27824586 on Build GRCh38
rs543664579 Chr.6: 27824556-27824556 on Build GRCh38
rs902562927 Chr.6: 27824717-27824717 on Build GRCh38
rs773266406 Chr.6: 27824329-27824329 on Build GRCh38
rs746425377 Chr.6: 27824438-27824438 on Build GRCh38
rs920379274 Chr.6: 27824560-27824560 on Build GRCh38
rs780788014 Chr.6: 27824295-27824295 on Build GRCh38
rs774493054 Chr.6: 27824736-27824736 on Build GRCh38
rs774541671 Chr.6: 27824247-27824247 on Build GRCh38
rs555943018 Chr.6: 27824298-27824298 on Build GRCh38
rs750050019 Chr.6: 27824397-27824397 on Build GRCh38
rs868123194 Chr.6: 27824644-27824644 on Build GRCh38
rs563687340 Chr.6: 27824576-27824576 on Build GRCh38
rs530773405 Chr.6: 27824635-27824635 on Build GRCh38
rs1042495081 Chr.6: 27824698-27824698 on Build GRCh38
rs750934985 Chr.6: 27824262-27824262 on Build GRCh38
rs745854626 Chr.6: 27824299-27824299 on Build GRCh38
rs769160731 Chr.6: 27824477-27824477 on Build GRCh38
rs113345516 Chr.6: 27824435-27824435 on Build GRCh38
rs987725796 Chr.6: 27824512-27824512 on Build GRCh38
rs370039295 Chr.6: 27824485-27824485 on Build GRCh38
rs947285126 Chr.6: 27824530-27824530 on Build GRCh38
rs377187343 Chr.6: 27824374-27824374 on Build GRCh38
rs760350021 Chr.6: 27824446-27824446 on Build GRCh38
rs774472207 Chr.6: 27824319-27824319 on Build GRCh38
rs771011750 Chr.6: 27824436-27824436 on Build GRCh38
rs201031164 Chr.6: 27824334-27824334 on Build GRCh38
rs528006754 Chr.6: 27824688-27824688 on Build GRCh38
rs201433804 Chr.6: 27824253-27824253 on Build GRCh38
rs773321936 Chr.6: 27824376-27824376 on Build GRCh38
rs1016333803 Chr.6: 27824499-27824499 on Build GRCh38
rs1037394969 Chr.6: 27824607-27824607 on Build GRCh38
rs534770168 Chr.6: 27824671-27824671 on Build GRCh38
rs983235669 Chr.6: 27824647-27824647 on Build GRCh38
rs578165187 Chr.6: 27824352-27824352 on Build GRCh38
rs754321135 Chr.6: 27824333-27824333 on Build GRCh38
rs144771004 Chr.6: 27824497-27824497 on Build GRCh38
rs943041972 Chr.6: 27824542-27824542 on Build GRCh38
rs776099558 Chr.6: 27824475-27824475 on Build GRCh38
rs763300464 Chr.6: 27824236-27824236 on Build GRCh38
rs147252800 Chr.6: 27824716-27824716 on Build GRCh38
rs148537877 Chr.6: 27824521-27824521 on Build GRCh38
rs996374311 Chr.6: 27824737-27824737 on Build GRCh38
rs779554897 Chr.6: 27824409-27824409 on Build GRCh38
rs371934187 Chr.6: 27824654-27824654 on Build GRCh38
rs767290620 Chr.6: 27824239-27824239 on Build GRCh38
rs765420405 Chr.6: 27824476-27824477 on Build GRCh38
rs542250499 Chr.6: 27824600-27824600 on Build GRCh38
rs376871813 Chr.6: 27824313-27824313 on Build GRCh38
rs781165358 Chr.6: 27824471-27824471 on Build GRCh38
rs2277104 Chr.6: 27824454-27824454 on Build GRCh38
rs869303647 Chr.6: 27824564-27824564 on Build GRCh38
rs1048047555 Chr.6: 27824574-27824574 on Build GRCh38
rs550896897 Chr.6: 27824489-27824489 on Build GRCh38
rs199585085 Chr.6: 27824439-27824439 on Build GRCh38
rs776758314 Chr.6: 27824437-27824437 on Build GRCh38
rs891783537 Chr.6: 27824287-27824287 on Build GRCh38
rs893922149 Chr.6: 27824599-27824599 on Build GRCh38
rs930145804 Chr.6: 27824563-27824563 on Build GRCh38
rs536927709 Chr.6: 27824630-27824631 on Build GRCh38
rs766055274 Chr.6: 27824404-27824405 on Build GRCh38
rs543724783 Chr.6: 27824373-27824373 on Build GRCh38
rs778712914 Chr.6: 27824366-27824366 on Build GRCh38
rs571433614 Chr.6: 27824468-27824468 on Build GRCh38
rs754855207 Chr.6: 27824455-27824455 on Build GRCh38
rs758670827 Chr.6: 27824421-27824421 on Build GRCh38
rs550884484 Chr.6: 27824640-27824640 on Build GRCh38
rs535138994 Chr.6: 27824531-27824531 on Build GRCh38
rs570061868 Chr.6: 27824504-27824504 on Build GRCh38
rs545591827 Chr.6: 27824621-27824621 on Build GRCh38
rs764517642 Chr.6: 27824442-27824442 on Build GRCh38
rs746632790 Chr.6: 27824283-27824283 on Build GRCh38
rs559379256 Chr.6: 27824603-27824603 on Build GRCh38
rs901230857 Chr.6: 27824559-27824559 on Build GRCh38
rs530458325 Chr.6: 27824653-27824653 on Build GRCh38
rs754914455 Chr.6: 27824448-27824448 on Build GRCh38
rs949896038 Chr.6: 27824581-27824581 on Build GRCh38

View Full Product Details