Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27095495-27096094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOTAIRM1
Entrez Gene ID
100506311
Gene Name
HOXA transcript antisense RNA, myeloid-specific 1
Gene Aliases
HOXA-AS1, HOXA1-AS1, NCRNA00179
UniGene
Hs.445239
Gene Chromosome Location
Chr.7: 27096094-27100258 on Build GRCh38
Transcripts
NR_038366.1 NR_038367.1 NM_005522.4 NM_153620.2

Gene Symbol
HOXA1
Entrez Gene ID
3198
Gene Name
homeobox A1
Gene Aliases
BSAS, HOX1, HOX1F
UniGene
Hs.67397
Gene Chromosome Location
Chr.7: 27092993-27096006 on Build GRCh38
Transcripts
NR_038366.1 NR_038367.1 NM_005522.4 NM_153620.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6514828 c.290C>T p.P97L Chr.7: 27095623-27095623 on Build GRCh38
6285112 c.265T>G p.S89A Chr.7: 27095648-27095648 on Build GRCh38
3880383 c.122C>T p.A41V Chr.7: 27095791-27095791 on Build GRCh38
74937 c.47G>C p.S16T Chr.7: 27095866-27095866 on Build GRCh38
3880381 c.378C>T p.C126C Chr.7: 27095535-27095535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886062262 Chr.7: 27095842-27095842 on Build GRCh38
rs997172905 Chr.7: 27095676-27095676 on Build GRCh38
rs764322837 Chr.7: 27095689-27095690 on Build GRCh38
rs572829050 Chr.7: 27095654-27095654 on Build GRCh38
rs752885733 Chr.7: 27095544-27095544 on Build GRCh38
rs774629222 Chr.7: 27095925-27095926 on Build GRCh38
rs750312129 Chr.7: 27095738-27095738 on Build GRCh38
rs104894018 Chr.7: 27095837-27095837 on Build GRCh38
rs865943119 Chr.7: 27095789-27095789 on Build GRCh38
rs570731900 Chr.7: 27096023-27096023 on Build GRCh38
rs370355386 Chr.7: 27095885-27095885 on Build GRCh38
rs368221577 Chr.7: 27096022-27096022 on Build GRCh38
rs767288977 Chr.7: 27095825-27095825 on Build GRCh38
rs376159031 Chr.7: 27095978-27095978 on Build GRCh38
rs917828771 Chr.7: 27095728-27095728 on Build GRCh38
rs104894017 Chr.7: 27095829-27095829 on Build GRCh38
rs927725372 Chr.7: 27096022-27096022 on Build GRCh38
rs1023408773 Chr.7: 27095980-27095980 on Build GRCh38
rs199540608 Chr.7: 27095944-27095944 on Build GRCh38
rs781616537 Chr.7: 27095848-27095848 on Build GRCh38
rs760765803 Chr.7: 27095678-27095678 on Build GRCh38
rs781331719 Chr.7: 27095694-27095694 on Build GRCh38
rs766431051 Chr.7: 27095537-27095537 on Build GRCh38
rs753969638 Chr.7: 27095698-27095700 on Build GRCh38
rs763704532 Chr.7: 27095950-27095950 on Build GRCh38
rs77832452 Chr.7: 27095739-27095739 on Build GRCh38
rs960142176 Chr.7: 27095970-27095970 on Build GRCh38
rs761654698 Chr.7: 27095730-27095730 on Build GRCh38
rs544314279 Chr.7: 27095690-27095698 on Build GRCh38
rs761181029 Chr.7: 27095534-27095534 on Build GRCh38
rs758237482 Chr.7: 27095697-27095698 on Build GRCh38
rs878979521 Chr.7: 27095737-27095737 on Build GRCh38
rs775236854 Chr.7: 27095863-27095865 on Build GRCh38
rs200922808 Chr.7: 27095793-27095793 on Build GRCh38
rs768796271 Chr.7: 27095704-27095706 on Build GRCh38
rs964954049 Chr.7: 27095998-27095998 on Build GRCh38
rs981104456 Chr.7: 27096002-27096002 on Build GRCh38
rs751378994 Chr.7: 27095729-27095729 on Build GRCh38
rs747464910 Chr.7: 27095698-27095706 on Build GRCh38
rs748952987 Chr.7: 27095704-27095704 on Build GRCh38
rs111580588 Chr.7: 27095577-27095577 on Build GRCh38
rs894460452 Chr.7: 27095655-27095655 on Build GRCh38
rs761162491 Chr.7: 27095651-27095651 on Build GRCh38
rs140318803 Chr.7: 27095818-27095818 on Build GRCh38
rs559987637 Chr.7: 27095724-27095724 on Build GRCh38
rs769077854 Chr.7: 27095911-27095914 on Build GRCh38
rs377149193 Chr.7: 27095619-27095619 on Build GRCh38
rs150376380 Chr.7: 27095834-27095834 on Build GRCh38
rs1021758952 Chr.7: 27095868-27095868 on Build GRCh38
rs886062261 Chr.7: 27095748-27095748 on Build GRCh38
rs765261087 Chr.7: 27095661-27095661 on Build GRCh38
rs762135120 Chr.7: 27095822-27095822 on Build GRCh38
rs146782650 Chr.7: 27095719-27095719 on Build GRCh38
rs746373446 Chr.7: 27095773-27095773 on Build GRCh38
rs372667623 Chr.7: 27095960-27095960 on Build GRCh38
rs747419833 Chr.7: 27095709-27095709 on Build GRCh38
rs533892779 Chr.7: 27095719-27095720 on Build GRCh38
rs950475320 Chr.7: 27096018-27096018 on Build GRCh38
rs768791109 Chr.7: 27095873-27095873 on Build GRCh38
rs904519138 Chr.7: 27096024-27096024 on Build GRCh38
rs749847386 Chr.7: 27095870-27095870 on Build GRCh38
rs370896322 Chr.7: 27095826-27095826 on Build GRCh38
rs867601078 Chr.7: 27095662-27095662 on Build GRCh38
rs776482521 Chr.7: 27095918-27095918 on Build GRCh38
rs773622256 Chr.7: 27095623-27095623 on Build GRCh38
rs764729360 Chr.7: 27095836-27095836 on Build GRCh38
rs773671587 Chr.7: 27095528-27095528 on Build GRCh38
rs575212090 Chr.7: 27095682-27095682 on Build GRCh38
rs754443433 Chr.7: 27095835-27095835 on Build GRCh38
rs374581356 Chr.7: 27095852-27095852 on Build GRCh38
rs533047031 Chr.7: 27096061-27096061 on Build GRCh38
rs368428758 Chr.7: 27095695-27095697 on Build GRCh38
rs777714750 Chr.7: 27095561-27095561 on Build GRCh38
rs371597524 Chr.7: 27095541-27095541 on Build GRCh38
rs759482597 Chr.7: 27095919-27095919 on Build GRCh38
rs754087421 Chr.7: 27096027-27096027 on Build GRCh38
rs772220119 Chr.7: 27095890-27095890 on Build GRCh38
rs745370848 Chr.7: 27095556-27095557 on Build GRCh38
rs778487133 Chr.7: 27095760-27095760 on Build GRCh38
rs780377792 Chr.7: 27095858-27095858 on Build GRCh38
rs372765755 Chr.7: 27095586-27095586 on Build GRCh38
rs898886363 Chr.7: 27095549-27095549 on Build GRCh38
rs752271771 Chr.7: 27095942-27095942 on Build GRCh38
rs758043074 Chr.7: 27095847-27095847 on Build GRCh38
rs779608126 Chr.7: 27095740-27095740 on Build GRCh38
rs906374305 Chr.7: 27095610-27095610 on Build GRCh38
rs754599509 Chr.7: 27095746-27095746 on Build GRCh38
rs376243279 Chr.7: 27095725-27095725 on Build GRCh38
rs199620262 Chr.7: 27095703-27095703 on Build GRCh38
rs920268318 Chr.7: 27096003-27096003 on Build GRCh38
rs769853692 Chr.7: 27095721-27095721 on Build GRCh38
rs45491004 Chr.7: 27096034-27096034 on Build GRCh38
rs752896589 Chr.7: 27095665-27095665 on Build GRCh38
rs370583131 Chr.7: 27095691-27095691 on Build GRCh38
rs373107510 Chr.7: 27095722-27095722 on Build GRCh38
rs146737549 Chr.7: 27095771-27095771 on Build GRCh38
rs878954224 Chr.7: 27096029-27096029 on Build GRCh38
rs367605225 Chr.7: 27095857-27095857 on Build GRCh38
rs762160733 Chr.7: 27095931-27095932 on Build GRCh38
rs1030782280 Chr.7: 27096021-27096021 on Build GRCh38
rs778078126 Chr.7: 27095708-27095708 on Build GRCh38
rs767586523 Chr.7: 27095692-27095697 on Build GRCh38
rs748841201 Chr.7: 27095888-27095890 on Build GRCh38
rs771218252 Chr.7: 27095770-27095770 on Build GRCh38
rs886062260 Chr.7: 27095697-27095697 on Build GRCh38
rs2074398 Chr.7: 27095700-27095700 on Build GRCh38
rs1047520159 Chr.7: 27095596-27095596 on Build GRCh38
rs1033634182 Chr.7: 27095638-27095638 on Build GRCh38
rs769552019 Chr.7: 27095685-27095686 on Build GRCh38
rs773032918 Chr.7: 27095891-27095891 on Build GRCh38
rs989150649 Chr.7: 27096033-27096033 on Build GRCh38
rs983179827 Chr.7: 27096021-27096022 on Build GRCh38
rs750568000 Chr.7: 27095691-27095692 on Build GRCh38
rs199827755 Chr.7: 27095574-27095574 on Build GRCh38
rs10951154 Chr.7: 27095695-27095695 on Build GRCh38
rs757334841 Chr.7: 27095698-27095703 on Build GRCh38
rs774858049 Chr.7: 27095690-27095707 on Build GRCh38
rs770272069 Chr.7: 27095790-27095790 on Build GRCh38
rs777153568 Chr.7: 27095717-27095717 on Build GRCh38
rs1053063606 Chr.7: 27096030-27096030 on Build GRCh38
rs1008225872 Chr.7: 27095859-27095859 on Build GRCh38
rs890518053 Chr.7: 27096032-27096032 on Build GRCh38
rs369218661 Chr.7: 27095567-27095567 on Build GRCh38
rs766226303 Chr.7: 27095914-27095914 on Build GRCh38
rs144288739 Chr.7: 27095762-27095762 on Build GRCh38
rs45602734 Chr.7: 27096026-27096026 on Build GRCh38
rs587777901 Chr.7: 27095698-27095698 on Build GRCh38
rs977912034 Chr.7: 27095689-27095689 on Build GRCh38
rs753327260 Chr.7: 27095743-27095743 on Build GRCh38
rs758708392 Chr.7: 27095673-27095673 on Build GRCh38
rs760336148 Chr.7: 27095893-27095893 on Build GRCh38
rs769152039 Chr.7: 27095737-27095738 on Build GRCh38
rs745429376 Chr.7: 27095700-27095701 on Build GRCh38
rs751466974 Chr.7: 27095680-27095680 on Build GRCh38
rs765543665 Chr.7: 27095543-27095543 on Build GRCh38
rs998940526 Chr.7: 27095921-27095921 on Build GRCh38
rs927780998 Chr.7: 27095775-27095775 on Build GRCh38
rs766274047 Chr.7: 27095832-27095832 on Build GRCh38
rs764712990 Chr.7: 27095938-27095938 on Build GRCh38
rs776550739 Chr.7: 27095657-27095657 on Build GRCh38
rs763176817 Chr.7: 27095796-27095796 on Build GRCh38
rs771305944 Chr.7: 27095715-27095715 on Build GRCh38
rs760625693 Chr.7: 27095828-27095828 on Build GRCh38
rs546435383 Chr.7: 27096066-27096066 on Build GRCh38
rs1022228508 Chr.7: 27095697-27095697 on Build GRCh38
rs774979619 Chr.7: 27095690-27095701 on Build GRCh38
rs968808790 Chr.7: 27095688-27095688 on Build GRCh38
rs969293002 Chr.7: 27095995-27095995 on Build GRCh38
rs768882831 Chr.7: 27095814-27095814 on Build GRCh38
rs775163257 Chr.7: 27095609-27095609 on Build GRCh38
rs886062259 Chr.7: 27095658-27095658 on Build GRCh38
rs748371765 Chr.7: 27095888-27095888 on Build GRCh38
rs1008939351 Chr.7: 27096041-27096041 on Build GRCh38
rs766665411 Chr.7: 27095694-27095695 on Build GRCh38
rs758170303 Chr.7: 27095559-27095559 on Build GRCh38
rs965283358 Chr.7: 27096069-27096069 on Build GRCh38
rs772358886 Chr.7: 27095620-27095620 on Build GRCh38
rs941794162 Chr.7: 27095652-27095652 on Build GRCh38
rs745425180 Chr.7: 27095581-27095581 on Build GRCh38

View Full Product Details