Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92806862-92807445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA5
Entrez Gene ID
9950
Gene Name
golgin A5
Gene Aliases
GOLIM5, RFG5, ret-II
UniGene
Hs.104320
Gene Chromosome Location
Chr.14: 92794231-92839963 on Build GRCh38
Transcripts
NM_005113.3 XM_011537420.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1608071 c.772+6G>T p.? Chr.14: 92806969-92806969 on Build GRCh38
6554612 c.682C>T p.R228* Chr.14: 92806873-92806873 on Build GRCh38
1608070 c.741G>A p.S247S Chr.14: 92806932-92806932 on Build GRCh38
1678204 c.755C>T p.S252F Chr.14: 92806946-92806946 on Build GRCh38
958899 c.730G>T p.E244* Chr.14: 92806921-92806921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562186941 Chr.14: 92807270-92807270 on Build GRCh38
rs893146007 Chr.14: 92806977-92806977 on Build GRCh38
rs61075676 Chr.14: 92807329-92807333 on Build GRCh38
rs954762499 Chr.14: 92807036-92807036 on Build GRCh38
rs1001846456 Chr.14: 92807328-92807328 on Build GRCh38
rs990430909 Chr.14: 92807206-92807206 on Build GRCh38
rs866999187 Chr.14: 92807066-92807066 on Build GRCh38
rs368665594 Chr.14: 92806940-92806940 on Build GRCh38
rs1048235997 Chr.14: 92807375-92807375 on Build GRCh38
rs111956343 Chr.14: 92807025-92807025 on Build GRCh38
rs540738752 Chr.14: 92807070-92807070 on Build GRCh38
rs371093608 Chr.14: 92807415-92807415 on Build GRCh38
rs964359277 Chr.14: 92807075-92807075 on Build GRCh38
rs781371576 Chr.14: 92807202-92807202 on Build GRCh38
rs1026178536 Chr.14: 92807052-92807052 on Build GRCh38
rs531381179 Chr.14: 92807251-92807251 on Build GRCh38
rs200228925 Chr.14: 92806958-92806958 on Build GRCh38
rs979493899 Chr.14: 92807114-92807114 on Build GRCh38
rs751513397 Chr.14: 92807321-92807325 on Build GRCh38
rs1037523547 Chr.14: 92807422-92807422 on Build GRCh38
rs1037573269 Chr.14: 92807223-92807223 on Build GRCh38
rs1002942901 Chr.14: 92807026-92807026 on Build GRCh38
rs924808091 Chr.14: 92807179-92807179 on Build GRCh38
rs112701736 Chr.14: 92807065-92807065 on Build GRCh38
rs757511882 Chr.14: 92807162-92807162 on Build GRCh38
rs758423919 Chr.14: 92807023-92807023 on Build GRCh38
rs562832405 Chr.14: 92807250-92807250 on Build GRCh38
rs142797011 Chr.14: 92807328-92807332 on Build GRCh38
rs956371207 Chr.14: 92807189-92807189 on Build GRCh38
rs921258473 Chr.14: 92807117-92807117 on Build GRCh38
rs369129790 Chr.14: 92807321-92807321 on Build GRCh38
rs761602617 Chr.14: 92806978-92806978 on Build GRCh38
rs544825654 Chr.14: 92807002-92807002 on Build GRCh38
rs551207423 Chr.14: 92807273-92807273 on Build GRCh38
rs113080946 Chr.14: 92807191-92807191 on Build GRCh38
rs558042533 Chr.14: 92807028-92807028 on Build GRCh38
rs45539743 Chr.14: 92807226-92807226 on Build GRCh38
rs371880770 Chr.14: 92806955-92806955 on Build GRCh38
rs765581032 Chr.14: 92807071-92807071 on Build GRCh38
rs932935197 Chr.14: 92807145-92807146 on Build GRCh38
rs974404678 Chr.14: 92807076-92807076 on Build GRCh38
rs541591410 Chr.14: 92807416-92807416 on Build GRCh38
rs779322105 Chr.14: 92806897-92806897 on Build GRCh38
rs748501039 Chr.14: 92806907-92806907 on Build GRCh38
rs541917826 Chr.14: 92807318-92807322 on Build GRCh38
rs150641129 Chr.14: 92806931-92806931 on Build GRCh38
rs886280726 Chr.14: 92807394-92807394 on Build GRCh38
rs7147752 Chr.14: 92807161-92807161 on Build GRCh38
rs771334225 Chr.14: 92806935-92806935 on Build GRCh38
rs1034875044 Chr.14: 92807014-92807014 on Build GRCh38
rs938787312 Chr.14: 92807267-92807267 on Build GRCh38
rs200911083 Chr.14: 92807320-92807320 on Build GRCh38
rs1048787396 Chr.14: 92807256-92807256 on Build GRCh38
rs914784386 Chr.14: 92807212-92807212 on Build GRCh38
rs1044443621 Chr.14: 92807232-92807232 on Build GRCh38
rs767296247 Chr.14: 92806985-92806985 on Build GRCh38
rs772332709 Chr.14: 92806927-92806927 on Build GRCh38
rs201869490 Chr.14: 92807321-92807321 on Build GRCh38
rs747227189 Chr.14: 92806932-92806932 on Build GRCh38
rs151281964 Chr.14: 92807317-92807321 on Build GRCh38
rs977808877 Chr.14: 92807168-92807168 on Build GRCh38
rs774177457 Chr.14: 92806967-92806967 on Build GRCh38
rs948673491 Chr.14: 92806986-92806986 on Build GRCh38
rs564551705 Chr.14: 92807020-92807020 on Build GRCh38
rs1036131707 Chr.14: 92806893-92806893 on Build GRCh38

View Full Product Details