Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 226735941-226736522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRS1
Entrez Gene ID
3667
Gene Name
insulin receptor substrate 1
Gene Aliases
HIRS-1
UniGene
Hs.471508
Gene Chromosome Location
Chr.2: 226731317-226798790 on Build GRCh38
Transcripts
NM_005544.2

SNP Detail

SNP ID Chromosome Location
rs15625 Chr.2: 226736165-226736165 on Build GRCh38
rs543801247 Chr.2: 226736280-226736280 on Build GRCh38
rs772013181 Chr.2: 226736039-226736039 on Build GRCh38
rs1005977909 Chr.2: 226736232-226736232 on Build GRCh38
rs935131551 Chr.2: 226736009-226736009 on Build GRCh38
rs893435426 Chr.2: 226736088-226736088 on Build GRCh38
rs531329335 Chr.2: 226736481-226736481 on Build GRCh38
rs903941708 Chr.2: 226736222-226736222 on Build GRCh38
rs1026098381 Chr.2: 226736386-226736386 on Build GRCh38
rs994303056 Chr.2: 226736183-226736183 on Build GRCh38
rs1042486900 Chr.2: 226736216-226736216 on Build GRCh38
rs113636756 Chr.2: 226736042-226736043 on Build GRCh38
rs796645133 Chr.2: 226736043-226736043 on Build GRCh38
rs889060636 Chr.2: 226736302-226736302 on Build GRCh38
rs966070999 Chr.2: 226735992-226735992 on Build GRCh38
rs1050334239 Chr.2: 226736136-226736136 on Build GRCh38
rs952198055 Chr.2: 226736074-226736074 on Build GRCh38
rs184730373 Chr.2: 226736400-226736400 on Build GRCh38
rs574119624 Chr.2: 226736305-226736305 on Build GRCh38
rs984978811 Chr.2: 226736468-226736468 on Build GRCh38
rs1023867865 Chr.2: 226736258-226736258 on Build GRCh38
rs1000026810 Chr.2: 226736262-226736262 on Build GRCh38
rs959498338 Chr.2: 226736474-226736474 on Build GRCh38
rs143728323 Chr.2: 226736339-226736339 on Build GRCh38
rs901257699 Chr.2: 226736172-226736172 on Build GRCh38
rs948171811 Chr.2: 226735991-226735991 on Build GRCh38
rs914990227 Chr.2: 226736466-226736466 on Build GRCh38
rs969649885 Chr.2: 226736278-226736278 on Build GRCh38
rs1018762369 Chr.2: 226736160-226736160 on Build GRCh38
rs550454148 Chr.2: 226735967-226735967 on Build GRCh38
rs181715451 Chr.2: 226736489-226736489 on Build GRCh38
rs982292544 Chr.2: 226735993-226735993 on Build GRCh38
rs756998827 Chr.2: 226736021-226736023 on Build GRCh38
rs926762338 Chr.2: 226736473-226736473 on Build GRCh38
rs541739645 Chr.2: 226736332-226736332 on Build GRCh38
rs889050881 Chr.2: 226736152-226736152 on Build GRCh38
rs55924404 Chr.2: 226736433-226736433 on Build GRCh38
rs927677878 Chr.2: 226736007-226736007 on Build GRCh38
rs140076732 Chr.2: 226736211-226736211 on Build GRCh38
rs79783536 Chr.2: 226736155-226736155 on Build GRCh38
rs938223414 Chr.2: 226736200-226736200 on Build GRCh38
rs567702851 Chr.2: 226736490-226736490 on Build GRCh38
rs763629883 Chr.2: 226736340-226736340 on Build GRCh38
rs1056895547 Chr.2: 226736201-226736201 on Build GRCh38
rs562084888 Chr.2: 226736295-226736295 on Build GRCh38
rs916840619 Chr.2: 226735981-226735981 on Build GRCh38
rs917936269 Chr.2: 226736209-226736209 on Build GRCh38
rs778740347 Chr.2: 226736117-226736117 on Build GRCh38
rs918961316 Chr.2: 226736073-226736073 on Build GRCh38
rs117173955 Chr.2: 226736395-226736395 on Build GRCh38
rs965742967 Chr.2: 226736368-226736368 on Build GRCh38
rs1007445568 Chr.2: 226736338-226736338 on Build GRCh38
rs758054175 Chr.2: 226736370-226736370 on Build GRCh38
rs971856913 Chr.2: 226736066-226736066 on Build GRCh38

View Full Product Details