Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42162923-42163514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH4
Entrez Gene ID
23415
Gene Name
potassium voltage-gated channel subfamily H member 4
Gene Aliases
BEC2, ELK1, Kv12.3
UniGene
Hs.304081
Gene Chromosome Location
Chr.17: 42156892-42181278 on Build GRCh38
Transcripts
NM_012285.2 XM_017024400.1 XM_017024401.1 XM_017024402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4694343 c.2579_2580insC p.T861fs*12 Chr.17: 42163232-42163233 on Build GRCh38
5892330 c.2576C>T p.P859L Chr.17: 42163236-42163236 on Build GRCh38
3819615 c.2545C>A p.P849T Chr.17: 42163267-42163267 on Build GRCh38
979429 c.2517G>T p.E839D Chr.17: 42163295-42163295 on Build GRCh38
3517628 c.2577C>T p.P859P Chr.17: 42163235-42163235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015801439 Chr.17: 42163424-42163424 on Build GRCh38
rs778049184 Chr.17: 42163431-42163431 on Build GRCh38
rs1005916591 Chr.17: 42163262-42163262 on Build GRCh38
rs1035579845 Chr.17: 42163405-42163405 on Build GRCh38
rs765507809 Chr.17: 42163375-42163375 on Build GRCh38
rs1041774427 Chr.17: 42163093-42163093 on Build GRCh38
rs752912390 Chr.17: 42163385-42163385 on Build GRCh38
rs185379443 Chr.17: 42163033-42163033 on Build GRCh38
rs190657461 Chr.17: 42163172-42163172 on Build GRCh38
rs769396326 Chr.17: 42163281-42163281 on Build GRCh38
rs1034307637 Chr.17: 42163448-42163448 on Build GRCh38
rs991991897 Chr.17: 42162965-42162965 on Build GRCh38
rs914400908 Chr.17: 42163430-42163430 on Build GRCh38
rs778706807 Chr.17: 42163134-42163134 on Build GRCh38
rs947449963 Chr.17: 42162975-42162975 on Build GRCh38
rs774337935 Chr.17: 42163337-42163337 on Build GRCh38
rs759778362 Chr.17: 42163225-42163225 on Build GRCh38
rs746280863 Chr.17: 42162946-42162946 on Build GRCh38
rs768089250 Chr.17: 42163316-42163316 on Build GRCh38
rs926677561 Chr.17: 42163002-42163002 on Build GRCh38
rs752047908 Chr.17: 42163249-42163249 on Build GRCh38
rs138731053 Chr.17: 42163241-42163241 on Build GRCh38
rs996990187 Chr.17: 42163169-42163169 on Build GRCh38
rs748071039 Chr.17: 42163199-42163199 on Build GRCh38
rs772151776 Chr.17: 42163342-42163342 on Build GRCh38
rs968818158 Chr.17: 42163456-42163456 on Build GRCh38
rs958789850 Chr.17: 42163422-42163422 on Build GRCh38
rs560039729 Chr.17: 42162976-42162976 on Build GRCh38
rs767754924 Chr.17: 42163197-42163198 on Build GRCh38
rs971034201 Chr.17: 42162986-42162986 on Build GRCh38
rs937975937 Chr.17: 42163077-42163077 on Build GRCh38
rs754745192 Chr.17: 42162973-42162973 on Build GRCh38
rs982767435 Chr.17: 42162999-42163000 on Build GRCh38
rs773078891 Chr.17: 42163362-42163362 on Build GRCh38
rs369870132 Chr.17: 42163351-42163351 on Build GRCh38
rs552362566 Chr.17: 42163364-42163364 on Build GRCh38
rs915226885 Chr.17: 42163493-42163493 on Build GRCh38
rs758429962 Chr.17: 42163244-42163244 on Build GRCh38
rs541826826 Chr.17: 42163163-42163163 on Build GRCh38
rs774201863 Chr.17: 42163202-42163202 on Build GRCh38
rs888169269 Chr.17: 42163411-42163411 on Build GRCh38
rs781475897 Chr.17: 42163270-42163270 on Build GRCh38
rs752782421 Chr.17: 42163239-42163239 on Build GRCh38
rs368659322 Chr.17: 42163217-42163217 on Build GRCh38
rs530624604 Chr.17: 42163253-42163253 on Build GRCh38
rs761717664 Chr.17: 42163339-42163339 on Build GRCh38
rs374223230 Chr.17: 42163238-42163238 on Build GRCh38
rs752902107 Chr.17: 42163200-42163201 on Build GRCh38
rs376800803 Chr.17: 42163219-42163219 on Build GRCh38
rs758307226 Chr.17: 42163427-42163427 on Build GRCh38
rs1041559829 Chr.17: 42163204-42163204 on Build GRCh38
rs944294718 Chr.17: 42163147-42163147 on Build GRCh38
rs756221609 Chr.17: 42163268-42163270 on Build GRCh38
rs754101738 Chr.17: 42163223-42163223 on Build GRCh38
rs989408744 Chr.17: 42163488-42163488 on Build GRCh38
rs900011084 Chr.17: 42163165-42163165 on Build GRCh38
rs143978398 Chr.17: 42163401-42163401 on Build GRCh38
rs746235008 Chr.17: 42163272-42163272 on Build GRCh38
rs748805614 Chr.17: 42163297-42163297 on Build GRCh38
rs778322670 Chr.17: 42163248-42163248 on Build GRCh38

View Full Product Details