Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5686714-5687082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD11B1L
Entrez Gene ID
374875
Gene Name
hydroxysteroid 11-beta dehydrogenase 1 like
Gene Aliases
11-DH3, 11-beta-HSD3, HSD3, SCDR10, SCDR10B, SDR26C2
UniGene
Hs.631840
Gene Chromosome Location
Chr.19: 5680765-5688523 on Build GRCh38
Transcripts
NM_001267868.1 NM_001267869.1 NM_001267870.1 NM_001267871.1 NM_198533.2 NM_198704.2 NM_198705.2 NM_198706.2 NM_198707.2 NM_198708.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5741371 c.341A>G p.N114S Chr.19: 5686924-5686924 on Build GRCh38
4806453 c.321G>T p.G107G Chr.19: 5686904-5686904 on Build GRCh38
5960136 c.354delC p.A119fs*19 Chr.19: 5686937-5686937 on Build GRCh38
6431882 c.407A>G p.Q136R Chr.19: 5686990-5686990 on Build GRCh38
5960135 c.354delC p.A119fs*19 Chr.19: 5686937-5686937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894968694 Chr.19: 5687015-5687015 on Build GRCh38
rs761110049 Chr.19: 5687011-5687011 on Build GRCh38
rs746940980 Chr.19: 5686931-5686931 on Build GRCh38
rs553392555 Chr.19: 5686973-5686973 on Build GRCh38
rs991402061 Chr.19: 5686971-5686971 on Build GRCh38
rs1043172981 Chr.19: 5686925-5686925 on Build GRCh38
rs890351528 Chr.19: 5686792-5686792 on Build GRCh38
rs950411552 Chr.19: 5686794-5686794 on Build GRCh38
rs998753607 Chr.19: 5686762-5686762 on Build GRCh38
rs933597598 Chr.19: 5686868-5686868 on Build GRCh38
rs992074378 Chr.19: 5686758-5686758 on Build GRCh38
rs753797634 Chr.19: 5686873-5686873 on Build GRCh38
rs747977283 Chr.19: 5686937-5686937 on Build GRCh38
rs760001903 Chr.19: 5686964-5686964 on Build GRCh38
rs757488459 Chr.19: 5687039-5687039 on Build GRCh38
rs1052016779 Chr.19: 5686901-5686901 on Build GRCh38
rs757372612 Chr.19: 5686892-5686892 on Build GRCh38
rs762308388 Chr.19: 5686954-5686954 on Build GRCh38
rs974860009 Chr.19: 5686865-5686865 on Build GRCh38
rs754078307 Chr.19: 5687037-5687037 on Build GRCh38
rs945993566 Chr.19: 5686914-5686914 on Build GRCh38
rs746809651 Chr.19: 5686910-5686911 on Build GRCh38
rs376953099 Chr.19: 5687031-5687031 on Build GRCh38
rs536367552 Chr.19: 5686807-5686807 on Build GRCh38
rs567485735 Chr.19: 5686746-5686746 on Build GRCh38
rs1042735834 Chr.19: 5686743-5686743 on Build GRCh38
rs1033957328 Chr.19: 5686996-5686996 on Build GRCh38
rs1017398571 Chr.19: 5686864-5686864 on Build GRCh38
rs950394570 Chr.19: 5687038-5687038 on Build GRCh38
rs1013959813 Chr.19: 5687034-5687034 on Build GRCh38
rs573248095 Chr.19: 5687001-5687001 on Build GRCh38
rs983421741 Chr.19: 5686953-5686953 on Build GRCh38
rs774056260 Chr.19: 5686850-5686850 on Build GRCh38
rs759058739 Chr.19: 5686902-5686902 on Build GRCh38
rs775794565 Chr.19: 5686995-5686995 on Build GRCh38
rs778091679 Chr.19: 5686851-5686851 on Build GRCh38
rs770662285 Chr.19: 5686993-5686994 on Build GRCh38
rs768756023 Chr.19: 5686933-5686933 on Build GRCh38
rs768036784 Chr.19: 5686979-5686979 on Build GRCh38
rs908931320 Chr.19: 5686809-5686809 on Build GRCh38
rs539071106 Chr.19: 5687047-5687047 on Build GRCh38
rs765378547 Chr.19: 5687042-5687042 on Build GRCh38
rs769291082 Chr.19: 5686946-5686946 on Build GRCh38
rs764169138 Chr.19: 5687013-5687013 on Build GRCh38
rs942217349 Chr.19: 5686833-5686833 on Build GRCh38
rs916745553 Chr.19: 5687033-5687033 on Build GRCh38
rs1024853768 Chr.19: 5687035-5687035 on Build GRCh38
rs917608177 Chr.19: 5686788-5686788 on Build GRCh38
rs963122358 Chr.19: 5687063-5687063 on Build GRCh38
rs922105568 Chr.19: 5686866-5686866 on Build GRCh38
rs113901360 Chr.19: 5687048-5687048 on Build GRCh38
rs960946441 Chr.19: 5686967-5686967 on Build GRCh38
rs781111075 Chr.19: 5686934-5686934 on Build GRCh38
rs770458522 Chr.19: 5686956-5686956 on Build GRCh38
rs772922135 Chr.19: 5686948-5686948 on Build GRCh38
rs983165081 Chr.19: 5686802-5686802 on Build GRCh38
rs773914890 Chr.19: 5686961-5686961 on Build GRCh38

View Full Product Details