Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 178558531-178559010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNMB2
Entrez Gene ID
10242
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 2
Gene Aliases
-
UniGene
Hs.478368
Gene Chromosome Location
Chr.3: 178536298-178844429 on Build GRCh38
Transcripts
NM_001278911.1 NM_005832.4 NM_181361.2 XM_011512325.2 NR_126560.1 NR_126561.1

Gene Symbol
KCNMB2-AS1
Entrez Gene ID
104797538
Gene Name
KCNMB2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.478363
Gene Chromosome Location
Chr.3: 178525467-178860405 on Build GRCh38
Transcripts
NM_001278911.1 NM_005832.4 NM_181361.2 XM_011512325.2 NR_126560.1 NR_126561.1

SNP Detail

SNP ID Chromosome Location
rs910463561 Chr.3: 178558602-178558602 on Build GRCh38
rs772348122 Chr.3: 178558751-178558751 on Build GRCh38
rs966457147 Chr.3: 178558753-178558753 on Build GRCh38
rs769820675 Chr.3: 178558663-178558663 on Build GRCh38
rs773334618 Chr.3: 178558690-178558690 on Build GRCh38
rs1007319997 Chr.3: 178558814-178558814 on Build GRCh38
rs958163406 Chr.3: 178558606-178558607 on Build GRCh38
rs367952025 Chr.3: 178558672-178558672 on Build GRCh38
rs907264187 Chr.3: 178558646-178558646 on Build GRCh38
rs1020905379 Chr.3: 178558855-178558855 on Build GRCh38
rs4857775 Chr.3: 178558590-178558590 on Build GRCh38
rs957937863 Chr.3: 178558770-178558770 on Build GRCh38
rs936627715 Chr.3: 178558576-178558576 on Build GRCh38
rs754893488 Chr.3: 178558978-178558978 on Build GRCh38
rs776822766 Chr.3: 178558893-178558893 on Build GRCh38
rs78543989 Chr.3: 178558782-178558782 on Build GRCh38
rs775648052 Chr.3: 178558755-178558755 on Build GRCh38
rs181057146 Chr.3: 178558614-178558614 on Build GRCh38
rs954652160 Chr.3: 178558796-178558796 on Build GRCh38
rs918403742 Chr.3: 178558891-178558891 on Build GRCh38
rs900679271 Chr.3: 178558720-178558720 on Build GRCh38
rs371449259 Chr.3: 178558738-178558738 on Build GRCh38
rs997644605 Chr.3: 178558731-178558731 on Build GRCh38
rs368645371 Chr.3: 178558975-178558975 on Build GRCh38
rs116735258 Chr.3: 178558954-178558954 on Build GRCh38
rs764399937 Chr.3: 178558890-178558890 on Build GRCh38
rs747985294 Chr.3: 178558647-178558647 on Build GRCh38
rs762207667 Chr.3: 178558965-178558965 on Build GRCh38
rs563199045 Chr.3: 178558775-178558775 on Build GRCh38
rs1037259840 Chr.3: 178558763-178558763 on Build GRCh38
rs1017818007 Chr.3: 178558826-178558826 on Build GRCh38
rs373651737 Chr.3: 178558821-178558821 on Build GRCh38
rs929431138 Chr.3: 178558896-178558896 on Build GRCh38
rs989380538 Chr.3: 178558840-178558840 on Build GRCh38
rs1045796870 Chr.3: 178558617-178558617 on Build GRCh38
rs368669413 Chr.3: 178558847-178558850 on Build GRCh38
rs188953666 Chr.3: 178558603-178558603 on Build GRCh38
rs541150074 Chr.3: 178558613-178558613 on Build GRCh38
rs559158634 Chr.3: 178558839-178558839 on Build GRCh38
rs887756671 Chr.3: 178558673-178558673 on Build GRCh38
rs1048276259 Chr.3: 178558912-178558912 on Build GRCh38
rs909249558 Chr.3: 178558951-178558951 on Build GRCh38
rs1039304712 Chr.3: 178558957-178558957 on Build GRCh38
rs957908520 Chr.3: 178558830-178558830 on Build GRCh38
rs966695860 Chr.3: 178558884-178558884 on Build GRCh38
rs184046158 Chr.3: 178558790-178558790 on Build GRCh38
rs1051965083 Chr.3: 178558977-178558977 on Build GRCh38
rs986409922 Chr.3: 178558591-178558591 on Build GRCh38
rs981884511 Chr.3: 178558861-178558861 on Build GRCh38
rs1017863138 Chr.3: 178558702-178558702 on Build GRCh38
rs867765727 Chr.3: 178558668-178558668 on Build GRCh38
rs982142008 Chr.3: 178558895-178558895 on Build GRCh38
rs929312857 Chr.3: 178558664-178558664 on Build GRCh38
rs1030067542 Chr.3: 178558579-178558579 on Build GRCh38
rs990729992 Chr.3: 178558772-178558772 on Build GRCh38
rs999321119 Chr.3: 178558985-178558985 on Build GRCh38
rs978229645 Chr.3: 178558754-178558754 on Build GRCh38
rs868400447 Chr.3: 178558679-178558679 on Build GRCh38

View Full Product Details