Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35440586-35441185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0319L
Entrez Gene ID
79932
Gene Name
KIAA0319 like
Gene Aliases
AAVR
UniGene
Hs.456507
Gene Chromosome Location
Chr.1: 35433490-35557670 on Build GRCh38
Transcripts
NM_024874.4 XM_006710907.1 XM_006710909.1 XM_006710910.1 XM_011542179.2 XM_011542180.1 XM_017002366.1 XM_017002367.1 XM_017002368.1 XM_017002369.1 XM_017002370.1 XM_017002371.1 XM_017002372.1 XM_017002374.1 XM_017002375.1 XM_017002376.1 XM_017002377.1 XM_017002378.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4165172 c.2871-3T>G p.? Chr.1: 35441141-35441141 on Build GRCh38
6283398 c.2913G>A p.L971L Chr.1: 35441096-35441096 on Build GRCh38
4165173 c.2871-3T>G p.? Chr.1: 35441141-35441141 on Build GRCh38
6693159 c.2921C>T p.T974M Chr.1: 35441088-35441088 on Build GRCh38
4842911 c.2956C>T p.R986* Chr.1: 35441053-35441053 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188489395 Chr.1: 35440608-35440608 on Build GRCh38
rs1021886088 Chr.1: 35440844-35440844 on Build GRCh38
rs1026827445 Chr.1: 35441029-35441029 on Build GRCh38
rs536581169 Chr.1: 35440943-35440943 on Build GRCh38
rs757710365 Chr.1: 35441048-35441048 on Build GRCh38
rs761507231 Chr.1: 35441118-35441118 on Build GRCh38
rs1038367329 Chr.1: 35440753-35440753 on Build GRCh38
rs374372036 Chr.1: 35441010-35441010 on Build GRCh38
rs551202764 Chr.1: 35440765-35440765 on Build GRCh38
rs756712402 Chr.1: 35441040-35441040 on Build GRCh38
rs992616463 Chr.1: 35440831-35440831 on Build GRCh38
rs150125031 Chr.1: 35441124-35441124 on Build GRCh38
rs948219487 Chr.1: 35440849-35440849 on Build GRCh38
rs777390125 Chr.1: 35440832-35440832 on Build GRCh38
rs775379755 Chr.1: 35441000-35441000 on Build GRCh38
rs892227495 Chr.1: 35441126-35441126 on Build GRCh38
rs963934130 Chr.1: 35440752-35440752 on Build GRCh38
rs545357266 Chr.1: 35440851-35440851 on Build GRCh38
rs760643239 Chr.1: 35441004-35441004 on Build GRCh38
rs554743586 Chr.1: 35440709-35440709 on Build GRCh38
rs1000795977 Chr.1: 35440649-35440649 on Build GRCh38
rs202079417 Chr.1: 35441052-35441052 on Build GRCh38
rs776419422 Chr.1: 35441101-35441101 on Build GRCh38
rs975997555 Chr.1: 35440626-35440627 on Build GRCh38
rs908627449 Chr.1: 35440689-35440689 on Build GRCh38
rs1056181051 Chr.1: 35440829-35440829 on Build GRCh38
rs185565136 Chr.1: 35440816-35440816 on Build GRCh38
rs952610085 Chr.1: 35440777-35440777 on Build GRCh38
rs904128930 Chr.1: 35440635-35440635 on Build GRCh38
rs774414365 Chr.1: 35441121-35441121 on Build GRCh38
rs936921202 Chr.1: 35441161-35441161 on Build GRCh38
rs752840271 Chr.1: 35441152-35441152 on Build GRCh38
rs996643556 Chr.1: 35441011-35441011 on Build GRCh38
rs745369609 Chr.1: 35440969-35440969 on Build GRCh38
rs974088404 Chr.1: 35440961-35440961 on Build GRCh38
rs767028989 Chr.1: 35440615-35440615 on Build GRCh38
rs896693114 Chr.1: 35440610-35440610 on Build GRCh38
rs746897102 Chr.1: 35441088-35441088 on Build GRCh38
rs992919413 Chr.1: 35441106-35441106 on Build GRCh38
rs1026875380 Chr.1: 35440766-35440766 on Build GRCh38
rs192928652 Chr.1: 35440795-35440795 on Build GRCh38
rs763833992 Chr.1: 35441006-35441006 on Build GRCh38
rs952600329 Chr.1: 35441044-35441044 on Build GRCh38
rs550118286 Chr.1: 35441156-35441156 on Build GRCh38
rs1030961071 Chr.1: 35440684-35440684 on Build GRCh38
rs918067999 Chr.1: 35440838-35440838 on Build GRCh38
rs771632902 Chr.1: 35440981-35440981 on Build GRCh38
rs982738131 Chr.1: 35441086-35441086 on Build GRCh38
rs755095859 Chr.1: 35441033-35441033 on Build GRCh38
rs982411335 Chr.1: 35440788-35440788 on Build GRCh38
rs1015717074 Chr.1: 35440902-35440902 on Build GRCh38
rs767746210 Chr.1: 35441035-35441035 on Build GRCh38
rs771535112 Chr.1: 35441064-35441064 on Build GRCh38
rs572081247 Chr.1: 35440698-35440698 on Build GRCh38
rs527714013 Chr.1: 35440923-35440923 on Build GRCh38
rs1045321667 Chr.1: 35440631-35440631 on Build GRCh38
rs766513650 Chr.1: 35441020-35441020 on Build GRCh38
rs755784142 Chr.1: 35440890-35440890 on Build GRCh38
rs567749039 Chr.1: 35440850-35440850 on Build GRCh38
rs751875020 Chr.1: 35441025-35441025 on Build GRCh38
rs531012233 Chr.1: 35440738-35440738 on Build GRCh38
rs149759448 Chr.1: 35440927-35440927 on Build GRCh38
rs778144519 Chr.1: 35441041-35441041 on Build GRCh38
rs745588176 Chr.1: 35441053-35441053 on Build GRCh38
rs141395302 Chr.1: 35441087-35441087 on Build GRCh38
rs919898437 Chr.1: 35440997-35440997 on Build GRCh38
rs769687335 Chr.1: 35441120-35441120 on Build GRCh38
rs903554630 Chr.1: 35441158-35441158 on Build GRCh38
rs77453248 Chr.1: 35440883-35440883 on Build GRCh38
rs570953456 Chr.1: 35440706-35440706 on Build GRCh38
rs753216479 Chr.1: 35441038-35441038 on Build GRCh38
rs368261954 Chr.1: 35440665-35440665 on Build GRCh38
rs137877511 Chr.1: 35441090-35441090 on Build GRCh38
rs776309406 Chr.1: 35441008-35441008 on Build GRCh38
rs980869552 Chr.1: 35441155-35441155 on Build GRCh38
rs759869733 Chr.1: 35441123-35441123 on Build GRCh38

View Full Product Details